Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 21496654-21497158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC10
Entrez Gene ID
399726
Gene Name
cancer susceptibility candidate 10
Gene Aliases
C10orf114, bA418C1.3
UniGene
Hs.315568
Gene Chromosome Location
Chr.10: 21494492-21497284 on Build GRCh38
Transcripts
NM_001010911.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1297160 c.20C>T p.S7L Chr.10: 21496795-21496795 on Build GRCh38
916993 c.55C>T p.R19C Chr.10: 21496760-21496760 on Build GRCh38
4013299 c.14A>T p.E5V Chr.10: 21496801-21496801 on Build GRCh38
5777410 c.11G>A p.R4Q Chr.10: 21496804-21496804 on Build GRCh38
2132749 c.5A>G p.Q2R Chr.10: 21496810-21496810 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761303769 Chr.10: 21496744-21496744 on Build GRCh38
rs775318635 Chr.10: 21496724-21496724 on Build GRCh38
rs201321677 Chr.10: 21496720-21496720 on Build GRCh38
rs940522592 Chr.10: 21497095-21497095 on Build GRCh38
rs531467183 Chr.10: 21496706-21496706 on Build GRCh38
rs765550018 Chr.10: 21496765-21496765 on Build GRCh38
rs907604470 Chr.10: 21496896-21496896 on Build GRCh38
rs779733651 Chr.10: 21496786-21496786 on Build GRCh38
rs763031223 Chr.10: 21496677-21496677 on Build GRCh38
rs904132059 Chr.10: 21496970-21496970 on Build GRCh38
rs536129636 Chr.10: 21496964-21496964 on Build GRCh38
rs764373352 Chr.10: 21496776-21496776 on Build GRCh38
rs952025778 Chr.10: 21497090-21497090 on Build GRCh38
rs892534932 Chr.10: 21496983-21496983 on Build GRCh38
rs925492143 Chr.10: 21497052-21497052 on Build GRCh38
rs940485692 Chr.10: 21496911-21496911 on Build GRCh38
rs769736766 Chr.10: 21496722-21496722 on Build GRCh38
rs1048229017 Chr.10: 21496903-21496903 on Build GRCh38
rs1001054791 Chr.10: 21496694-21496694 on Build GRCh38
rs370701694 Chr.10: 21496741-21496741 on Build GRCh38
rs754320709 Chr.10: 21496748-21496748 on Build GRCh38
rs200193158 Chr.10: 21496700-21496700 on Build GRCh38
rs984666067 Chr.10: 21496857-21496857 on Build GRCh38
rs758607267 Chr.10: 21496772-21496772 on Build GRCh38
rs138941395 Chr.10: 21496947-21496947 on Build GRCh38
rs570851799 Chr.10: 21496874-21496874 on Build GRCh38
rs532377641 Chr.10: 21496686-21496686 on Build GRCh38
rs753049977 Chr.10: 21496768-21496768 on Build GRCh38
rs75185404 Chr.10: 21497103-21497103 on Build GRCh38
rs760050892 Chr.10: 21496850-21496850 on Build GRCh38
rs975431436 Chr.10: 21497056-21497056 on Build GRCh38
rs896736123 Chr.10: 21496682-21496682 on Build GRCh38
rs554280451 Chr.10: 21496854-21496854 on Build GRCh38
rs771591995 Chr.10: 21496844-21496844 on Build GRCh38
rs754267787 Chr.10: 21496683-21496683 on Build GRCh38
rs144565194 Chr.10: 21496789-21496789 on Build GRCh38
rs138673157 Chr.10: 21496779-21496779 on Build GRCh38
rs901669552 Chr.10: 21497099-21497099 on Build GRCh38
rs772668077 Chr.10: 21496849-21496849 on Build GRCh38
rs756830249 Chr.10: 21496807-21496808 on Build GRCh38
rs371609504 Chr.10: 21496709-21496709 on Build GRCh38
rs985021932 Chr.10: 21497092-21497092 on Build GRCh38
rs777915107 Chr.10: 21496707-21496707 on Build GRCh38
rs554610170 Chr.10: 21497043-21497043 on Build GRCh38
rs1040166183 Chr.10: 21497091-21497091 on Build GRCh38
rs1045259817 Chr.10: 21496968-21496968 on Build GRCh38
rs779723726 Chr.10: 21496804-21496804 on Build GRCh38
rs551951274 Chr.10: 21496692-21496692 on Build GRCh38
rs149294945 Chr.10: 21496780-21496780 on Build GRCh38
rs574335458 Chr.10: 21497117-21497117 on Build GRCh38
rs781248537 Chr.10: 21496717-21496717 on Build GRCh38
rs774023534 Chr.10: 21496838-21496838 on Build GRCh38
rs200550375 Chr.10: 21496737-21496737 on Build GRCh38
rs886919500 Chr.10: 21496935-21496935 on Build GRCh38
rs1008349710 Chr.10: 21497012-21497012 on Build GRCh38
rs367593647 Chr.10: 21496835-21496835 on Build GRCh38
rs1028898469 Chr.10: 21497059-21497059 on Build GRCh38
rs749092808 Chr.10: 21496816-21496816 on Build GRCh38
rs762514735 Chr.10: 21496738-21496738 on Build GRCh38
rs756046910 Chr.10: 21496791-21496791 on Build GRCh38
rs907741642 Chr.10: 21497094-21497094 on Build GRCh38
rs368305390 Chr.10: 21496707-21496707 on Build GRCh38
rs964012729 Chr.10: 21497047-21497047 on Build GRCh38
rs1031195789 Chr.10: 21496980-21496980 on Build GRCh38
rs540049715 Chr.10: 21496892-21496892 on Build GRCh38
rs757417538 Chr.10: 21496859-21496859 on Build GRCh38
rs968447062 Chr.10: 21497031-21497031 on Build GRCh38
rs758581959 Chr.10: 21496702-21496702 on Build GRCh38
rs879520353 Chr.10: 21496972-21496972 on Build GRCh38
rs566401036 Chr.10: 21496684-21496684 on Build GRCh38
rs768293231 Chr.10: 21496739-21496739 on Build GRCh38
rs991835444 Chr.10: 21497096-21497096 on Build GRCh38
rs987191017 Chr.10: 21496736-21496736 on Build GRCh38
rs879791160 Chr.10: 21496955-21496955 on Build GRCh38
rs917677585 Chr.10: 21497114-21497114 on Build GRCh38
rs747705762 Chr.10: 21496839-21496839 on Build GRCh38
rs1045100842 Chr.10: 21497124-21497124 on Build GRCh38
rs766911776 Chr.10: 21496745-21496745 on Build GRCh38
rs759985474 Chr.10: 21496758-21496758 on Build GRCh38
rs556783340 Chr.10: 21496920-21496920 on Build GRCh38

View Full Product Details