Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26751475-26752023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI1
Entrez Gene ID
10006
Gene Name
abl interactor 1
Gene Aliases
ABI-1, ABLBP4, E3B1, NAP1BP, SSH3BP, SSH3BP1
UniGene
Hs.508148
Gene Chromosome Location
Chr.10: 26746596-26861087 on Build GRCh38
Transcripts
NM_001012750.2 NM_001012751.2 NM_001012752.2 NM_001178116.1 NM_001178119.1 NM_001178120.1 NM_001178121.1 NM_001178122.1 NM_001178123.1 NM_001178124.1 NM_001178125.1 NM_005470.3 XM_005252332.3 XM_005252333.3 XM_006717359.2 XM_006717360.2 XM_006717361.2 XM_017015458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4013524 c.1252C>T p.P418S Chr.10: 26751697-26751697 on Build GRCh38
7085775 c.1242A>C p.P414P Chr.10: 26751707-26751707 on Build GRCh38
4462158 c.1235C>T p.P412L Chr.10: 26751714-26751714 on Build GRCh38
6293592 c.1188G>A p.P396P Chr.10: 26751761-26751761 on Build GRCh38
1721131 c.1252_1254delCCA p.P418delP Chr.10: 26751695-26751697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs193123094 Chr.10: 26751740-26751740 on Build GRCh38
rs748928252 Chr.10: 26751599-26751599 on Build GRCh38
rs777267993 Chr.10: 26751799-26751799 on Build GRCh38
rs761333410 Chr.10: 26751591-26751592 on Build GRCh38
rs572768904 Chr.10: 26751707-26751707 on Build GRCh38
rs992824481 Chr.10: 26751517-26751517 on Build GRCh38
rs139017043 Chr.10: 26751538-26751538 on Build GRCh38
rs200148529 Chr.10: 26751736-26751736 on Build GRCh38
rs199540578 Chr.10: 26751686-26751686 on Build GRCh38
rs567613849 Chr.10: 26751986-26751986 on Build GRCh38
rs758873983 Chr.10: 26751762-26751762 on Build GRCh38
rs998847568 Chr.10: 26751858-26751858 on Build GRCh38
rs759386547 Chr.10: 26751642-26751642 on Build GRCh38
rs916152285 Chr.10: 26751575-26751575 on Build GRCh38
rs943198230 Chr.10: 26751739-26751739 on Build GRCh38
rs1006336671 Chr.10: 26751885-26751885 on Build GRCh38
rs910470720 Chr.10: 26751647-26751647 on Build GRCh38
rs375017858 Chr.10: 26751565-26751565 on Build GRCh38
rs765661298 Chr.10: 26751739-26751740 on Build GRCh38
rs200697468 Chr.10: 26751703-26751703 on Build GRCh38
rs760722965 Chr.10: 26751728-26751728 on Build GRCh38
rs529754521 Chr.10: 26751941-26751941 on Build GRCh38
rs557421551 Chr.10: 26751507-26751507 on Build GRCh38
rs750066446 Chr.10: 26751753-26751753 on Build GRCh38
rs201504652 Chr.10: 26751702-26751702 on Build GRCh38
rs770491097 Chr.10: 26751608-26751608 on Build GRCh38
rs771996944 Chr.10: 26751654-26751654 on Build GRCh38
rs377604542 Chr.10: 26751796-26751796 on Build GRCh38
rs530616563 Chr.10: 26751818-26751818 on Build GRCh38
rs901811694 Chr.10: 26751725-26751725 on Build GRCh38
rs753897727 Chr.10: 26751734-26751734 on Build GRCh38
rs574061991 Chr.10: 26751537-26751537 on Build GRCh38
rs377364870 Chr.10: 26751592-26751592 on Build GRCh38
rs753243560 Chr.10: 26751529-26751529 on Build GRCh38
rs368655056 Chr.10: 26751869-26751870 on Build GRCh38
rs758958587 Chr.10: 26751571-26751571 on Build GRCh38
rs767819355 Chr.10: 26751520-26751520 on Build GRCh38
rs370908388 Chr.10: 26751705-26751705 on Build GRCh38
rs895313217 Chr.10: 26751917-26751917 on Build GRCh38
rs760815058 Chr.10: 26751547-26751547 on Build GRCh38
rs764270194 Chr.10: 26751552-26751552 on Build GRCh38
rs767223620 Chr.10: 26751672-26751673 on Build GRCh38
rs957439431 Chr.10: 26751934-26751934 on Build GRCh38
rs778500074 Chr.10: 26751829-26751829 on Build GRCh38
rs903223042 Chr.10: 26751848-26751849 on Build GRCh38
rs560996442 Chr.10: 26751916-26751916 on Build GRCh38
rs764185239 Chr.10: 26751732-26751732 on Build GRCh38
rs901767927 Chr.10: 26751748-26751748 on Build GRCh38
rs374177554 Chr.10: 26751894-26751894 on Build GRCh38
rs373351238 Chr.10: 26751589-26751589 on Build GRCh38
rs756296985 Chr.10: 26751807-26751807 on Build GRCh38
rs2242232 Chr.10: 26751583-26751583 on Build GRCh38
rs12766451 Chr.10: 26751768-26751768 on Build GRCh38
rs546545284 Chr.10: 26751961-26751961 on Build GRCh38
rs757445491 Chr.10: 26751554-26751554 on Build GRCh38
rs780683919 Chr.10: 26751765-26751765 on Build GRCh38
rs774141875 Chr.10: 26751609-26751609 on Build GRCh38
rs201804543 Chr.10: 26751695-26751697 on Build GRCh38
rs1023105019 Chr.10: 26751997-26751997 on Build GRCh38
rs755374808 Chr.10: 26751694-26751695 on Build GRCh38
rs779298350 Chr.10: 26751559-26751559 on Build GRCh38

View Full Product Details