Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26763802-26764315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI1
Entrez Gene ID
10006
Gene Name
abl interactor 1
Gene Aliases
ABI-1, ABLBP4, E3B1, NAP1BP, SSH3BP, SSH3BP1
UniGene
Hs.508148
Gene Chromosome Location
Chr.10: 26746596-26861087 on Build GRCh38
Transcripts
NM_001012750.2 NM_001012751.2 NM_001012752.2 NM_001178116.1 NM_001178119.1 NM_001178120.1 NM_001178121.1 NM_001178122.1 NM_001178123.1 NM_001178124.1 NM_001178125.1 NM_005470.3 XM_005252332.3 XM_005252333.3 XM_006717359.2 XM_006717360.2 XM_006717361.2 XM_011519289.2 XM_017015458.1 XM_017015459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6846484 c.901+1G>A p.? Chr.10: 26763879-26763879 on Build GRCh38
427421 c.895G>T p.V299L Chr.10: 26763886-26763886 on Build GRCh38
5733796 c.854C>T p.P285L Chr.10: 26763927-26763927 on Build GRCh38
4832966 c.830C>G p.S277C Chr.10: 26763951-26763951 on Build GRCh38
5789459 c.898A>C p.I300L Chr.10: 26763883-26763883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936427665 Chr.10: 26763911-26763911 on Build GRCh38
rs201631906 Chr.10: 26763873-26763873 on Build GRCh38
rs1014497441 Chr.10: 26764121-26764121 on Build GRCh38
rs758537942 Chr.10: 26763917-26763917 on Build GRCh38
rs943947396 Chr.10: 26764038-26764038 on Build GRCh38
rs994614438 Chr.10: 26764048-26764048 on Build GRCh38
rs559735069 Chr.10: 26763950-26763950 on Build GRCh38
rs770294372 Chr.10: 26763947-26763947 on Build GRCh38
rs924957959 Chr.10: 26764157-26764157 on Build GRCh38
rs966402088 Chr.10: 26764154-26764154 on Build GRCh38
rs746139789 Chr.10: 26763884-26763887 on Build GRCh38
rs764104766 Chr.10: 26763899-26763899 on Build GRCh38
rs759149814 Chr.10: 26763970-26763970 on Build GRCh38
rs1018908592 Chr.10: 26764289-26764289 on Build GRCh38
rs775265512 Chr.10: 26763980-26763980 on Build GRCh38
rs752428017 Chr.10: 26763886-26763886 on Build GRCh38
rs114533437 Chr.10: 26764107-26764107 on Build GRCh38
rs1041388368 Chr.10: 26764044-26764044 on Build GRCh38
rs1025852746 Chr.10: 26764202-26764202 on Build GRCh38
rs139609364 Chr.10: 26763901-26763901 on Build GRCh38
rs895158364 Chr.10: 26764050-26764050 on Build GRCh38
rs977691119 Chr.10: 26764155-26764155 on Build GRCh38
rs960506321 Chr.10: 26764120-26764120 on Build GRCh38
rs781477425 Chr.10: 26763842-26763846 on Build GRCh38
rs1001630436 Chr.10: 26764091-26764091 on Build GRCh38
rs985195175 Chr.10: 26764242-26764242 on Build GRCh38
rs957756089 Chr.10: 26764230-26764230 on Build GRCh38
rs780392579 Chr.10: 26763920-26763920 on Build GRCh38
rs532628894 Chr.10: 26764037-26764037 on Build GRCh38
rs759048348 Chr.10: 26763869-26763869 on Build GRCh38
rs149749581 Chr.10: 26763943-26763943 on Build GRCh38
rs771186900 Chr.10: 26764062-26764062 on Build GRCh38
rs1020992282 Chr.10: 26764150-26764150 on Build GRCh38
rs560326444 Chr.10: 26764034-26764034 on Build GRCh38
rs925075724 Chr.10: 26763907-26763907 on Build GRCh38
rs768958948 Chr.10: 26763931-26763931 on Build GRCh38
rs904351367 Chr.10: 26763978-26763978 on Build GRCh38
rs745974378 Chr.10: 26763913-26763913 on Build GRCh38
rs780557681 Chr.10: 26764115-26764115 on Build GRCh38
rs546254922 Chr.10: 26764046-26764046 on Build GRCh38
rs757296325 Chr.10: 26763906-26763906 on Build GRCh38
rs190033938 Chr.10: 26764082-26764082 on Build GRCh38
rs777010816 Chr.10: 26763936-26763936 on Build GRCh38
rs984363257 Chr.10: 26763838-26763838 on Build GRCh38
rs531693242 Chr.10: 26764088-26764088 on Build GRCh38
rs193032056 Chr.10: 26763849-26763849 on Build GRCh38
rs769698777 Chr.10: 26763935-26763937 on Build GRCh38
rs201595826 Chr.10: 26763834-26763834 on Build GRCh38
rs747350366 Chr.10: 26763925-26763925 on Build GRCh38
rs778792155 Chr.10: 26763910-26763910 on Build GRCh38
rs984374530 Chr.10: 26764243-26764243 on Build GRCh38
rs760429761 Chr.10: 26763990-26763990 on Build GRCh38
rs540396727 Chr.10: 26763975-26763975 on Build GRCh38
rs756016243 Chr.10: 26763896-26763896 on Build GRCh38
rs910927799 Chr.10: 26763853-26763853 on Build GRCh38
rs535356059 Chr.10: 26764128-26764128 on Build GRCh38

View Full Product Details