Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27005021-27005570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26
Entrez Gene ID
22852
Gene Name
ankyrin repeat domain 26
Gene Aliases
THC2, bA145E8.1
UniGene
Hs.361041
Gene Chromosome Location
Chr.10: 26938195-27100498 on Build GRCh38
Transcripts
NM_001256053.1 NM_014915.2 XM_006717423.2 XM_006717425.3 XM_011519416.2 XM_017015928.1 XM_017015929.1 XM_017015930.1 XM_017015931.1 XM_017015932.1 XM_017015933.1

SNP Detail

SNP ID Chromosome Location
rs997938415 Chr.10: 27005062-27005062 on Build GRCh38
rs1038836345 Chr.10: 27005269-27005269 on Build GRCh38
rs760890734 Chr.10: 27005108-27005108 on Build GRCh38
rs1016324395 Chr.10: 27005131-27005131 on Build GRCh38
rs574433218 Chr.10: 27005206-27005206 on Build GRCh38
rs539837222 Chr.10: 27005501-27005501 on Build GRCh38
rs773128620 Chr.10: 27005265-27005265 on Build GRCh38
rs768854633 Chr.10: 27005229-27005229 on Build GRCh38
rs144522802 Chr.10: 27005216-27005216 on Build GRCh38
rs892846998 Chr.10: 27005267-27005267 on Build GRCh38
rs949650676 Chr.10: 27005219-27005219 on Build GRCh38
rs1014942327 Chr.10: 27005522-27005523 on Build GRCh38
rs988411257 Chr.10: 27005502-27005502 on Build GRCh38
rs928298711 Chr.10: 27005253-27005253 on Build GRCh38
rs990982357 Chr.10: 27005193-27005193 on Build GRCh38
rs971423724 Chr.10: 27005448-27005448 on Build GRCh38
rs1023539101 Chr.10: 27005060-27005060 on Build GRCh38
rs548846121 Chr.10: 27005184-27005184 on Build GRCh38
rs3215884 Chr.10: 27005521-27005522 on Build GRCh38
rs570729257 Chr.10: 27005439-27005439 on Build GRCh38
rs766391764 Chr.10: 27005339-27005339 on Build GRCh38
rs886447888 Chr.10: 27005338-27005338 on Build GRCh38
rs1010564125 Chr.10: 27005422-27005422 on Build GRCh38
rs759479576 Chr.10: 27005489-27005489 on Build GRCh38
rs997424197 Chr.10: 27005322-27005322 on Build GRCh38
rs767758965 Chr.10: 27005545-27005545 on Build GRCh38
rs543630204 Chr.10: 27005528-27005528 on Build GRCh38
rs191989707 Chr.10: 27005316-27005316 on Build GRCh38
rs1023788815 Chr.10: 27005440-27005440 on Build GRCh38
rs765804997 Chr.10: 27005144-27005149 on Build GRCh38
rs900393350 Chr.10: 27005053-27005053 on Build GRCh38
rs78425651 Chr.10: 27005394-27005394 on Build GRCh38
rs1052182505 Chr.10: 27005375-27005375 on Build GRCh38
rs935534519 Chr.10: 27005353-27005354 on Build GRCh38
rs184207908 Chr.10: 27005410-27005410 on Build GRCh38
rs924159595 Chr.10: 27005500-27005500 on Build GRCh38
rs531917417 Chr.10: 27005146-27005146 on Build GRCh38
rs983465637 Chr.10: 27005182-27005182 on Build GRCh38
rs1003423446 Chr.10: 27005482-27005482 on Build GRCh38
rs1030324440 Chr.10: 27005100-27005100 on Build GRCh38
rs1004848805 Chr.10: 27005341-27005341 on Build GRCh38
rs969362704 Chr.10: 27005075-27005075 on Build GRCh38
rs200102083 Chr.10: 27005521-27005521 on Build GRCh38
rs375997697 Chr.10: 27005268-27005268 on Build GRCh38
rs1030371894 Chr.10: 27005330-27005330 on Build GRCh38
rs934443442 Chr.10: 27005264-27005264 on Build GRCh38
rs377417678 Chr.10: 27005420-27005420 on Build GRCh38
rs1052462705 Chr.10: 27005340-27005340 on Build GRCh38
rs904876392 Chr.10: 27005458-27005458 on Build GRCh38
rs916832892 Chr.10: 27005218-27005218 on Build GRCh38
rs886046941 Chr.10: 27005060-27005060 on Build GRCh38
rs886046942 Chr.10: 27005448-27005448 on Build GRCh38
rs200859644 Chr.10: 27005522-27005522 on Build GRCh38
rs950817085 Chr.10: 27005107-27005107 on Build GRCh38

View Full Product Details