Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 31809096-31809619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP12
Entrez Gene ID
94134
Gene Name
Rho GTPase activating protein 12
Gene Aliases
-
UniGene
Hs.499264
Gene Chromosome Location
Chr.10: 31805398-31928876 on Build GRCh38
Transcripts
NM_001270695.1 NM_001270696.1 NM_001270697.1 NM_001270698.1 NM_001270699.1 NM_018287.6 XM_005252644.1 XM_011519761.1 XM_017016954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4388563 c.2133G>T p.E711D Chr.10: 31809124-31809124 on Build GRCh38
3358738 c.2144T>G p.L715W Chr.10: 31809113-31809113 on Build GRCh38
7214980 c.2051-3T>C p.? Chr.10: 31809310-31809310 on Build GRCh38
917737 c.2155A>C p.K719Q Chr.10: 31809102-31809102 on Build GRCh38
1238209 c.2080A>T p.S694C Chr.10: 31809278-31809278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777194557 Chr.10: 31809285-31809285 on Build GRCh38
rs141159821 Chr.10: 31809301-31809301 on Build GRCh38
rs377180449 Chr.10: 31809287-31809287 on Build GRCh38
rs780320103 Chr.10: 31809307-31809307 on Build GRCh38
rs992000532 Chr.10: 31809488-31809488 on Build GRCh38
rs779166850 Chr.10: 31809205-31809205 on Build GRCh38
rs374781140 Chr.10: 31809327-31809327 on Build GRCh38
rs376707593 Chr.10: 31809189-31809189 on Build GRCh38
rs769482377 Chr.10: 31809142-31809142 on Build GRCh38
rs778010821 Chr.10: 31809327-31809328 on Build GRCh38
rs201598945 Chr.10: 31809200-31809200 on Build GRCh38
rs754118008 Chr.10: 31809201-31809201 on Build GRCh38
rs529859627 Chr.10: 31809273-31809273 on Build GRCh38
rs200457588 Chr.10: 31809222-31809222 on Build GRCh38
rs750758339 Chr.10: 31809230-31809230 on Build GRCh38
rs760623977 Chr.10: 31809342-31809342 on Build GRCh38
rs533175767 Chr.10: 31809326-31809327 on Build GRCh38
rs775459793 Chr.10: 31809344-31809346 on Build GRCh38
rs1022440681 Chr.10: 31809364-31809364 on Build GRCh38
rs779424434 Chr.10: 31809156-31809157 on Build GRCh38
rs751734948 Chr.10: 31809406-31809410 on Build GRCh38
rs569666038 Chr.10: 31809511-31809511 on Build GRCh38
rs1047939694 Chr.10: 31809433-31809433 on Build GRCh38
rs957697673 Chr.10: 31809544-31809544 on Build GRCh38
rs777036598 Chr.10: 31809341-31809342 on Build GRCh38
rs916507840 Chr.10: 31809491-31809491 on Build GRCh38
rs1033748133 Chr.10: 31809539-31809539 on Build GRCh38
rs140324216 Chr.10: 31809235-31809235 on Build GRCh38
rs747624562 Chr.10: 31809139-31809139 on Build GRCh38
rs1024759288 Chr.10: 31809489-31809489 on Build GRCh38
rs771962757 Chr.10: 31809331-31809331 on Build GRCh38
rs764282807 Chr.10: 31809344-31809344 on Build GRCh38
rs886639414 Chr.10: 31809160-31809160 on Build GRCh38
rs977841548 Chr.10: 31809454-31809454 on Build GRCh38
rs769342736 Chr.10: 31809586-31809586 on Build GRCh38
rs112808486 Chr.10: 31809315-31809315 on Build GRCh38
rs751898208 Chr.10: 31809267-31809267 on Build GRCh38
rs2275808 Chr.10: 31809187-31809187 on Build GRCh38
rs374495749 Chr.10: 31809199-31809199 on Build GRCh38
rs766088064 Chr.10: 31809176-31809176 on Build GRCh38
rs995869946 Chr.10: 31809312-31809312 on Build GRCh38
rs771270671 Chr.10: 31809329-31809329 on Build GRCh38
rs752875373 Chr.10: 31809197-31809197 on Build GRCh38
rs906272421 Chr.10: 31809529-31809529 on Build GRCh38
rs762799437 Chr.10: 31809168-31809168 on Build GRCh38
rs989413312 Chr.10: 31809590-31809590 on Build GRCh38
rs532771211 Chr.10: 31809143-31809143 on Build GRCh38
rs573378187 Chr.10: 31809233-31809233 on Build GRCh38
rs754716419 Chr.10: 31809355-31809355 on Build GRCh38
rs774992457 Chr.10: 31809593-31809593 on Build GRCh38
rs886628495 Chr.10: 31809472-31809475 on Build GRCh38
rs1019209760 Chr.10: 31809291-31809291 on Build GRCh38
rs781091787 Chr.10: 31809367-31809367 on Build GRCh38
rs775565198 Chr.10: 31809338-31809338 on Build GRCh38
rs964697181 Chr.10: 31809303-31809303 on Build GRCh38
rs780586597 Chr.10: 31809249-31809249 on Build GRCh38
rs1001898995 Chr.10: 31809532-31809532 on Build GRCh38
rs749356151 Chr.10: 31809456-31809456 on Build GRCh38
rs761433212 Chr.10: 31809163-31809167 on Build GRCh38
rs772120424 Chr.10: 31809214-31809215 on Build GRCh38
rs770581460 Chr.10: 31809298-31809298 on Build GRCh38
rs368668457 Chr.10: 31809190-31809190 on Build GRCh38
rs570236859 Chr.10: 31809372-31809372 on Build GRCh38
rs1030785578 Chr.10: 31809476-31809476 on Build GRCh38
rs774056928 Chr.10: 31809310-31809310 on Build GRCh38
rs149135755 Chr.10: 31809481-31809481 on Build GRCh38

View Full Product Details