Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32919739-32920259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB1
Entrez Gene ID
3688
Gene Name
integrin subunit beta 1
Gene Aliases
CD29, FNRB, GPIIA, MDF2, MSK12, VLA-BETA, VLAB
UniGene
Hs.643813
Gene Chromosome Location
Chr.10: 32900318-32958365 on Build GRCh38
Transcripts
NM_002211.3 NM_033668.2 NM_133376.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4786438 c.1289T>C p.I430T Chr.10: 32920065-32920065 on Build GRCh38
4013879 c.1461C>T p.G487G Chr.10: 32919893-32919893 on Build GRCh38
5077903 c.1313delA p.K438fs*17 Chr.10: 32920041-32920041 on Build GRCh38
4013882 c.1352C>T p.T451M Chr.10: 32920002-32920002 on Build GRCh38
4802141 c.1286G>T p.S429I Chr.10: 32920068-32920068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047793723 Chr.10: 32920189-32920189 on Build GRCh38
rs571657526 Chr.10: 32919782-32919782 on Build GRCh38
rs775874993 Chr.10: 32920066-32920066 on Build GRCh38
rs754935937 Chr.10: 32919949-32919949 on Build GRCh38
rs201228879 Chr.10: 32920002-32920002 on Build GRCh38
rs543305169 Chr.10: 32919993-32919993 on Build GRCh38
rs201867204 Chr.10: 32919861-32919861 on Build GRCh38
rs775002495 Chr.10: 32919846-32919846 on Build GRCh38
rs747344725 Chr.10: 32920219-32920219 on Build GRCh38
rs200740357 Chr.10: 32920133-32920133 on Build GRCh38
rs751579836 Chr.10: 32920086-32920087 on Build GRCh38
rs778171708 Chr.10: 32919995-32919995 on Build GRCh38
rs754080821 Chr.10: 32920111-32920111 on Build GRCh38
rs189199433 Chr.10: 32919763-32919763 on Build GRCh38
rs765443724 Chr.10: 32919851-32919852 on Build GRCh38
rs1009966763 Chr.10: 32920172-32920172 on Build GRCh38
rs45580338 Chr.10: 32920102-32920102 on Build GRCh38
rs796452248 Chr.10: 32920160-32920160 on Build GRCh38
rs963889011 Chr.10: 32920224-32920224 on Build GRCh38
rs199980084 Chr.10: 32919996-32919996 on Build GRCh38
rs556334352 Chr.10: 32920093-32920094 on Build GRCh38
rs749312525 Chr.10: 32920001-32920001 on Build GRCh38
rs759740155 Chr.10: 32919849-32919850 on Build GRCh38
rs752504227 Chr.10: 32919854-32919854 on Build GRCh38
rs1009137692 Chr.10: 32919891-32919891 on Build GRCh38
rs760981561 Chr.10: 32920067-32920067 on Build GRCh38
rs755977363 Chr.10: 32920108-32920108 on Build GRCh38
rs375565127 Chr.10: 32920091-32920091 on Build GRCh38
rs529782489 Chr.10: 32920114-32920114 on Build GRCh38
rs372581400 Chr.10: 32919808-32919808 on Build GRCh38
rs199714737 Chr.10: 32920230-32920230 on Build GRCh38
rs1057335969 Chr.10: 32919784-32919784 on Build GRCh38
rs947611698 Chr.10: 32919761-32919761 on Build GRCh38
rs3780876 Chr.10: 32919792-32919792 on Build GRCh38
rs761506383 Chr.10: 32919876-32919876 on Build GRCh38
rs748314340 Chr.10: 32920181-32920181 on Build GRCh38
rs180672316 Chr.10: 32919950-32919950 on Build GRCh38
rs143061769 Chr.10: 32919890-32919890 on Build GRCh38
rs974531843 Chr.10: 32919988-32919988 on Build GRCh38
rs749899655 Chr.10: 32919892-32919892 on Build GRCh38
rs756354856 Chr.10: 32919991-32919991 on Build GRCh38
rs775869254 Chr.10: 32920210-32920210 on Build GRCh38
rs201957851 Chr.10: 32920115-32920115 on Build GRCh38
rs11557912 Chr.10: 32920068-32920068 on Build GRCh38
rs200632770 Chr.10: 32920040-32920040 on Build GRCh38
rs377263167 Chr.10: 32919904-32919904 on Build GRCh38
rs201585457 Chr.10: 32920200-32920200 on Build GRCh38
rs752496276 Chr.10: 32920105-32920105 on Build GRCh38
rs201973516 Chr.10: 32919893-32919893 on Build GRCh38
rs759990495 Chr.10: 32920176-32920176 on Build GRCh38
rs147320657 Chr.10: 32920016-32920016 on Build GRCh38
rs201328160 Chr.10: 32920090-32920090 on Build GRCh38
rs1018646625 Chr.10: 32920201-32920201 on Build GRCh38
rs1008839055 Chr.10: 32920202-32920202 on Build GRCh38
rs111522124 Chr.10: 32919962-32919962 on Build GRCh38
rs765754224 Chr.10: 32920184-32920184 on Build GRCh38
rs764807466 Chr.10: 32920072-32920072 on Build GRCh38
rs764045193 Chr.10: 32920085-32920086 on Build GRCh38
rs771524178 Chr.10: 32919840-32919840 on Build GRCh38
rs567846688 Chr.10: 32920186-32920186 on Build GRCh38
rs1026236432 Chr.10: 32919849-32919849 on Build GRCh38
rs202125974 Chr.10: 32919856-32919856 on Build GRCh38
rs572473980 Chr.10: 32919814-32919814 on Build GRCh38
rs955335980 Chr.10: 32919831-32919837 on Build GRCh38
rs768997768 Chr.10: 32920220-32920220 on Build GRCh38
rs948857459 Chr.10: 32919798-32919798 on Build GRCh38
rs151175196 Chr.10: 32919929-32919929 on Build GRCh38
rs746356744 Chr.10: 32920018-32920018 on Build GRCh38
rs745956908 Chr.10: 32920196-32920196 on Build GRCh38
rs202197186 Chr.10: 32920084-32920084 on Build GRCh38
rs540994419 Chr.10: 32920094-32920094 on Build GRCh38
rs199630291 Chr.10: 32919848-32919848 on Build GRCh38
rs1044764505 Chr.10: 32919799-32919799 on Build GRCh38
rs927567254 Chr.10: 32920104-32920104 on Build GRCh38
rs201353503 Chr.10: 32919968-32919968 on Build GRCh38
rs758178134 Chr.10: 32919870-32919871 on Build GRCh38
rs753045262 Chr.10: 32919868-32919868 on Build GRCh38
rs754917875 Chr.10: 32920097-32920097 on Build GRCh38
rs994721165 Chr.10: 32919823-32919823 on Build GRCh38
rs140045688 Chr.10: 32920019-32920019 on Build GRCh38
rs11009146 Chr.10: 32920079-32920079 on Build GRCh38
rs199513073 Chr.10: 32919875-32919875 on Build GRCh38
rs1010081226 Chr.10: 32919806-32919806 on Build GRCh38
rs556640329 Chr.10: 32919764-32919764 on Build GRCh38
rs934828855 Chr.10: 32919766-32919766 on Build GRCh38

View Full Product Details