Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 37151928-37152458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD30A
Entrez Gene ID
91074
Gene Name
ankyrin repeat domain 30A
Gene Aliases
NY-BR-1
UniGene
Hs.373787
Gene Chromosome Location
Chr.10: 37125754-37235831 on Build GRCh38
Transcripts
NM_052997.2 XM_011519757.2 XM_011519758.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5515329 c.1495G>T p.E499* Chr.10: 37152077-37152077 on Build GRCh38
917958 c.1509G>T p.K503N Chr.10: 37152091-37152091 on Build GRCh38
917957 c.1486T>C p.F496L Chr.10: 37152068-37152068 on Build GRCh38
4467314 c.1480C>T p.P494S Chr.10: 37152062-37152062 on Build GRCh38
6129225 c.1478A>G p.D493G Chr.10: 37152060-37152060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766677311 Chr.10: 37152136-37152136 on Build GRCh38
rs113854383 Chr.10: 37152401-37152401 on Build GRCh38
rs902810623 Chr.10: 37152242-37152242 on Build GRCh38
rs767239968 Chr.10: 37152074-37152074 on Build GRCh38
rs367660636 Chr.10: 37151974-37151974 on Build GRCh38
rs780270117 Chr.10: 37152103-37152103 on Build GRCh38
rs185425251 Chr.10: 37152431-37152431 on Build GRCh38
rs770335333 Chr.10: 37152127-37152127 on Build GRCh38
rs190082898 Chr.10: 37152168-37152168 on Build GRCh38
rs773419215 Chr.10: 37152128-37152128 on Build GRCh38
rs377410013 Chr.10: 37152066-37152066 on Build GRCh38
rs914295789 Chr.10: 37152216-37152216 on Build GRCh38
rs1148257 Chr.10: 37152144-37152144 on Build GRCh38
rs887856512 Chr.10: 37152319-37152319 on Build GRCh38
rs758153783 Chr.10: 37152160-37152160 on Build GRCh38
rs374037740 Chr.10: 37152110-37152110 on Build GRCh38
rs17605939 Chr.10: 37152212-37152212 on Build GRCh38
rs879019577 Chr.10: 37152111-37152111 on Build GRCh38
rs17590141 Chr.10: 37152226-37152226 on Build GRCh38
rs760765417 Chr.10: 37152050-37152050 on Build GRCh38
rs749263602 Chr.10: 37152009-37152009 on Build GRCh38
rs753444984 Chr.10: 37152147-37152147 on Build GRCh38
rs17605918 Chr.10: 37152026-37152026 on Build GRCh38
rs772934791 Chr.10: 37152400-37152400 on Build GRCh38
rs17605897 Chr.10: 37151966-37151966 on Build GRCh38
rs17605946 Chr.10: 37152218-37152218 on Build GRCh38
rs747225637 Chr.10: 37152166-37152166 on Build GRCh38
rs1029107527 Chr.10: 37152298-37152298 on Build GRCh38
rs558014233 Chr.10: 37152138-37152139 on Build GRCh38
rs17590155 Chr.10: 37152250-37152250 on Build GRCh38
rs17605961 Chr.10: 37152296-37152296 on Build GRCh38
rs774295493 Chr.10: 37152014-37152014 on Build GRCh38
rs940562287 Chr.10: 37151993-37151993 on Build GRCh38
rs17605925 Chr.10: 37152145-37152145 on Build GRCh38
rs868558109 Chr.10: 37152428-37152428 on Build GRCh38
rs771790844 Chr.10: 37152102-37152102 on Build GRCh38
rs17590120 Chr.10: 37152038-37152038 on Build GRCh38
rs17590127 Chr.10: 37152105-37152105 on Build GRCh38
rs373632677 Chr.10: 37152149-37152149 on Build GRCh38
rs147869221 Chr.10: 37152138-37152138 on Build GRCh38
rs559880936 Chr.10: 37152310-37152310 on Build GRCh38
rs984742815 Chr.10: 37151979-37151979 on Build GRCh38
rs981343886 Chr.10: 37152369-37152369 on Build GRCh38
rs17590169 Chr.10: 37152416-37152416 on Build GRCh38
rs937115641 Chr.10: 37152035-37152035 on Build GRCh38
rs752323974 Chr.10: 37152076-37152076 on Build GRCh38
rs17605982 Chr.10: 37152415-37152415 on Build GRCh38
rs778866106 Chr.10: 37152151-37152151 on Build GRCh38
rs763337065 Chr.10: 37152132-37152132 on Build GRCh38
rs11011050 Chr.10: 37152272-37152272 on Build GRCh38
rs370765794 Chr.10: 37152095-37152095 on Build GRCh38
rs777955611 Chr.10: 37152239-37152239 on Build GRCh38
rs779717572 Chr.10: 37152163-37152163 on Build GRCh38
rs907778818 Chr.10: 37151986-37151986 on Build GRCh38
rs981290224 Chr.10: 37152197-37152197 on Build GRCh38
rs879237341 Chr.10: 37152246-37152246 on Build GRCh38
rs143203945 Chr.10: 37152240-37152240 on Build GRCh38
rs752340658 Chr.10: 37152140-37152140 on Build GRCh38
rs928485613 Chr.10: 37152205-37152205 on Build GRCh38
rs541948867 Chr.10: 37152175-37152175 on Build GRCh38
rs1036000296 Chr.10: 37152385-37152385 on Build GRCh38
rs17605911 Chr.10: 37152022-37152022 on Build GRCh38
rs17590162 Chr.10: 37152287-37152287 on Build GRCh38
rs764180219 Chr.10: 37152054-37152054 on Build GRCh38
rs17605968 Chr.10: 37152396-37152396 on Build GRCh38
rs558043459 Chr.10: 37152077-37152077 on Build GRCh38
rs267602482 Chr.10: 37152081-37152081 on Build GRCh38
rs921962572 Chr.10: 37151973-37151973 on Build GRCh38
rs17605975 Chr.10: 37152402-37152402 on Build GRCh38
rs902756773 Chr.10: 37152094-37152094 on Build GRCh38
rs17590148 Chr.10: 37152249-37152249 on Build GRCh38
rs1014886700 Chr.10: 37152321-37152321 on Build GRCh38
rs757334629 Chr.10: 37152092-37152092 on Build GRCh38
rs768656363 Chr.10: 37152116-37152116 on Build GRCh38
rs562256802 Chr.10: 37152190-37152190 on Build GRCh38
rs538041220 Chr.10: 37152064-37152064 on Build GRCh38
rs776756864 Chr.10: 37152121-37152121 on Build GRCh38
rs45590233 Chr.10: 37152004-37152004 on Build GRCh38
rs535182700 Chr.10: 37152008-37152008 on Build GRCh38
rs770794534 Chr.10: 37152012-37152012 on Build GRCh38
rs753764850 Chr.10: 37152087-37152087 on Build GRCh38
rs17590176 Chr.10: 37152432-37152432 on Build GRCh38
rs777064189 Chr.10: 37152055-37152055 on Build GRCh38
rs879174134 Chr.10: 37152108-37152108 on Build GRCh38
rs985183233 Chr.10: 37152174-37152174 on Build GRCh38
rs996729800 Chr.10: 37152276-37152276 on Build GRCh38
rs750345537 Chr.10: 37152152-37152152 on Build GRCh38
rs1006223799 Chr.10: 37152320-37152320 on Build GRCh38
rs17590113 Chr.10: 37152027-37152027 on Build GRCh38
rs17605932 Chr.10: 37152206-37152206 on Build GRCh38
rs760373559 Chr.10: 37152139-37152139 on Build GRCh38
rs17605953 Chr.10: 37152289-37152289 on Build GRCh38
rs45515098 Chr.10: 37152063-37152063 on Build GRCh38
rs928433310 Chr.10: 37152028-37152028 on Build GRCh38
rs17605904 Chr.10: 37152011-37152011 on Build GRCh38
rs757368299 Chr.10: 37152086-37152086 on Build GRCh38
rs548753348 Chr.10: 37152429-37152429 on Build GRCh38
rs760413320 Chr.10: 37152142-37152142 on Build GRCh38
rs1040618839 Chr.10: 37152262-37152262 on Build GRCh38
rs775397098 Chr.10: 37152031-37152031 on Build GRCh38
rs180949067 Chr.10: 37152196-37152196 on Build GRCh38
rs781062734 Chr.10: 37152125-37152125 on Build GRCh38
rs961990991 Chr.10: 37152346-37152346 on Build GRCh38

View Full Product Details