Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 44370070-44370566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL12
Entrez Gene ID
6387
Gene Name
C-X-C motif chemokine ligand 12
Gene Aliases
IRH, PBSF, SCYB12, SDF1, TLSF, TPAR1
UniGene
Hs.522891
Gene Chromosome Location
Chr.10: 44292088-44385097 on Build GRCh38
Transcripts
NM_000609.6 NM_001277990.1

SNP Detail

SNP ID Chromosome Location
rs138232619 Chr.10: 44370322-44370322 on Build GRCh38
rs974888999 Chr.10: 44370193-44370193 on Build GRCh38
rs553118430 Chr.10: 44370474-44370474 on Build GRCh38
rs978164872 Chr.10: 44370411-44370411 on Build GRCh38
rs936460021 Chr.10: 44370421-44370421 on Build GRCh38
rs917563618 Chr.10: 44370107-44370107 on Build GRCh38
rs1804430 Chr.10: 44370291-44370291 on Build GRCh38
rs923923221 Chr.10: 44370286-44370286 on Build GRCh38
rs962861136 Chr.10: 44370320-44370320 on Build GRCh38
rs1006898299 Chr.10: 44370346-44370346 on Build GRCh38
rs921758190 Chr.10: 44370438-44370438 on Build GRCh38
rs964846912 Chr.10: 44370121-44370121 on Build GRCh38
rs547197609 Chr.10: 44370479-44370479 on Build GRCh38
rs1051592807 Chr.10: 44370485-44370485 on Build GRCh38
rs562214696 Chr.10: 44370117-44370117 on Build GRCh38
rs79160520 Chr.10: 44370483-44370483 on Build GRCh38
rs758265186 Chr.10: 44370273-44370273 on Build GRCh38
rs113379535 Chr.10: 44370518-44370518 on Build GRCh38
rs555451028 Chr.10: 44370366-44370366 on Build GRCh38
rs751054162 Chr.10: 44370293-44370293 on Build GRCh38
rs1065297 Chr.10: 44370528-44370528 on Build GRCh38
rs933383554 Chr.10: 44370262-44370262 on Build GRCh38
rs974941371 Chr.10: 44370372-44370372 on Build GRCh38
rs1033873524 Chr.10: 44370327-44370327 on Build GRCh38
rs959823497 Chr.10: 44370338-44370338 on Build GRCh38
rs752599395 Chr.10: 44370221-44370221 on Build GRCh38
rs1038898248 Chr.10: 44370281-44370281 on Build GRCh38
rs755831068 Chr.10: 44370507-44370507 on Build GRCh38
rs966365706 Chr.10: 44370392-44370392 on Build GRCh38
rs530830766 Chr.10: 44370484-44370484 on Build GRCh38
rs1804429 Chr.10: 44370393-44370393 on Build GRCh38
rs755035212 Chr.10: 44370435-44370437 on Build GRCh38
rs978536813 Chr.10: 44370267-44370267 on Build GRCh38
rs74930726 Chr.10: 44370294-44370294 on Build GRCh38
rs566554462 Chr.10: 44370288-44370288 on Build GRCh38
rs766555469 Chr.10: 44370411-44370412 on Build GRCh38
rs964944992 Chr.10: 44370369-44370369 on Build GRCh38
rs573610322 Chr.10: 44370382-44370382 on Build GRCh38
rs7777 Chr.10: 44370289-44370289 on Build GRCh38
rs906328437 Chr.10: 44370297-44370297 on Build GRCh38
rs10347 Chr.10: 44370304-44370304 on Build GRCh38
rs567621626 Chr.10: 44370107-44370107 on Build GRCh38
rs903645963 Chr.10: 44370237-44370237 on Build GRCh38
rs181570691 Chr.10: 44370387-44370387 on Build GRCh38
rs888086602 Chr.10: 44370344-44370344 on Build GRCh38
rs1037021714 Chr.10: 44370526-44370526 on Build GRCh38
rs558065974 Chr.10: 44370300-44370300 on Build GRCh38
rs992381101 Chr.10: 44370342-44370342 on Build GRCh38
rs184589176 Chr.10: 44370496-44370496 on Build GRCh38
rs868724307 Chr.10: 44370396-44370396 on Build GRCh38
rs535647460 Chr.10: 44370118-44370118 on Build GRCh38

View Full Product Details