Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 71749094-71749602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf54
Entrez Gene ID
64115
Gene Name
chromosome 10 open reading frame 54
Gene Aliases
B7-H5, B7H5, DD1alpha, GI24, PD-1H, PP2135, SISP1, VISTA
UniGene
Hs.47382
Gene Chromosome Location
Chr.10: 71747556-71773580 on Build GRCh38
Transcripts
NM_022153.1 NM_022124.5 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_011540052.2 XM_017016499.1 XM_017016500.1 XM_017016501.1 XM_017016502.1 XM_017016503.1 XM_017016504.1 XM_017016505.1

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
UniGene
Hs.656032
Gene Chromosome Location
Chr.10: 71396934-71815947 on Build GRCh38
Transcripts
NM_022153.1 NM_022124.5 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_011540052.2 XM_017016499.1 XM_017016500.1 XM_017016501.1 XM_017016502.1 XM_017016503.1 XM_017016504.1 XM_017016505.1

SNP Detail

SNP ID Chromosome Location
rs987023821 Chr.10: 71749196-71749196 on Build GRCh38
rs1033335198 Chr.10: 71749234-71749234 on Build GRCh38
rs1016022557 Chr.10: 71749136-71749136 on Build GRCh38
rs924663745 Chr.10: 71749298-71749301 on Build GRCh38
rs910544841 Chr.10: 71749343-71749343 on Build GRCh38
rs567113078 Chr.10: 71749571-71749571 on Build GRCh38
rs996316204 Chr.10: 71749456-71749456 on Build GRCh38
rs917424100 Chr.10: 71749259-71749259 on Build GRCh38
rs1005924323 Chr.10: 71749409-71749409 on Build GRCh38
rs930329387 Chr.10: 71749370-71749370 on Build GRCh38
rs370866549 Chr.10: 71749209-71749209 on Build GRCh38
rs925426694 Chr.10: 71749244-71749244 on Build GRCh38
rs977513787 Chr.10: 71749295-71749295 on Build GRCh38
rs971572898 Chr.10: 71749520-71749520 on Build GRCh38
rs1005867367 Chr.10: 71749130-71749130 on Build GRCh38
rs900273908 Chr.10: 71749453-71749453 on Build GRCh38
rs753929513 Chr.10: 71749506-71749506 on Build GRCh38
rs548463885 Chr.10: 71749568-71749568 on Build GRCh38
rs577240197 Chr.10: 71749184-71749184 on Build GRCh38
rs544891344 Chr.10: 71749190-71749190 on Build GRCh38
rs896296944 Chr.10: 71749288-71749288 on Build GRCh38
rs959068972 Chr.10: 71749472-71749472 on Build GRCh38
rs975646622 Chr.10: 71749171-71749171 on Build GRCh38
rs139069800 Chr.10: 71749518-71749518 on Build GRCh38
rs575636974 Chr.10: 71749239-71749239 on Build GRCh38
rs542937626 Chr.10: 71749310-71749310 on Build GRCh38
rs775737474 Chr.10: 71749218-71749218 on Build GRCh38
rs971520504 Chr.10: 71749270-71749270 on Build GRCh38
rs955555073 Chr.10: 71749489-71749489 on Build GRCh38
rs886318605 Chr.10: 71749364-71749364 on Build GRCh38
rs540177041 Chr.10: 71749461-71749461 on Build GRCh38
rs991325330 Chr.10: 71749562-71749562 on Build GRCh38
rs1033798745 Chr.10: 71749469-71749469 on Build GRCh38
rs1054893151 Chr.10: 71749330-71749330 on Build GRCh38
rs34038038 Chr.10: 71749431-71749432 on Build GRCh38
rs112822200 Chr.10: 71749191-71749191 on Build GRCh38
rs368455934 Chr.10: 71749499-71749499 on Build GRCh38
rs143089953 Chr.10: 71749537-71749537 on Build GRCh38
rs1047696810 Chr.10: 71749362-71749362 on Build GRCh38
rs530373639 Chr.10: 71749567-71749567 on Build GRCh38
rs78063283 Chr.10: 71749556-71749556 on Build GRCh38
rs12771006 Chr.10: 71749444-71749444 on Build GRCh38
rs938094020 Chr.10: 71749247-71749247 on Build GRCh38
rs866591595 Chr.10: 71749207-71749207 on Build GRCh38
rs558792886 Chr.10: 71749132-71749132 on Build GRCh38
rs991749668 Chr.10: 71749245-71749245 on Build GRCh38
rs774057721 Chr.10: 71749139-71749139 on Build GRCh38
rs57734913 Chr.10: 71749122-71749122 on Build GRCh38
rs1030050680 Chr.10: 71749477-71749477 on Build GRCh38
rs190393650 Chr.10: 71749560-71749560 on Build GRCh38
rs993211823 Chr.10: 71749150-71749159 on Build GRCh38
rs979498657 Chr.10: 71749538-71749538 on Build GRCh38
rs1054992705 Chr.10: 71749264-71749264 on Build GRCh38
rs186774890 Chr.10: 71749344-71749344 on Build GRCh38

View Full Product Details