Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 71751566-71752066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf54
Entrez Gene ID
64115
Gene Name
chromosome 10 open reading frame 54
Gene Aliases
B7-H5, B7H5, DD1alpha, GI24, PD-1H, PP2135, SISP1, VISTA
UniGene
Hs.47382
Gene Chromosome Location
Chr.10: 71747556-71773580 on Build GRCh38
Transcripts
NM_022153.1 NM_022124.5 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_011540052.2 XM_017016499.1 XM_017016500.1 XM_017016501.1 XM_017016502.1 XM_017016503.1 XM_017016504.1 XM_017016505.1

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
UniGene
Hs.656032
Gene Chromosome Location
Chr.10: 71396934-71815947 on Build GRCh38
Transcripts
NM_022153.1 NM_022124.5 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_011540052.2 XM_017016499.1 XM_017016500.1 XM_017016501.1 XM_017016502.1 XM_017016503.1 XM_017016504.1 XM_017016505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6551040 c.736G>A p.E246K Chr.10: 71751830-71751830 on Build GRCh38
243266 c.766G>A p.E256K Chr.10: 71751800-71751800 on Build GRCh38
5371083 c.815C>T p.S272F Chr.10: 71751751-71751751 on Build GRCh38
4386598 c.748C>A p.P250T Chr.10: 71751818-71751818 on Build GRCh38
6129969 c.765C>T p.P255P Chr.10: 71751801-71751801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145779213 Chr.10: 71751761-71751761 on Build GRCh38
rs774690287 Chr.10: 71751707-71751707 on Build GRCh38
rs751329363 Chr.10: 71751840-71751840 on Build GRCh38
rs377080999 Chr.10: 71751600-71751600 on Build GRCh38
rs759786059 Chr.10: 71751796-71751796 on Build GRCh38
rs567964395 Chr.10: 71751991-71751991 on Build GRCh38
rs775558013 Chr.10: 71751676-71751676 on Build GRCh38
rs948558353 Chr.10: 71752046-71752046 on Build GRCh38
rs141431398 Chr.10: 71751850-71751850 on Build GRCh38
rs117530736 Chr.10: 71752016-71752016 on Build GRCh38
rs765834365 Chr.10: 71751709-71751709 on Build GRCh38
rs148949180 Chr.10: 71751821-71751821 on Build GRCh38
rs767882153 Chr.10: 71751800-71751800 on Build GRCh38
rs763117232 Chr.10: 71751695-71751695 on Build GRCh38
rs748907481 Chr.10: 71751648-71751648 on Build GRCh38
rs112574903 Chr.10: 71751894-71751894 on Build GRCh38
rs773223258 Chr.10: 71751772-71751772 on Build GRCh38
rs201880754 Chr.10: 71751837-71751837 on Build GRCh38
rs200047559 Chr.10: 71751857-71751857 on Build GRCh38
rs139821326 Chr.10: 71751717-71751717 on Build GRCh38
rs908056977 Chr.10: 71751672-71751672 on Build GRCh38
rs903240052 Chr.10: 71751715-71751715 on Build GRCh38
rs777258542 Chr.10: 71751645-71751645 on Build GRCh38
rs773315626 Chr.10: 71751690-71751690 on Build GRCh38
rs754934975 Chr.10: 71751845-71751845 on Build GRCh38
rs200507926 Chr.10: 71751692-71751692 on Build GRCh38
rs143240262 Chr.10: 71751727-71751727 on Build GRCh38
rs908095514 Chr.10: 71752001-71752001 on Build GRCh38
rs775856574 Chr.10: 71751884-71751884 on Build GRCh38
rs778993914 Chr.10: 71751859-71751859 on Build GRCh38
rs771125955 Chr.10: 71751780-71751780 on Build GRCh38
rs761012326 Chr.10: 71751900-71751900 on Build GRCh38
rs565800501 Chr.10: 71752020-71752020 on Build GRCh38
rs535064782 Chr.10: 71751608-71751608 on Build GRCh38
rs375739009 Chr.10: 71751740-71751740 on Build GRCh38
rs766588848 Chr.10: 71751702-71751702 on Build GRCh38
rs924738618 Chr.10: 71751961-71751961 on Build GRCh38
rs151170391 Chr.10: 71751847-71751847 on Build GRCh38
rs369048655 Chr.10: 71751906-71751906 on Build GRCh38
rs139710615 Chr.10: 71751673-71751673 on Build GRCh38
rs769960085 Chr.10: 71751687-71751687 on Build GRCh38
rs892314630 Chr.10: 71751822-71751822 on Build GRCh38
rs987695520 Chr.10: 71751954-71751954 on Build GRCh38
rs191188213 Chr.10: 71751613-71751613 on Build GRCh38
rs1022113557 Chr.10: 71751854-71751854 on Build GRCh38
rs142813365 Chr.10: 71751836-71751836 on Build GRCh38
rs745662633 Chr.10: 71751661-71751661 on Build GRCh38
rs761185707 Chr.10: 71751803-71751803 on Build GRCh38
rs746051994 Chr.10: 71751872-71751872 on Build GRCh38
rs775738494 Chr.10: 71751801-71751801 on Build GRCh38
rs551980223 Chr.10: 71751903-71751903 on Build GRCh38
rs974008674 Chr.10: 71752029-71752029 on Build GRCh38
rs555787002 Chr.10: 71751599-71751599 on Build GRCh38
rs751983965 Chr.10: 71751985-71751985 on Build GRCh38
rs754813770 Chr.10: 71752034-71752034 on Build GRCh38
rs374701065 Chr.10: 71751905-71751905 on Build GRCh38
rs1002500893 Chr.10: 71751782-71751782 on Build GRCh38
rs750143709 Chr.10: 71751823-71751823 on Build GRCh38
rs540037467 Chr.10: 71751658-71751658 on Build GRCh38
rs74649893 Chr.10: 71751901-71751901 on Build GRCh38
rs552980068 Chr.10: 71752023-71752023 on Build GRCh38
rs760000245 Chr.10: 71751711-71751711 on Build GRCh38
rs751948153 Chr.10: 71751619-71751619 on Build GRCh38
rs779383749 Chr.10: 71752041-71752041 on Build GRCh38
rs746929027 Chr.10: 71751760-71751760 on Build GRCh38
rs756992203 Chr.10: 71751652-71751652 on Build GRCh38
rs1040437072 Chr.10: 71751709-71751709 on Build GRCh38
rs372452777 Chr.10: 71751739-71751739 on Build GRCh38
rs755407596 Chr.10: 71751623-71751623 on Build GRCh38
rs199673713 Chr.10: 71751726-71751726 on Build GRCh38
rs767886762 Chr.10: 71751714-71751714 on Build GRCh38
rs772195812 Chr.10: 71751879-71751879 on Build GRCh38
rs535451458 Chr.10: 71751994-71751994 on Build GRCh38
rs140530381 Chr.10: 71751911-71751911 on Build GRCh38
rs753412246 Chr.10: 71751725-71751725 on Build GRCh38
rs961111498 Chr.10: 71751807-71751807 on Build GRCh38
rs762570652 Chr.10: 71751716-71751716 on Build GRCh38
rs183458408 Chr.10: 71751640-71751640 on Build GRCh38
rs762830091 Chr.10: 71751641-71751641 on Build GRCh38
rs774602402 Chr.10: 71751781-71751781 on Build GRCh38
rs531541026 Chr.10: 71751921-71751921 on Build GRCh38
rs748156820 Chr.10: 71751686-71751686 on Build GRCh38
rs190774933 Chr.10: 71751957-71751957 on Build GRCh38
rs763610736 Chr.10: 71751721-71751721 on Build GRCh38
rs752718137 Chr.10: 71751710-71751710 on Build GRCh38
rs745646326 Chr.10: 71751732-71751732 on Build GRCh38
rs779853858 Chr.10: 71751838-71751838 on Build GRCh38
rs764702023 Chr.10: 71751818-71751818 on Build GRCh38
rs749432318 Chr.10: 71751777-71751777 on Build GRCh38
rs761467389 Chr.10: 71751819-71751819 on Build GRCh38
rs766939172 Chr.10: 71751908-71751908 on Build GRCh38

View Full Product Details