Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 71760786-71761291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf54
Entrez Gene ID
64115
Gene Name
chromosome 10 open reading frame 54
Gene Aliases
B7-H5, B7H5, DD1alpha, GI24, PD-1H, PP2135, SISP1, VISTA
UniGene
Hs.47382
Gene Chromosome Location
Chr.10: 71747556-71773580 on Build GRCh38
Transcripts
NM_022153.1 NM_022124.5 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_011540052.2 XM_017016499.1 XM_017016500.1 XM_017016501.1 XM_017016502.1 XM_017016503.1 XM_017016504.1 XM_017016505.1

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
UniGene
Hs.656032
Gene Chromosome Location
Chr.10: 71396934-71815947 on Build GRCh38
Transcripts
NM_022153.1 NM_022124.5 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_011540052.2 XM_017016499.1 XM_017016500.1 XM_017016501.1 XM_017016502.1 XM_017016503.1 XM_017016504.1 XM_017016505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
224972 c.545C>T p.P182L Chr.10: 71760891-71760891 on Build GRCh38
3751944 c.561T>G p.D187E Chr.10: 71760875-71760875 on Build GRCh38
6435975 c.541T>C p.Y181H Chr.10: 71760895-71760895 on Build GRCh38
4651812 c.542A>G p.Y181C Chr.10: 71760894-71760894 on Build GRCh38
73881 c.520G>C p.A174P Chr.10: 71760916-71760916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376240469 Chr.10: 71760831-71760831 on Build GRCh38
rs896651315 Chr.10: 71761050-71761050 on Build GRCh38
rs535425747 Chr.10: 71761021-71761021 on Build GRCh38
rs371897554 Chr.10: 71760932-71760932 on Build GRCh38
rs544899889 Chr.10: 71761190-71761190 on Build GRCh38
rs558697325 Chr.10: 71761217-71761217 on Build GRCh38
rs3747868 Chr.10: 71761071-71761071 on Build GRCh38
rs774386938 Chr.10: 71760934-71760934 on Build GRCh38
rs554664968 Chr.10: 71761122-71761122 on Build GRCh38
rs747038589 Chr.10: 71760946-71760946 on Build GRCh38
rs747801389 Chr.10: 71760915-71760915 on Build GRCh38
rs776961306 Chr.10: 71760836-71760836 on Build GRCh38
rs200269030 Chr.10: 71760928-71760928 on Build GRCh38
rs930986659 Chr.10: 71760940-71760940 on Build GRCh38
rs774057568 Chr.10: 71761205-71761205 on Build GRCh38
rs764197826 Chr.10: 71760832-71760833 on Build GRCh38
rs11557291 Chr.10: 71760925-71760925 on Build GRCh38
rs143030978 Chr.10: 71760892-71760892 on Build GRCh38
rs993455074 Chr.10: 71761213-71761213 on Build GRCh38
rs572586662 Chr.10: 71761142-71761142 on Build GRCh38
rs867185788 Chr.10: 71760891-71760891 on Build GRCh38
rs752499363 Chr.10: 71761086-71761086 on Build GRCh38
rs917366673 Chr.10: 71760878-71760878 on Build GRCh38
rs1057074776 Chr.10: 71761024-71761024 on Build GRCh38
rs768732460 Chr.10: 71760948-71760948 on Build GRCh38
rs180989338 Chr.10: 71760972-71760972 on Build GRCh38
rs3747869 Chr.10: 71760875-71760875 on Build GRCh38
rs765456775 Chr.10: 71760975-71760975 on Build GRCh38
rs754553759 Chr.10: 71760903-71760903 on Build GRCh38
rs952408709 Chr.10: 71761242-71761242 on Build GRCh38
rs777657488 Chr.10: 71760922-71760922 on Build GRCh38
rs545508585 Chr.10: 71761145-71761145 on Build GRCh38
rs569225952 Chr.10: 71760963-71760963 on Build GRCh38
rs1008140560 Chr.10: 71761196-71761199 on Build GRCh38
rs540452540 Chr.10: 71761144-71761144 on Build GRCh38
rs534829898 Chr.10: 71761023-71761023 on Build GRCh38
rs200189378 Chr.10: 71760848-71760848 on Build GRCh38
rs375563247 Chr.10: 71760998-71760998 on Build GRCh38
rs377371485 Chr.10: 71760838-71760838 on Build GRCh38
rs961185314 Chr.10: 71761207-71761207 on Build GRCh38
rs755835959 Chr.10: 71760918-71760918 on Build GRCh38
rs188723368 Chr.10: 71761102-71761102 on Build GRCh38
rs751025857 Chr.10: 71760896-71760896 on Build GRCh38
rs532708167 Chr.10: 71761164-71761164 on Build GRCh38
rs3833469 Chr.10: 71760861-71760861 on Build GRCh38
rs761380688 Chr.10: 71761214-71761214 on Build GRCh38
rs909735601 Chr.10: 71760931-71760931 on Build GRCh38
rs1035054622 Chr.10: 71761167-71761167 on Build GRCh38
rs41305993 Chr.10: 71761069-71761069 on Build GRCh38
rs780594227 Chr.10: 71760905-71760905 on Build GRCh38
rs958983071 Chr.10: 71760859-71760859 on Build GRCh38
rs903923434 Chr.10: 71761125-71761125 on Build GRCh38
rs866639457 Chr.10: 71760833-71760833 on Build GRCh38
rs781265157 Chr.10: 71760997-71760997 on Build GRCh38
rs553191453 Chr.10: 71761056-71761056 on Build GRCh38
rs201924865 Chr.10: 71760888-71760888 on Build GRCh38
rs370808519 Chr.10: 71760982-71760982 on Build GRCh38
rs575196887 Chr.10: 71761095-71761095 on Build GRCh38
rs149127816 Chr.10: 71761068-71761068 on Build GRCh38

View Full Product Details