Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 76876901-76877399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNMA1
Entrez Gene ID
3778
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
UniGene
Hs.144795
Gene Chromosome Location
Chr.10: 76869601-77637819 on Build GRCh38
Transcripts
NM_001014797.2 NM_001271518.1 NM_001322829.1 NM_001322830.1 NM_001322832.1 NM_001322835.1 NM_001322838.1 XM_005269776.3 XM_005269778.1 XM_005269781.2 XM_005269787.3 XM_005269789.2 XM_005269792.2 XM_005269796.2 XM_011539773.1 XM_011539775.1 XM_011539776.1 XM_011539780.1 XM_011539781.2 XM_011539785.1 XM_017016207.1 XM_017016208.1 XM_017016210.1 XM_017016211.1 XM_017016212.1 XM_017016213.1 XM_017016216.1 XM_017016217.1 XM_017016223.1

SNP Detail

SNP ID Chromosome Location
rs930635654 Chr.10: 76877205-76877205 on Build GRCh38
rs748254202 Chr.10: 76877195-76877195 on Build GRCh38
rs749606661 Chr.10: 76877104-76877104 on Build GRCh38
rs1001594994 Chr.10: 76877213-76877213 on Build GRCh38
rs949339512 Chr.10: 76877202-76877202 on Build GRCh38
rs912186379 Chr.10: 76877193-76877193 on Build GRCh38
rs207471158 Chr.10: 76876982-76876982 on Build GRCh38
rs886958569 Chr.10: 76876958-76876958 on Build GRCh38
rs978937500 Chr.10: 76877111-76877111 on Build GRCh38
rs1056983039 Chr.10: 76877263-76877263 on Build GRCh38
rs953176506 Chr.10: 76877371-76877371 on Build GRCh38
rs1004008281 Chr.10: 76876971-76876971 on Build GRCh38
rs907223156 Chr.10: 76877206-76877206 on Build GRCh38
rs184033826 Chr.10: 76877077-76877077 on Build GRCh38
rs962275534 Chr.10: 76877324-76877324 on Build GRCh38
rs531470195 Chr.10: 76877329-76877329 on Build GRCh38
rs1015134613 Chr.10: 76877302-76877302 on Build GRCh38
rs952324112 Chr.10: 76876991-76876991 on Build GRCh38
rs1029974162 Chr.10: 76877327-76877327 on Build GRCh38
rs552414379 Chr.10: 76877175-76877175 on Build GRCh38
rs531630962 Chr.10: 76877241-76877241 on Build GRCh38
rs1035614519 Chr.10: 76876975-76876975 on Build GRCh38
rs375222113 Chr.10: 76877219-76877219 on Build GRCh38
rs542032963 Chr.10: 76876995-76876995 on Build GRCh38
rs922958693 Chr.10: 76877083-76877083 on Build GRCh38
rs778933212 Chr.10: 76877192-76877192 on Build GRCh38
rs527752817 Chr.10: 76877037-76877037 on Build GRCh38
rs562179463 Chr.10: 76877005-76877005 on Build GRCh38
rs1034508156 Chr.10: 76876981-76876981 on Build GRCh38
rs975737195 Chr.10: 76877082-76877082 on Build GRCh38
rs570492621 Chr.10: 76877204-76877204 on Build GRCh38
rs562589840 Chr.10: 76877245-76877245 on Build GRCh38
rs868051972 Chr.10: 76877270-76877270 on Build GRCh38
rs1031381699 Chr.10: 76877114-76877114 on Build GRCh38
rs1047776309 Chr.10: 76876946-76876946 on Build GRCh38
rs907917251 Chr.10: 76877220-76877220 on Build GRCh38
rs763799203 Chr.10: 76877138-76877141 on Build GRCh38
rs956412792 Chr.10: 76877128-76877128 on Build GRCh38
rs925112303 Chr.10: 76877146-76877147 on Build GRCh38
rs756661504 Chr.10: 76876983-76876983 on Build GRCh38
rs974746065 Chr.10: 76877203-76877203 on Build GRCh38
rs572577611 Chr.10: 76877059-76877059 on Build GRCh38
rs913819843 Chr.10: 76877202-76877202 on Build GRCh38
rs998200129 Chr.10: 76877326-76877326 on Build GRCh38
rs895759062 Chr.10: 76877344-76877344 on Build GRCh38
rs188936080 Chr.10: 76877096-76877096 on Build GRCh38
rs564761250 Chr.10: 76877081-76877081 on Build GRCh38
rs1046189377 Chr.10: 76876925-76876925 on Build GRCh38

View Full Product Details