Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80092056-80092569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM254
Entrez Gene ID
80195
Gene Name
transmembrane protein 254
Gene Aliases
C10orf57, bA369J21.6
UniGene
Hs.169982
Gene Chromosome Location
Chr.10: 80078646-80092552 on Build GRCh38
Transcripts
NM_001270367.1 NM_001270368.1 NM_001270369.1 NM_001270370.1 NM_001270371.1 NM_001270372.1 NM_001270373.1 NM_001270374.1 NM_025125.3 NR_072984.1 NR_072985.1 NR_072986.1 NR_072987.1 XM_006717978.2

SNP Detail

SNP ID Chromosome Location
rs974070917 Chr.10: 80092307-80092307 on Build GRCh38
rs184257001 Chr.10: 80092237-80092237 on Build GRCh38
rs780138855 Chr.10: 80092436-80092436 on Build GRCh38
rs368925025 Chr.10: 80092115-80092115 on Build GRCh38
rs931661136 Chr.10: 80092430-80092430 on Build GRCh38
rs578236251 Chr.10: 80092311-80092311 on Build GRCh38
rs935148540 Chr.10: 80092519-80092519 on Build GRCh38
rs564026657 Chr.10: 80092402-80092402 on Build GRCh38
rs3156 Chr.10: 80092303-80092303 on Build GRCh38
rs1051496781 Chr.10: 80092457-80092457 on Build GRCh38
rs111237914 Chr.10: 80092325-80092325 on Build GRCh38
rs577690155 Chr.10: 80092446-80092446 on Build GRCh38
rs757001076 Chr.10: 80092162-80092162 on Build GRCh38
rs911673906 Chr.10: 80092461-80092463 on Build GRCh38
rs113104047 Chr.10: 80092243-80092243 on Build GRCh38
rs113358488 Chr.10: 80092545-80092545 on Build GRCh38
rs557861704 Chr.10: 80092376-80092376 on Build GRCh38
rs1021590120 Chr.10: 80092185-80092185 on Build GRCh38
rs543287422 Chr.10: 80092476-80092476 on Build GRCh38
rs573811615 Chr.10: 80092213-80092213 on Build GRCh38
rs113163061 Chr.10: 80092253-80092253 on Build GRCh38
rs142228076 Chr.10: 80092413-80092413 on Build GRCh38
rs550565483 Chr.10: 80092196-80092196 on Build GRCh38
rs78066698 Chr.10: 80092097-80092097 on Build GRCh38
rs566103341 Chr.10: 80092248-80092248 on Build GRCh38
rs1011981900 Chr.10: 80092111-80092111 on Build GRCh38
rs14598 Chr.10: 80092469-80092469 on Build GRCh38
rs559119945 Chr.10: 80092533-80092533 on Build GRCh38
rs557225681 Chr.10: 80092133-80092133 on Build GRCh38
rs770150921 Chr.10: 80092170-80092170 on Build GRCh38

View Full Product Details