Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80157503-80158045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA11
Entrez Gene ID
311
Gene Name
annexin A11
Gene Aliases
ANX11, CAP50
UniGene
Hs.530291
Gene Chromosome Location
Chr.10: 80153953-80205677 on Build GRCh38
Transcripts
NM_001157.2 NM_001278407.1 NM_001278408.1 NM_001278409.1 NM_145868.1 NM_145869.1 XM_005269741.4 XM_005269742.1 XM_006717813.1 XM_006717814.3 XM_011539735.1 XM_011539736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5926721 c.175-5T>G p.? Chr.10: 80158030-80158030 on Build GRCh38
5441495 c.1381C>T p.R461C Chr.10: 80157718-80157718 on Build GRCh38
4662143 c.237G>C p.G79G Chr.10: 80157760-80157760 on Build GRCh38
3440969 c.1302C>T p.A434A Chr.10: 80158000-80158000 on Build GRCh38
3440968 c.1457C>T p.S486L Chr.10: 80157642-80157642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768113693 Chr.10: 80157775-80157775 on Build GRCh38
rs762980671 Chr.10: 80157967-80157967 on Build GRCh38
rs776038968 Chr.10: 80157956-80157956 on Build GRCh38
rs774676209 Chr.10: 80157667-80157667 on Build GRCh38
rs867842506 Chr.10: 80157746-80157746 on Build GRCh38
rs376025310 Chr.10: 80157992-80157992 on Build GRCh38
rs148593238 Chr.10: 80157733-80157733 on Build GRCh38
rs756336958 Chr.10: 80157923-80157923 on Build GRCh38
rs146644733 Chr.10: 80157675-80157675 on Build GRCh38
rs146495937 Chr.10: 80157674-80157674 on Build GRCh38
rs753789357 Chr.10: 80157681-80157681 on Build GRCh38
rs142956872 Chr.10: 80157706-80157706 on Build GRCh38
rs564117121 Chr.10: 80157529-80157529 on Build GRCh38
rs779282732 Chr.10: 80157734-80157734 on Build GRCh38
rs557024909 Chr.10: 80157856-80157856 on Build GRCh38
rs746232888 Chr.10: 80157946-80157946 on Build GRCh38
rs950907127 Chr.10: 80157761-80157761 on Build GRCh38
rs764120084 Chr.10: 80157924-80157924 on Build GRCh38
rs1048821917 Chr.10: 80157881-80157881 on Build GRCh38
rs533129675 Chr.10: 80157562-80157562 on Build GRCh38
rs372366508 Chr.10: 80157945-80157945 on Build GRCh38
rs774164541 Chr.10: 80157989-80157989 on Build GRCh38
rs142001721 Chr.10: 80157744-80157744 on Build GRCh38
rs774408735 Chr.10: 80157920-80157920 on Build GRCh38
rs749809198 Chr.10: 80157732-80157732 on Build GRCh38
rs901613582 Chr.10: 80157541-80157541 on Build GRCh38
rs544924707 Chr.10: 80157951-80157951 on Build GRCh38
rs746183884 Chr.10: 80157737-80157737 on Build GRCh38
rs765293205 Chr.10: 80158002-80158002 on Build GRCh38
rs978553622 Chr.10: 80157558-80157558 on Build GRCh38
rs199870592 Chr.10: 80158000-80158000 on Build GRCh38
rs750209333 Chr.10: 80157695-80157695 on Build GRCh38
rs752833540 Chr.10: 80157711-80157711 on Build GRCh38
rs767785759 Chr.10: 80157804-80157804 on Build GRCh38
rs12767142 Chr.10: 80157873-80157873 on Build GRCh38
rs879227548 Chr.10: 80157765-80157765 on Build GRCh38
rs765169730 Chr.10: 80157692-80157692 on Build GRCh38
rs763891108 Chr.10: 80157590-80157590 on Build GRCh38
rs374382845 Chr.10: 80157707-80157707 on Build GRCh38
rs369428921 Chr.10: 80157606-80157606 on Build GRCh38
rs749668544 Chr.10: 80157650-80157650 on Build GRCh38
rs529536583 Chr.10: 80157628-80157628 on Build GRCh38
rs758911878 Chr.10: 80158007-80158007 on Build GRCh38
rs79950900 Chr.10: 80157637-80157637 on Build GRCh38
rs140115990 Chr.10: 80157991-80157991 on Build GRCh38
rs771342827 Chr.10: 80157660-80157660 on Build GRCh38
rs778082972 Chr.10: 80157642-80157642 on Build GRCh38
rs778864661 Chr.10: 80157766-80157766 on Build GRCh38
rs944658328 Chr.10: 80157855-80157855 on Build GRCh38
rs368132715 Chr.10: 80157676-80157676 on Build GRCh38
rs113046621 Chr.10: 80157871-80157871 on Build GRCh38
rs761885687 Chr.10: 80157783-80157783 on Build GRCh38
rs956787240 Chr.10: 80158021-80158021 on Build GRCh38
rs922896835 Chr.10: 80157872-80157872 on Build GRCh38
rs866151231 Chr.10: 80157705-80157705 on Build GRCh38
rs1012638618 Chr.10: 80157728-80157728 on Build GRCh38
rs201343738 Chr.10: 80157786-80157786 on Build GRCh38
rs371405651 Chr.10: 80157961-80157961 on Build GRCh38
rs199749354 Chr.10: 80157596-80157596 on Build GRCh38
rs12775374 Chr.10: 80157564-80157564 on Build GRCh38
rs968143559 Chr.10: 80157534-80157534 on Build GRCh38
rs867506376 Chr.10: 80157828-80157828 on Build GRCh38
rs537007303 Chr.10: 80157830-80157830 on Build GRCh38
rs955878628 Chr.10: 80157799-80157799 on Build GRCh38
rs778842350 Chr.10: 80157619-80157619 on Build GRCh38
rs1802932 Chr.10: 80157730-80157730 on Build GRCh38
rs766145443 Chr.10: 80157986-80157986 on Build GRCh38
rs768327712 Chr.10: 80157954-80157954 on Build GRCh38
rs760374358 Chr.10: 80157753-80157753 on Build GRCh38
rs747615399 Chr.10: 80157960-80157960 on Build GRCh38
rs748134350 Chr.10: 80157636-80157636 on Build GRCh38
rs773166335 Chr.10: 80157787-80157787 on Build GRCh38
rs771674112 Chr.10: 80157669-80157669 on Build GRCh38
rs752929121 Chr.10: 80157921-80157921 on Build GRCh38
rs371825050 Chr.10: 80157690-80157690 on Build GRCh38
rs781458762 Chr.10: 80157635-80157635 on Build GRCh38
rs77724375 Chr.10: 80157950-80157950 on Build GRCh38
rs773217818 Chr.10: 80157963-80157963 on Build GRCh38
rs909477417 Chr.10: 80157836-80157836 on Build GRCh38
rs369623714 Chr.10: 80157895-80157895 on Build GRCh38
rs988730045 Chr.10: 80157840-80157840 on Build GRCh38
rs753874099 Chr.10: 80157940-80157940 on Build GRCh38
rs776180846 Chr.10: 80157781-80157781 on Build GRCh38
rs911767215 Chr.10: 80157846-80157846 on Build GRCh38
rs757782267 Chr.10: 80157941-80157941 on Build GRCh38
rs368569582 Chr.10: 80157789-80157789 on Build GRCh38
rs776537022 Chr.10: 80157600-80157600 on Build GRCh38
rs750982225 Chr.10: 80158006-80158006 on Build GRCh38
rs188329789 Chr.10: 80157621-80157621 on Build GRCh38
rs34130731 Chr.10: 80157725-80157726 on Build GRCh38
rs961003614 Chr.10: 80157717-80157717 on Build GRCh38
rs150671497 Chr.10: 80157745-80157745 on Build GRCh38
rs1079242 Chr.10: 80157952-80157952 on Build GRCh38
rs755226655 Chr.10: 80157634-80157634 on Build GRCh38
rs369416316 Chr.10: 80157907-80157907 on Build GRCh38
rs143921438 Chr.10: 80157649-80157649 on Build GRCh38
rs1000931056 Chr.10: 80157974-80157974 on Build GRCh38
rs756080261 Chr.10: 80157712-80157712 on Build GRCh38

View Full Product Details