Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 85600723-85601232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRID1
Entrez Gene ID
2894
Gene Name
glutamate ionotropic receptor delta type subunit 1
Gene Aliases
GluD1
UniGene
Hs.530653
Gene Chromosome Location
Chr.10: 85599555-86366493 on Build GRCh38
Transcripts
NM_017551.2 NR_038986.1

Gene Symbol
GRID1-AS1
Entrez Gene ID
100507470
Gene Name
GRID1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.585483
Gene Chromosome Location
Chr.10: 85577731-85607215 on Build GRCh38
Transcripts
NM_017551.2 NR_038986.1

SNP Detail

SNP ID Chromosome Location
rs1022721759 Chr.10: 85600978-85600978 on Build GRCh38
rs904235666 Chr.10: 85600874-85600874 on Build GRCh38
rs904221837 Chr.10: 85601081-85601081 on Build GRCh38
rs35668931 Chr.10: 85600779-85600779 on Build GRCh38
rs3812654 Chr.10: 85600936-85600936 on Build GRCh38
rs1052619158 Chr.10: 85601017-85601017 on Build GRCh38
rs866608508 Chr.10: 85601201-85601201 on Build GRCh38
rs895194326 Chr.10: 85601190-85601190 on Build GRCh38
rs538854921 Chr.10: 85600979-85600979 on Build GRCh38
rs1008656036 Chr.10: 85601174-85601174 on Build GRCh38
rs574802795 Chr.10: 85601099-85601099 on Build GRCh38
rs543027541 Chr.10: 85600768-85600768 on Build GRCh38
rs41313467 Chr.10: 85601160-85601160 on Build GRCh38
rs577799708 Chr.10: 85601180-85601180 on Build GRCh38
rs1003736882 Chr.10: 85600945-85600945 on Build GRCh38
rs76899086 Chr.10: 85601065-85601065 on Build GRCh38
rs780612279 Chr.10: 85601183-85601183 on Build GRCh38
rs139782709 Chr.10: 85601200-85601200 on Build GRCh38
rs141657638 Chr.10: 85600782-85600782 on Build GRCh38
rs895236433 Chr.10: 85600964-85600964 on Build GRCh38
rs529589315 Chr.10: 85600817-85600817 on Build GRCh38
rs928150173 Chr.10: 85601006-85601006 on Build GRCh38
rs561790875 Chr.10: 85601087-85601087 on Build GRCh38
rs189407731 Chr.10: 85600780-85600780 on Build GRCh38
rs760937948 Chr.10: 85600948-85600948 on Build GRCh38
rs139309715 Chr.10: 85601098-85601098 on Build GRCh38
rs181676847 Chr.10: 85600785-85600785 on Build GRCh38
rs908985571 Chr.10: 85600991-85600991 on Build GRCh38
rs866497868 Chr.10: 85601057-85601057 on Build GRCh38
rs543574486 Chr.10: 85600847-85600847 on Build GRCh38
rs1009880176 Chr.10: 85600862-85600862 on Build GRCh38
rs1052725235 Chr.10: 85600834-85600834 on Build GRCh38
rs773888344 Chr.10: 85600893-85600893 on Build GRCh38
rs1022854724 Chr.10: 85600776-85600776 on Build GRCh38
rs761128847 Chr.10: 85600900-85600900 on Build GRCh38
rs748668642 Chr.10: 85600755-85600755 on Build GRCh38
rs774847406 Chr.10: 85601204-85601204 on Build GRCh38
rs910095237 Chr.10: 85600879-85600879 on Build GRCh38
rs532975086 Chr.10: 85600789-85600789 on Build GRCh38
rs112932451 Chr.10: 85600922-85600922 on Build GRCh38
rs973351517 Chr.10: 85600939-85600939 on Build GRCh38
rs115949885 Chr.10: 85601181-85601181 on Build GRCh38
rs146201379 Chr.10: 85600863-85600863 on Build GRCh38
rs554342357 Chr.10: 85600990-85600990 on Build GRCh38
rs1028818056 Chr.10: 85600887-85600887 on Build GRCh38
rs531308611 Chr.10: 85600832-85600832 on Build GRCh38
rs2664404 Chr.10: 85600966-85600966 on Build GRCh38
rs932305845 Chr.10: 85600841-85600841 on Build GRCh38
rs762688881 Chr.10: 85600826-85600826 on Build GRCh38
rs954742537 Chr.10: 85600889-85600889 on Build GRCh38
rs531984550 Chr.10: 85600949-85600949 on Build GRCh38
rs1031570637 Chr.10: 85601005-85601005 on Build GRCh38
rs897315106 Chr.10: 85600944-85600944 on Build GRCh38
rs2616672 Chr.10: 85600955-85600955 on Build GRCh38
rs1057044963 Chr.10: 85601188-85601188 on Build GRCh38
rs996336020 Chr.10: 85600876-85600876 on Build GRCh38
rs373665615 Chr.10: 85600838-85600838 on Build GRCh38
rs757366645 Chr.10: 85601016-85601016 on Build GRCh38
rs1014821258 Chr.10: 85600935-85600935 on Build GRCh38
rs1000054491 Chr.10: 85601002-85601002 on Build GRCh38
rs909541740 Chr.10: 85601121-85601121 on Build GRCh38
rs891456353 Chr.10: 85600846-85600846 on Build GRCh38
rs907879196 Chr.10: 85600781-85600781 on Build GRCh38
rs991249787 Chr.10: 85600757-85600757 on Build GRCh38
rs185845078 Chr.10: 85600823-85600823 on Build GRCh38
rs750995261 Chr.10: 85601058-85601058 on Build GRCh38
rs566144720 Chr.10: 85600992-85600992 on Build GRCh38
rs939536547 Chr.10: 85601014-85601014 on Build GRCh38
rs41313469 Chr.10: 85601033-85601033 on Build GRCh38

View Full Product Details