Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86983007-86983551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP11
Entrez Gene ID
119385
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 11
Gene Aliases
-
UniGene
Hs.511787
Gene Chromosome Location
Chr.10: 86970741-87010203 on Build GRCh38
Transcripts
NM_133447.1

SNP Detail

SNP ID Chromosome Location
rs527513448 Chr.10: 86983312-86983312 on Build GRCh38
rs1035643403 Chr.10: 86983060-86983060 on Build GRCh38
rs867172873 Chr.10: 86983383-86983383 on Build GRCh38
rs1057347750 Chr.10: 86983323-86983323 on Build GRCh38
rs181856235 Chr.10: 86983185-86983185 on Build GRCh38
rs925444979 Chr.10: 86983145-86983145 on Build GRCh38
rs942140491 Chr.10: 86983264-86983264 on Build GRCh38
rs766144368 Chr.10: 86983474-86983474 on Build GRCh38
rs968509565 Chr.10: 86983090-86983090 on Build GRCh38
rs773269620 Chr.10: 86983412-86983412 on Build GRCh38
rs923587054 Chr.10: 86983178-86983178 on Build GRCh38
rs4104990 Chr.10: 86983391-86983391 on Build GRCh38
rs117778404 Chr.10: 86983528-86983528 on Build GRCh38
rs1023039117 Chr.10: 86983082-86983082 on Build GRCh38
rs761854413 Chr.10: 86983334-86983334 on Build GRCh38
rs897903906 Chr.10: 86983316-86983316 on Build GRCh38
rs968333509 Chr.10: 86983403-86983403 on Build GRCh38
rs993328292 Chr.10: 86983318-86983318 on Build GRCh38
rs1013220511 Chr.10: 86983338-86983338 on Build GRCh38
rs577467769 Chr.10: 86983055-86983055 on Build GRCh38
rs960224195 Chr.10: 86983068-86983068 on Build GRCh38
rs933550848 Chr.10: 86983208-86983208 on Build GRCh38
rs978483451 Chr.10: 86983131-86983131 on Build GRCh38
rs370586579 Chr.10: 86983211-86983211 on Build GRCh38
rs988300737 Chr.10: 86983174-86983174 on Build GRCh38
rs562180389 Chr.10: 86983468-86983468 on Build GRCh38
rs116717319 Chr.10: 86983262-86983262 on Build GRCh38
rs991192103 Chr.10: 86983081-86983081 on Build GRCh38
rs956825574 Chr.10: 86983153-86983153 on Build GRCh38
rs116876871 Chr.10: 86983501-86983501 on Build GRCh38
rs763260484 Chr.10: 86983422-86983422 on Build GRCh38
rs776842171 Chr.10: 86983052-86983052 on Build GRCh38

View Full Product Details