Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86993144-86993644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP11
Entrez Gene ID
119385
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 11
Gene Aliases
-
UniGene
Hs.511787
Gene Chromosome Location
Chr.10: 86970741-87010203 on Build GRCh38
Transcripts
NM_133447.1

SNP Detail

SNP ID Chromosome Location
rs111428970 Chr.10: 86993231-86993231 on Build GRCh38
rs558876474 Chr.10: 86993565-86993565 on Build GRCh38
rs368436297 Chr.10: 86993324-86993324 on Build GRCh38
rs140678241 Chr.10: 86993469-86993469 on Build GRCh38
rs544384658 Chr.10: 86993301-86993301 on Build GRCh38
rs375139833 Chr.10: 86993557-86993557 on Build GRCh38
rs753640744 Chr.10: 86993492-86993492 on Build GRCh38
rs187780115 Chr.10: 86993615-86993615 on Build GRCh38
rs146841657 Chr.10: 86993425-86993425 on Build GRCh38
rs1046221032 Chr.10: 86993490-86993490 on Build GRCh38
rs2131391 Chr.10: 86993208-86993208 on Build GRCh38
rs906273888 Chr.10: 86993504-86993504 on Build GRCh38
rs547894786 Chr.10: 86993424-86993424 on Build GRCh38
rs914558283 Chr.10: 86993293-86993293 on Build GRCh38
rs570663465 Chr.10: 86993388-86993388 on Build GRCh38
rs540777398 Chr.10: 86993297-86993297 on Build GRCh38
rs530837782 Chr.10: 86993261-86993261 on Build GRCh38
rs971581028 Chr.10: 86993256-86993256 on Build GRCh38
rs886744029 Chr.10: 86993461-86993461 on Build GRCh38
rs990561144 Chr.10: 86993280-86993280 on Build GRCh38
rs752229262 Chr.10: 86993522-86993522 on Build GRCh38
rs1003461482 Chr.10: 86993521-86993521 on Build GRCh38
rs576280744 Chr.10: 86993555-86993555 on Build GRCh38
rs554982920 Chr.10: 86993257-86993257 on Build GRCh38
rs574865302 Chr.10: 86993279-86993279 on Build GRCh38
rs1035048336 Chr.10: 86993275-86993275 on Build GRCh38
rs111480848 Chr.10: 86993545-86993545 on Build GRCh38
rs1048554394 Chr.10: 86993459-86993459 on Build GRCh38
rs369396639 Chr.10: 86993390-86993390 on Build GRCh38
rs536210237 Chr.10: 86993433-86993433 on Build GRCh38
rs921185922 Chr.10: 86993438-86993438 on Build GRCh38
rs977388617 Chr.10: 86993604-86993604 on Build GRCh38
rs931206042 Chr.10: 86993456-86993456 on Build GRCh38
rs1011804539 Chr.10: 86993546-86993546 on Build GRCh38
rs912677517 Chr.10: 86993612-86993612 on Build GRCh38
rs183172701 Chr.10: 86993444-86993444 on Build GRCh38
rs560559368 Chr.10: 86993449-86993449 on Build GRCh38
rs199627109 Chr.10: 86993568-86993568 on Build GRCh38
rs1745902 Chr.10: 86993572-86993572 on Build GRCh38

View Full Product Details