Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87753000-87753497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
UniGene
Hs.435948
Gene Chromosome Location
Chr.10: 87751512-87841345 on Build GRCh38
Transcripts
NM_001321967.1 NM_001321968.1 NM_001321969.1 NM_032810.3 NR_135914.1 XM_005270252.4 XM_011540302.1 XM_017016847.1 XM_017016848.1 XM_017016849.1 XM_017016850.1

SNP Detail

SNP ID Chromosome Location
rs1008044215 Chr.10: 87753372-87753372 on Build GRCh38
rs373457160 Chr.10: 87753458-87753458 on Build GRCh38
rs754140259 Chr.10: 87753211-87753211 on Build GRCh38
rs986698194 Chr.10: 87753071-87753071 on Build GRCh38
rs188784348 Chr.10: 87753081-87753081 on Build GRCh38
rs1011742813 Chr.10: 87753327-87753327 on Build GRCh38
rs11546681 Chr.10: 87753365-87753365 on Build GRCh38
rs892850040 Chr.10: 87753318-87753318 on Build GRCh38
rs12253113 Chr.10: 87753082-87753082 on Build GRCh38
rs569705250 Chr.10: 87753328-87753328 on Build GRCh38
rs1002316930 Chr.10: 87753035-87753035 on Build GRCh38
rs908715456 Chr.10: 87753220-87753222 on Build GRCh38
rs949594732 Chr.10: 87753258-87753258 on Build GRCh38
rs931422468 Chr.10: 87753204-87753204 on Build GRCh38
rs764382355 Chr.10: 87753050-87753050 on Build GRCh38
rs757847509 Chr.10: 87753110-87753110 on Build GRCh38
rs7481 Chr.10: 87753361-87753361 on Build GRCh38
rs920021375 Chr.10: 87753181-87753181 on Build GRCh38
rs1034672929 Chr.10: 87753044-87753044 on Build GRCh38
rs781530943 Chr.10: 87753344-87753344 on Build GRCh38
rs780455136 Chr.10: 87753470-87753470 on Build GRCh38
rs937017599 Chr.10: 87753287-87753287 on Build GRCh38
rs751163214 Chr.10: 87753152-87753156 on Build GRCh38
rs757564565 Chr.10: 87753181-87753181 on Build GRCh38
rs1046696985 Chr.10: 87753259-87753259 on Build GRCh38
rs56939055 Chr.10: 87753061-87753061 on Build GRCh38
rs960458491 Chr.10: 87753045-87753045 on Build GRCh38
rs55897649 Chr.10: 87753168-87753168 on Build GRCh38
rs756472589 Chr.10: 87753028-87753028 on Build GRCh38
rs779256612 Chr.10: 87753260-87753260 on Build GRCh38
rs963970798 Chr.10: 87753434-87753434 on Build GRCh38
rs551996583 Chr.10: 87753419-87753419 on Build GRCh38
rs967228222 Chr.10: 87753070-87753070 on Build GRCh38
rs758503137 Chr.10: 87753446-87753446 on Build GRCh38
rs975843845 Chr.10: 87753153-87753153 on Build GRCh38
rs1019471084 Chr.10: 87753401-87753401 on Build GRCh38
rs1020092955 Chr.10: 87753333-87753333 on Build GRCh38
rs758114862 Chr.10: 87753052-87753053 on Build GRCh38

View Full Product Details