Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89709089-89709605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF20B
Entrez Gene ID
9585
Gene Name
kinesin family member 20B
Gene Aliases
CT90, KRMP1, MPHOSPH1, MPP-1, MPP1
UniGene
Hs.240
Gene Chromosome Location
Chr.10: 89701590-89774943 on Build GRCh38
Transcripts
NM_001284259.1 NM_016195.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4820501 c.283C>T p.Q95* Chr.10: 89709393-89709393 on Build GRCh38
7078144 c.177C>G p.L59L Chr.10: 89709196-89709196 on Build GRCh38
4469399 c.159C>T p.F53F Chr.10: 89709178-89709178 on Build GRCh38
921302 c.334A>C p.K112Q Chr.10: 89709444-89709444 on Build GRCh38
6130882 c.161A>T p.E54V Chr.10: 89709180-89709180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115396465 Chr.10: 89709374-89709374 on Build GRCh38
rs777647873 Chr.10: 89709124-89709125 on Build GRCh38
rs755293250 Chr.10: 89709502-89709502 on Build GRCh38
rs200362702 Chr.10: 89709388-89709388 on Build GRCh38
rs745587216 Chr.10: 89709209-89709209 on Build GRCh38
rs376346565 Chr.10: 89709457-89709457 on Build GRCh38
rs1038163377 Chr.10: 89709459-89709459 on Build GRCh38
rs201280490 Chr.10: 89709210-89709210 on Build GRCh38
rs201328071 Chr.10: 89709382-89709382 on Build GRCh38
rs142799530 Chr.10: 89709343-89709343 on Build GRCh38
rs758141800 Chr.10: 89709185-89709185 on Build GRCh38
rs761820930 Chr.10: 89709305-89709305 on Build GRCh38
rs780460247 Chr.10: 89709386-89709386 on Build GRCh38
rs778521871 Chr.10: 89709178-89709178 on Build GRCh38
rs755633032 Chr.10: 89709123-89709123 on Build GRCh38
rs773209778 Chr.10: 89709408-89709408 on Build GRCh38
rs780807749 Chr.10: 89709374-89709375 on Build GRCh38
rs755701011 Chr.10: 89709243-89709243 on Build GRCh38
rs775766868 Chr.10: 89709295-89709295 on Build GRCh38
rs929663619 Chr.10: 89709539-89709539 on Build GRCh38
rs758962805 Chr.10: 89709381-89709381 on Build GRCh38
rs866058124 Chr.10: 89709227-89709227 on Build GRCh38
rs1046975138 Chr.10: 89709222-89709222 on Build GRCh38
rs371431314 Chr.10: 89709124-89709124 on Build GRCh38
rs201599384 Chr.10: 89709446-89709446 on Build GRCh38
rs984012162 Chr.10: 89709262-89709262 on Build GRCh38
rs756898969 Chr.10: 89709385-89709385 on Build GRCh38
rs778370870 Chr.10: 89709500-89709500 on Build GRCh38
rs766114784 Chr.10: 89709189-89709189 on Build GRCh38
rs777502341 Chr.10: 89709245-89709245 on Build GRCh38
rs1027766416 Chr.10: 89709544-89709544 on Build GRCh38
rs746988402 Chr.10: 89709133-89709133 on Build GRCh38
rs776677145 Chr.10: 89709304-89709304 on Build GRCh38
rs559026944 Chr.10: 89709158-89709158 on Build GRCh38
rs774649782 Chr.10: 89709423-89709423 on Build GRCh38
rs947776881 Chr.10: 89709451-89709451 on Build GRCh38
rs146592890 Chr.10: 89709321-89709321 on Build GRCh38
rs564952243 Chr.10: 89709389-89709389 on Build GRCh38
rs765303097 Chr.10: 89709315-89709315 on Build GRCh38
rs373776246 Chr.10: 89709138-89709138 on Build GRCh38
rs1056194785 Chr.10: 89709171-89709171 on Build GRCh38
rs567440461 Chr.10: 89709557-89709557 on Build GRCh38
rs759189061 Chr.10: 89709427-89709427 on Build GRCh38
rs199753367 Chr.10: 89709149-89709149 on Build GRCh38
rs763235804 Chr.10: 89709409-89709409 on Build GRCh38
rs369401035 Chr.10: 89709320-89709320 on Build GRCh38
rs898192915 Chr.10: 89709196-89709196 on Build GRCh38
rs200864660 Chr.10: 89709323-89709323 on Build GRCh38
rs771120255 Chr.10: 89709410-89709410 on Build GRCh38
rs767507186 Chr.10: 89709336-89709336 on Build GRCh38
rs755646052 Chr.10: 89709349-89709349 on Build GRCh38
rs781608999 Chr.10: 89709395-89709395 on Build GRCh38
rs917394146 Chr.10: 89709267-89709267 on Build GRCh38
rs781555701 Chr.10: 89709504-89709504 on Build GRCh38
rs756696385 Chr.10: 89709328-89709328 on Build GRCh38
rs375623454 Chr.10: 89709329-89709329 on Build GRCh38
rs531351218 Chr.10: 89709330-89709330 on Build GRCh38
rs947679379 Chr.10: 89709180-89709180 on Build GRCh38
rs1018290188 Chr.10: 89709263-89709263 on Build GRCh38
rs890920094 Chr.10: 89709225-89709225 on Build GRCh38
rs367726513 Chr.10: 89709191-89709191 on Build GRCh38
rs756920160 Chr.10: 89709460-89709460 on Build GRCh38
rs765498226 Chr.10: 89709127-89709127 on Build GRCh38
rs1018194599 Chr.10: 89709414-89709414 on Build GRCh38
rs554776959 Chr.10: 89709279-89709279 on Build GRCh38
rs369439648 Chr.10: 89709355-89709355 on Build GRCh38
rs764211631 Chr.10: 89709120-89709120 on Build GRCh38
rs750093147 Chr.10: 89709501-89709501 on Build GRCh38
rs928793253 Chr.10: 89709162-89709162 on Build GRCh38
rs977736665 Chr.10: 89709126-89709126 on Build GRCh38
rs574782938 Chr.10: 89709298-89709298 on Build GRCh38
rs1047393727 Chr.10: 89709546-89709546 on Build GRCh38
rs41286896 Chr.10: 89709529-89709529 on Build GRCh38
rs747721266 Chr.10: 89709233-89709233 on Build GRCh38
rs748681486 Chr.10: 89709396-89709396 on Build GRCh38
rs1011647301 Chr.10: 89709284-89709284 on Build GRCh38
rs767453897 Chr.10: 89709438-89709438 on Build GRCh38
rs140554027 Chr.10: 89709248-89709248 on Build GRCh38
rs375851858 Chr.10: 89709198-89709198 on Build GRCh38
rs1129777 Chr.10: 89709168-89709168 on Build GRCh38
rs769216570 Chr.10: 89709302-89709302 on Build GRCh38
rs746431610 Chr.10: 89709285-89709285 on Build GRCh38
rs752634082 Chr.10: 89709441-89709441 on Build GRCh38
rs199632456 Chr.10: 89709399-89709399 on Build GRCh38

View Full Product Details