Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91908220-91908709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFBP3
Entrez Gene ID
143282
Gene Name
fibroblast growth factor binding protein 3
Gene Aliases
C10orf13, FGF-BP3
UniGene
Hs.466120
Gene Chromosome Location
Chr.10: 91906588-91909501 on Build GRCh38
Transcripts
NM_152429.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3441494 c.597C>T p.P199P Chr.10: 91908373-91908373 on Build GRCh38
7175187 c.744C>T p.L248L Chr.10: 91908226-91908226 on Build GRCh38
3849489 c.612C>G p.T204T Chr.10: 91908358-91908358 on Build GRCh38
3765523 c.319G>A p.A107T Chr.10: 91908651-91908651 on Build GRCh38
6888976 c.624G>A p.K208K Chr.10: 91908346-91908346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772191465 Chr.10: 91908466-91908466 on Build GRCh38
rs768607072 Chr.10: 91908478-91908478 on Build GRCh38
rs781429443 Chr.10: 91908395-91908395 on Build GRCh38
rs767330127 Chr.10: 91908276-91908276 on Build GRCh38
rs541312392 Chr.10: 91908677-91908677 on Build GRCh38
rs770993208 Chr.10: 91908342-91908342 on Build GRCh38
rs747029737 Chr.10: 91908571-91908572 on Build GRCh38
rs11186740 Chr.10: 91908383-91908383 on Build GRCh38
rs748484895 Chr.10: 91908397-91908397 on Build GRCh38
rs751166125 Chr.10: 91908452-91908452 on Build GRCh38
rs903837247 Chr.10: 91908507-91908507 on Build GRCh38
rs762953279 Chr.10: 91908331-91908331 on Build GRCh38
rs145041514 Chr.10: 91908243-91908243 on Build GRCh38
rs771048358 Chr.10: 91908407-91908407 on Build GRCh38
rs757710382 Chr.10: 91908685-91908685 on Build GRCh38
rs11819077 Chr.10: 91908667-91908667 on Build GRCh38
rs368583604 Chr.10: 91908631-91908631 on Build GRCh38
rs528736388 Chr.10: 91908490-91908490 on Build GRCh38
rs753871709 Chr.10: 91908440-91908440 on Build GRCh38
rs765270957 Chr.10: 91908249-91908249 on Build GRCh38
rs752628287 Chr.10: 91908277-91908277 on Build GRCh38
rs751273902 Chr.10: 91908521-91908521 on Build GRCh38
rs779688626 Chr.10: 91908449-91908449 on Build GRCh38
rs777058494 Chr.10: 91908247-91908247 on Build GRCh38
rs867654038 Chr.10: 91908644-91908644 on Build GRCh38
rs1029811679 Chr.10: 91908405-91908405 on Build GRCh38
rs756898341 Chr.10: 91908373-91908373 on Build GRCh38
rs772599504 Chr.10: 91908504-91908504 on Build GRCh38
rs747882850 Chr.10: 91908610-91908610 on Build GRCh38
rs758567355 Chr.10: 91908587-91908587 on Build GRCh38
rs369733632 Chr.10: 91908260-91908260 on Build GRCh38
rs779962770 Chr.10: 91908593-91908593 on Build GRCh38
rs192010535 Chr.10: 91908672-91908672 on Build GRCh38
rs919575160 Chr.10: 91908388-91908388 on Build GRCh38
rs969362860 Chr.10: 91908574-91908574 on Build GRCh38
rs983394739 Chr.10: 91908641-91908641 on Build GRCh38
rs778088756 Chr.10: 91908459-91908459 on Build GRCh38
rs945902543 Chr.10: 91908528-91908528 on Build GRCh38
rs374461098 Chr.10: 91908448-91908448 on Build GRCh38
rs772292050 Chr.10: 91908425-91908425 on Build GRCh38
rs747534914 Chr.10: 91908602-91908602 on Build GRCh38
rs754795996 Chr.10: 91908270-91908270 on Build GRCh38
rs748630930 Chr.10: 91908310-91908310 on Build GRCh38
rs375841530 Chr.10: 91908384-91908384 on Build GRCh38
rs757778107 Chr.10: 91908550-91908550 on Build GRCh38
rs751360927 Chr.10: 91908265-91908265 on Build GRCh38
rs147290202 Chr.10: 91908390-91908390 on Build GRCh38
rs759710869 Chr.10: 91908422-91908422 on Build GRCh38
rs372722249 Chr.10: 91908446-91908446 on Build GRCh38
rs757723020 Chr.10: 91908464-91908464 on Build GRCh38
rs753408023 Chr.10: 91908371-91908371 on Build GRCh38
rs754208602 Chr.10: 91908679-91908679 on Build GRCh38
rs904178477 Chr.10: 91908364-91908364 on Build GRCh38
rs368507225 Chr.10: 91908417-91908417 on Build GRCh38
rs750992710 Chr.10: 91908689-91908689 on Build GRCh38
rs568110442 Chr.10: 91908501-91908501 on Build GRCh38
rs769470835 Chr.10: 91908617-91908617 on Build GRCh38
rs992193902 Chr.10: 91908684-91908684 on Build GRCh38
rs759210551 Chr.10: 91908673-91908673 on Build GRCh38
rs867419584 Chr.10: 91908653-91908653 on Build GRCh38
rs570576273 Chr.10: 91908589-91908589 on Build GRCh38
rs531188271 Chr.10: 91908512-91908512 on Build GRCh38
rs773810665 Chr.10: 91908664-91908664 on Build GRCh38
rs1044344783 Chr.10: 91908690-91908690 on Build GRCh38
rs766529535 Chr.10: 91908261-91908261 on Build GRCh38
rs1107947 Chr.10: 91908353-91908353 on Build GRCh38
rs963321743 Chr.10: 91908652-91908652 on Build GRCh38
rs529857593 Chr.10: 91908300-91908300 on Build GRCh38
rs746360136 Chr.10: 91908572-91908572 on Build GRCh38
rs963816409 Chr.10: 91908394-91908394 on Build GRCh38
rs756048253 Chr.10: 91908285-91908285 on Build GRCh38
rs368404329 Chr.10: 91908338-91908338 on Build GRCh38
rs1023885060 Chr.10: 91908561-91908561 on Build GRCh38
rs768947496 Chr.10: 91908308-91908308 on Build GRCh38
rs778883939 Chr.10: 91908465-91908465 on Build GRCh38
rs543541902 Chr.10: 91908343-91908343 on Build GRCh38
rs747323462 Chr.10: 91908242-91908242 on Build GRCh38
rs80269321 Chr.10: 91908294-91908294 on Build GRCh38
rs765953507 Chr.10: 91908659-91908659 on Build GRCh38
rs1031061548 Chr.10: 91908533-91908533 on Build GRCh38
rs759781653 Chr.10: 91908350-91908350 on Build GRCh38
rs951024225 Chr.10: 91908627-91908627 on Build GRCh38
rs752498235 Chr.10: 91908356-91908356 on Build GRCh38
rs142767199 Chr.10: 91908427-91908427 on Build GRCh38
rs747262063 Chr.10: 91908601-91908601 on Build GRCh38
rs747225127 Chr.10: 91908477-91908477 on Build GRCh38
rs140442876 Chr.10: 91908301-91908301 on Build GRCh38
rs763980493 Chr.10: 91908438-91908438 on Build GRCh38
rs775852199 Chr.10: 91908467-91908467 on Build GRCh38
rs777459521 Chr.10: 91908291-91908291 on Build GRCh38
rs1045147251 Chr.10: 91908246-91908246 on Build GRCh38
rs779413128 Chr.10: 91908570-91908570 on Build GRCh38
rs749781645 Chr.10: 91908406-91908406 on Build GRCh38
rs1009124043 Chr.10: 91908241-91908241 on Build GRCh38
rs569023750 Chr.10: 91908362-91908362 on Build GRCh38
rs543137706 Chr.10: 91908553-91908553 on Build GRCh38
rs765511010 Chr.10: 91908688-91908688 on Build GRCh38
rs144016948 Chr.10: 91908389-91908389 on Build GRCh38
rs900792888 Chr.10: 91908461-91908461 on Build GRCh38
rs776578516 Chr.10: 91908608-91908608 on Build GRCh38
rs770288218 Chr.10: 91908400-91908400 on Build GRCh38
rs981005188 Chr.10: 91908660-91908660 on Build GRCh38
rs781586024 Chr.10: 91908309-91908309 on Build GRCh38
rs899516770 Chr.10: 91908604-91908604 on Build GRCh38
rs912274448 Chr.10: 91908635-91908635 on Build GRCh38
rs548334485 Chr.10: 91908500-91908500 on Build GRCh38
rs974380997 Chr.10: 91908578-91908578 on Build GRCh38
rs997897695 Chr.10: 91908618-91908618 on Build GRCh38
rs999850637 Chr.10: 91908598-91908598 on Build GRCh38
rs34164735 Chr.10: 91908502-91908503 on Build GRCh38
rs754595032 Chr.10: 91908546-91908546 on Build GRCh38
rs773290877 Chr.10: 91908322-91908322 on Build GRCh38
rs776982068 Chr.10: 91908647-91908647 on Build GRCh38
rs773233397 Chr.10: 91908253-91908253 on Build GRCh38
rs756988127 Chr.10: 91908298-91908298 on Build GRCh38
rs776523354 Chr.10: 91908481-91908481 on Build GRCh38
rs778539937 Chr.10: 91908376-91908376 on Build GRCh38
rs762589417 Chr.10: 91908511-91908511 on Build GRCh38
rs778317588 Chr.10: 91908403-91908403 on Build GRCh38
rs777552441 Chr.10: 91908417-91908418 on Build GRCh38
rs775165413 Chr.10: 91908426-91908426 on Build GRCh38
rs976510746 Chr.10: 91908404-91908404 on Build GRCh38
rs770235150 Chr.10: 91908315-91908315 on Build GRCh38
rs139228214 Chr.10: 91908409-91908409 on Build GRCh38
rs761861234 Chr.10: 91908484-91908484 on Build GRCh38
rs10881994 Chr.10: 91908651-91908651 on Build GRCh38
rs202089453 Chr.10: 91908462-91908462 on Build GRCh38
rs1056475384 Chr.10: 91908498-91908498 on Build GRCh38
rs529040118 Chr.10: 91908547-91908547 on Build GRCh38
rs756528084 Chr.10: 91908454-91908454 on Build GRCh38

View Full Product Details