Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91908596-91909085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFBP3
Entrez Gene ID
143282
Gene Name
fibroblast growth factor binding protein 3
Gene Aliases
C10orf13, FGF-BP3
UniGene
Hs.466120
Gene Chromosome Location
Chr.10: 91906588-91909501 on Build GRCh38
Transcripts
NM_152429.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3441495 c.93G>A p.G31G Chr.10: 91908877-91908877 on Build GRCh38
6663592 c.54_55insCTG p.L18_S19insL Chr.10: 91908915-91908916 on Build GRCh38
3765523 c.319G>A p.A107T Chr.10: 91908651-91908651 on Build GRCh38
5727285 c.26C>T p.S9L Chr.10: 91908944-91908944 on Build GRCh38
5682733 c.74C>T p.A25V Chr.10: 91908896-91908896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760394396 Chr.10: 91908941-91908941 on Build GRCh38
rs935829193 Chr.10: 91908969-91908969 on Build GRCh38
rs541312392 Chr.10: 91908677-91908677 on Build GRCh38
rs945880402 Chr.10: 91908693-91908693 on Build GRCh38
rs751628988 Chr.10: 91908740-91908740 on Build GRCh38
rs768132211 Chr.10: 91908943-91908943 on Build GRCh38
rs546083407 Chr.10: 91908906-91908906 on Build GRCh38
rs773627903 Chr.10: 91908947-91908947 on Build GRCh38
rs868142450 Chr.10: 91908873-91908873 on Build GRCh38
rs11819077 Chr.10: 91908667-91908667 on Build GRCh38
rs202182077 Chr.10: 91908869-91908869 on Build GRCh38
rs757710382 Chr.10: 91908685-91908685 on Build GRCh38
rs368583604 Chr.10: 91908631-91908631 on Build GRCh38
rs774954098 Chr.10: 91908940-91908940 on Build GRCh38
rs770347743 Chr.10: 91908755-91908789 on Build GRCh38
rs896609563 Chr.10: 91908713-91908713 on Build GRCh38
rs967167745 Chr.10: 91908929-91908929 on Build GRCh38
rs559887846 Chr.10: 91908920-91908920 on Build GRCh38
rs11555481 Chr.10: 91908752-91908752 on Build GRCh38
rs935892251 Chr.10: 91908747-91908747 on Build GRCh38
rs867654038 Chr.10: 91908644-91908644 on Build GRCh38
rs903868284 Chr.10: 91908697-91908697 on Build GRCh38
rs1041970538 Chr.10: 91908736-91908736 on Build GRCh38
rs756251241 Chr.10: 91908898-91908898 on Build GRCh38
rs773865761 Chr.10: 91908843-91908843 on Build GRCh38
rs192010535 Chr.10: 91908672-91908672 on Build GRCh38
rs371835191 Chr.10: 91908954-91908954 on Build GRCh38
rs766847271 Chr.10: 91908950-91908950 on Build GRCh38
rs983394739 Chr.10: 91908641-91908641 on Build GRCh38
rs554655505 Chr.10: 91908767-91908767 on Build GRCh38
rs752153652 Chr.10: 91908952-91908952 on Build GRCh38
rs899466971 Chr.10: 91908763-91908763 on Build GRCh38
rs766960625 Chr.10: 91908927-91908927 on Build GRCh38
rs755165527 Chr.10: 91908887-91908887 on Build GRCh38
rs969606470 Chr.10: 91908745-91908745 on Build GRCh38
rs745401747 Chr.10: 91908915-91908916 on Build GRCh38
rs949516389 Chr.10: 91908985-91908985 on Build GRCh38
rs370474127 Chr.10: 91908916-91908918 on Build GRCh38
rs769678936 Chr.10: 91908818-91908818 on Build GRCh38
rs752106546 Chr.10: 91908875-91908875 on Build GRCh38
rs745795679 Chr.10: 91908921-91908921 on Build GRCh38
rs754208602 Chr.10: 91908679-91908679 on Build GRCh38
rs769470835 Chr.10: 91908617-91908617 on Build GRCh38
rs750992710 Chr.10: 91908689-91908689 on Build GRCh38
rs992193902 Chr.10: 91908684-91908684 on Build GRCh38
rs759210551 Chr.10: 91908673-91908673 on Build GRCh38
rs763526872 Chr.10: 91908871-91908871 on Build GRCh38
rs965185435 Chr.10: 91908848-91908848 on Build GRCh38
rs867419584 Chr.10: 91908653-91908653 on Build GRCh38
rs200450389 Chr.10: 91908889-91908889 on Build GRCh38
rs1004373586 Chr.10: 91908868-91908868 on Build GRCh38
rs773810665 Chr.10: 91908664-91908664 on Build GRCh38
rs758787158 Chr.10: 91908703-91908703 on Build GRCh38
rs749280189 Chr.10: 91908825-91908825 on Build GRCh38
rs1044344783 Chr.10: 91908690-91908690 on Build GRCh38
rs935565129 Chr.10: 91908897-91908897 on Build GRCh38
rs1039635491 Chr.10: 91909027-91909027 on Build GRCh38
rs748980225 Chr.10: 91908900-91908900 on Build GRCh38
rs963321743 Chr.10: 91908652-91908652 on Build GRCh38
rs750348568 Chr.10: 91909021-91909021 on Build GRCh38
rs760226116 Chr.10: 91908857-91908857 on Build GRCh38
rs761439216 Chr.10: 91908899-91908899 on Build GRCh38
rs868007366 Chr.10: 91908862-91908862 on Build GRCh38
rs765953507 Chr.10: 91908659-91908659 on Build GRCh38
rs376112133 Chr.10: 91908931-91908932 on Build GRCh38
rs951024225 Chr.10: 91908627-91908627 on Build GRCh38
rs764163214 Chr.10: 91908994-91908994 on Build GRCh38
rs905035584 Chr.10: 91909041-91909041 on Build GRCh38
rs771569550 Chr.10: 91908847-91908847 on Build GRCh38
rs765660624 Chr.10: 91908865-91908865 on Build GRCh38
rs765511010 Chr.10: 91908688-91908688 on Build GRCh38
rs1052522347 Chr.10: 91908704-91908704 on Build GRCh38
rs773126180 Chr.10: 91908828-91908828 on Build GRCh38
rs1014075654 Chr.10: 91908722-91908722 on Build GRCh38
rs775184933 Chr.10: 91908853-91908853 on Build GRCh38
rs1026773205 Chr.10: 91909059-91909059 on Build GRCh38
rs995451670 Chr.10: 91908754-91908754 on Build GRCh38
rs200280119 Chr.10: 91908935-91908935 on Build GRCh38
rs981005188 Chr.10: 91908660-91908660 on Build GRCh38
rs912274448 Chr.10: 91908635-91908635 on Build GRCh38
rs568148727 Chr.10: 91908976-91908976 on Build GRCh38
rs997897695 Chr.10: 91908618-91908618 on Build GRCh38
rs1023492868 Chr.10: 91908731-91908731 on Build GRCh38
rs777399935 Chr.10: 91908822-91908822 on Build GRCh38
rs763287480 Chr.10: 91908948-91908948 on Build GRCh38
rs776982068 Chr.10: 91908647-91908647 on Build GRCh38
rs536091981 Chr.10: 91908890-91908890 on Build GRCh38
rs758981844 Chr.10: 91908846-91908846 on Build GRCh38
rs548449809 Chr.10: 91908963-91908963 on Build GRCh38
rs780202289 Chr.10: 91908732-91908732 on Build GRCh38
rs748365880 Chr.10: 91908817-91908817 on Build GRCh38
rs982453277 Chr.10: 91908798-91908798 on Build GRCh38
rs1045529730 Chr.10: 91908999-91908999 on Build GRCh38
rs115718182 Chr.10: 91909044-91909044 on Build GRCh38
rs10881994 Chr.10: 91908651-91908651 on Build GRCh38
rs534654065 Chr.10: 91908791-91908791 on Build GRCh38
rs543578833 Chr.10: 91908808-91908808 on Build GRCh38

View Full Product Details