Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92050083-92050584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB3
Entrez Gene ID
22849
Gene Name
cytoplasmic polyadenylation element binding protein 3
Gene Aliases
-
UniGene
Hs.131683
Gene Chromosome Location
Chr.10: 92046692-92291118 on Build GRCh38
Transcripts
NM_001178137.1 NM_014912.4 XM_005269630.4 XM_006717714.3 XM_006717715.3 XM_011539514.2 XM_011539515.2 XM_011539517.2 XM_011539519.2 XM_011539520.2 XM_017015927.1

SNP Detail

SNP ID Chromosome Location
rs993409712 Chr.10: 92050158-92050158 on Build GRCh38
rs1055762259 Chr.10: 92050499-92050499 on Build GRCh38
rs921957312 Chr.10: 92050118-92050118 on Build GRCh38
rs539363913 Chr.10: 92050246-92050246 on Build GRCh38
rs940784970 Chr.10: 92050210-92050210 on Build GRCh38
rs938701787 Chr.10: 92050483-92050483 on Build GRCh38
rs915990412 Chr.10: 92050509-92050509 on Build GRCh38
rs924127858 Chr.10: 92050111-92050112 on Build GRCh38
rs567327438 Chr.10: 92050276-92050276 on Build GRCh38
rs1024633491 Chr.10: 92050431-92050431 on Build GRCh38
rs189018022 Chr.10: 92050279-92050279 on Build GRCh38
rs1051760005 Chr.10: 92050151-92050151 on Build GRCh38
rs911999133 Chr.10: 92050169-92050169 on Build GRCh38
rs897778661 Chr.10: 92050113-92050113 on Build GRCh38
rs954346196 Chr.10: 92050179-92050179 on Build GRCh38
rs999242547 Chr.10: 92050484-92050485 on Build GRCh38
rs756184034 Chr.10: 92050112-92050112 on Build GRCh38
rs181769533 Chr.10: 92050516-92050516 on Build GRCh38
rs879326181 Chr.10: 92050522-92050522 on Build GRCh38
rs36105375 Chr.10: 92050537-92050538 on Build GRCh38
rs932009881 Chr.10: 92050151-92050151 on Build GRCh38
rs867786022 Chr.10: 92050385-92050385 on Build GRCh38
rs780708704 Chr.10: 92050475-92050477 on Build GRCh38
rs1011947793 Chr.10: 92050413-92050413 on Build GRCh38
rs148382215 Chr.10: 92050183-92050183 on Build GRCh38
rs1030023754 Chr.10: 92050168-92050168 on Build GRCh38
rs370082311 Chr.10: 92050457-92050457 on Build GRCh38
rs572775594 Chr.10: 92050395-92050395 on Build GRCh38
rs995436004 Chr.10: 92050289-92050290 on Build GRCh38
rs914741206 Chr.10: 92050303-92050303 on Build GRCh38
rs967627947 Chr.10: 92050304-92050304 on Build GRCh38
rs990731255 Chr.10: 92050290-92050290 on Build GRCh38
rs1007197474 Chr.10: 92050212-92050212 on Build GRCh38
rs977630726 Chr.10: 92050308-92050308 on Build GRCh38

View Full Product Details