Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92091695-92092238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB3
Entrez Gene ID
22849
Gene Name
cytoplasmic polyadenylation element binding protein 3
Gene Aliases
-
UniGene
Hs.131683
Gene Chromosome Location
Chr.10: 92046692-92291118 on Build GRCh38
Transcripts
NM_001178137.1 NM_014912.4 XM_005269630.4 XM_006717714.3 XM_006717715.3 XM_011539514.2 XM_011539515.2 XM_011539517.2 XM_011539518.2 XM_011539519.2 XM_011539520.2 XM_017015927.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4529807 c.1643G>A p.R548K Chr.10: 92091874-92091874 on Build GRCh38
7329769 c.1558G>A p.D520N Chr.10: 92091917-92091917 on Build GRCh38
6634544 c.1558G>T p.D520Y Chr.10: 92091917-92091917 on Build GRCh38
6634552 c.1580T>A p.I527N Chr.10: 92091937-92091937 on Build GRCh38
4182218 c.1568T>G p.F523C Chr.10: 92091907-92091907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758449954 Chr.10: 92091987-92091987 on Build GRCh38
rs745472426 Chr.10: 92092152-92092152 on Build GRCh38
rs1020555347 Chr.10: 92092158-92092158 on Build GRCh38
rs183115011 Chr.10: 92092165-92092165 on Build GRCh38
rs767977151 Chr.10: 92091836-92091836 on Build GRCh38
rs936390230 Chr.10: 92092060-92092060 on Build GRCh38
rs369153928 Chr.10: 92091930-92091930 on Build GRCh38
rs373528761 Chr.10: 92091804-92091804 on Build GRCh38
rs1229578 Chr.10: 92091957-92091957 on Build GRCh38
rs1050012788 Chr.10: 92092054-92092054 on Build GRCh38
rs1002893684 Chr.10: 92092191-92092191 on Build GRCh38
rs888804851 Chr.10: 92092212-92092212 on Build GRCh38
rs577416184 Chr.10: 92091849-92091849 on Build GRCh38
rs149011392 Chr.10: 92091778-92091778 on Build GRCh38
rs142967581 Chr.10: 92092132-92092132 on Build GRCh38
rs993640710 Chr.10: 92091916-92091916 on Build GRCh38
rs369829144 Chr.10: 92091798-92091798 on Build GRCh38
rs35476882 Chr.10: 92092004-92092005 on Build GRCh38
rs750240026 Chr.10: 92091958-92091958 on Build GRCh38
rs542262641 Chr.10: 92092073-92092073 on Build GRCh38
rs971386429 Chr.10: 92091721-92091721 on Build GRCh38
rs959019420 Chr.10: 92091734-92091734 on Build GRCh38
rs1034440637 Chr.10: 92091728-92091728 on Build GRCh38
rs374710766 Chr.10: 92091808-92091808 on Build GRCh38
rs559834389 Chr.10: 92092056-92092056 on Build GRCh38
rs371398249 Chr.10: 92091788-92091788 on Build GRCh38
rs775592518 Chr.10: 92091841-92091841 on Build GRCh38
rs768405541 Chr.10: 92091796-92091796 on Build GRCh38
rs561130149 Chr.10: 92092088-92092088 on Build GRCh38
rs532375852 Chr.10: 92092189-92092189 on Build GRCh38
rs189777891 Chr.10: 92092101-92092101 on Build GRCh38
rs759894214 Chr.10: 92091978-92091979 on Build GRCh38
rs552208060 Chr.10: 92092025-92092025 on Build GRCh38
rs559922936 Chr.10: 92091793-92091793 on Build GRCh38
rs1052268258 Chr.10: 92092116-92092116 on Build GRCh38
rs760097953 Chr.10: 92091819-92091819 on Build GRCh38
rs981405894 Chr.10: 92091723-92091723 on Build GRCh38
rs971403588 Chr.10: 92092051-92092051 on Build GRCh38
rs774855993 Chr.10: 92091816-92091816 on Build GRCh38
rs971951296 Chr.10: 92091845-92091845 on Build GRCh38
rs528470811 Chr.10: 92092064-92092064 on Build GRCh38
rs780119787 Chr.10: 92091992-92091992 on Build GRCh38
rs984652990 Chr.10: 92091755-92091755 on Build GRCh38
rs764406144 Chr.10: 92091864-92091864 on Build GRCh38
rs376994406 Chr.10: 92091986-92091986 on Build GRCh38
rs907413525 Chr.10: 92092162-92092162 on Build GRCh38
rs1050322772 Chr.10: 92092166-92092166 on Build GRCh38
rs184971435 Chr.10: 92091764-92091764 on Build GRCh38
rs888781664 Chr.10: 92091756-92091756 on Build GRCh38
rs746489226 Chr.10: 92091795-92091795 on Build GRCh38
rs753962299 Chr.10: 92091784-92091786 on Build GRCh38

View Full Product Details