Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95194120-95194631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM6
Entrez Gene ID
142827
Gene Name
acyl-CoA synthetase medium-chain family member 6
Gene Aliases
C10orf129, bA310E22.3
UniGene
Hs.134229
Gene Chromosome Location
Chr.10: 95194200-95228929 on Build GRCh38
Transcripts
NM_207321.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1644445 c.125T>A p.V42D Chr.10: 95194610-95194610 on Build GRCh38
1627695 c.24C>T p.F8F Chr.10: 95194509-95194509 on Build GRCh38
466124 c.68A>T p.Y23F Chr.10: 95194553-95194553 on Build GRCh38
1350015 c.63C>A p.C21* Chr.10: 95194548-95194548 on Build GRCh38
4169239 c.34C>T p.R12W Chr.10: 95194519-95194519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052854989 Chr.10: 95194434-95194434 on Build GRCh38
rs374342058 Chr.10: 95194197-95194197 on Build GRCh38
rs1034485244 Chr.10: 95194275-95194276 on Build GRCh38
rs757510884 Chr.10: 95194522-95194522 on Build GRCh38
rs747933826 Chr.10: 95194180-95194180 on Build GRCh38
rs763355381 Chr.10: 95194451-95194451 on Build GRCh38
rs913941647 Chr.10: 95194469-95194469 on Build GRCh38
rs545166181 Chr.10: 95194323-95194323 on Build GRCh38
rs554010135 Chr.10: 95194145-95194145 on Build GRCh38
rs567595841 Chr.10: 95194503-95194503 on Build GRCh38
rs993127917 Chr.10: 95194315-95194315 on Build GRCh38
rs577209199 Chr.10: 95194196-95194196 on Build GRCh38
rs888327837 Chr.10: 95194565-95194565 on Build GRCh38
rs187035186 Chr.10: 95194480-95194480 on Build GRCh38
rs965091573 Chr.10: 95194282-95194282 on Build GRCh38
rs866565580 Chr.10: 95194544-95194544 on Build GRCh38
rs531871604 Chr.10: 95194318-95194318 on Build GRCh38
rs11188225 Chr.10: 95194603-95194603 on Build GRCh38
rs542639443 Chr.10: 95194309-95194309 on Build GRCh38
rs777620019 Chr.10: 95194271-95194272 on Build GRCh38
rs768073933 Chr.10: 95194574-95194575 on Build GRCh38
rs775949612 Chr.10: 95194170-95194170 on Build GRCh38
rs952681336 Chr.10: 95194204-95194204 on Build GRCh38
rs778765969 Chr.10: 95194160-95194160 on Build GRCh38
rs926052736 Chr.10: 95194455-95194455 on Build GRCh38
rs1004249487 Chr.10: 95194218-95194218 on Build GRCh38
rs936945258 Chr.10: 95194423-95194423 on Build GRCh38
rs886141301 Chr.10: 95194189-95194189 on Build GRCh38
rs774048200 Chr.10: 95194566-95194566 on Build GRCh38
rs762693798 Chr.10: 95194465-95194465 on Build GRCh38
rs547739620 Chr.10: 95194490-95194490 on Build GRCh38
rs374138819 Chr.10: 95194518-95194518 on Build GRCh38
rs925486961 Chr.10: 95194353-95194353 on Build GRCh38
rs1004167462 Chr.10: 95194580-95194580 on Build GRCh38
rs867503196 Chr.10: 95194496-95194496 on Build GRCh38
rs562723715 Chr.10: 95194316-95194316 on Build GRCh38
rs963102299 Chr.10: 95194200-95194200 on Build GRCh38
rs149290267 Chr.10: 95194327-95194327 on Build GRCh38
rs1015268155 Chr.10: 95194242-95194242 on Build GRCh38
rs591157 Chr.10: 95194541-95194541 on Build GRCh38
rs937820865 Chr.10: 95194532-95194532 on Build GRCh38
rs779361696 Chr.10: 95194534-95194534 on Build GRCh38
rs530002609 Chr.10: 95194520-95194520 on Build GRCh38
rs201256506 Chr.10: 95194495-95194495 on Build GRCh38
rs139790584 Chr.10: 95194283-95194283 on Build GRCh38

View Full Product Details