Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95937857-95938362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf131
Entrez Gene ID
100127889
Gene Name
chromosome 10 open reading frame 131
Gene Aliases
bA690P14.3
UniGene
Hs.719076
Gene Chromosome Location
Chr.10: 95907965-95938658 on Build GRCh38
Transcripts
NM_001130446.2 NR_038444.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001130446.2 NR_038444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
921844 c.529T>C p.*177Q Chr.10: 95938207-95938207 on Build GRCh38
6457508 c.400A>G p.K134E Chr.10: 95938078-95938078 on Build GRCh38
1350052 c.314G>A p.R105H Chr.10: 95938004-95938004 on Build GRCh38
5826861 c.494C>A p.S165Y Chr.10: 95938184-95938184 on Build GRCh38
1350054 c.494C>A p.S165Y Chr.10: 95938184-95938184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778478419 Chr.10: 95938255-95938255 on Build GRCh38
rs780679716 Chr.10: 95938235-95938235 on Build GRCh38
rs772316083 Chr.10: 95937993-95938000 on Build GRCh38
rs1006289748 Chr.10: 95938293-95938293 on Build GRCh38
rs924320099 Chr.10: 95938081-95938081 on Build GRCh38
rs756649743 Chr.10: 95938197-95938197 on Build GRCh38
rs774373777 Chr.10: 95938004-95938004 on Build GRCh38
rs749809899 Chr.10: 95938257-95938257 on Build GRCh38
rs893005851 Chr.10: 95937894-95937894 on Build GRCh38
rs907127057 Chr.10: 95938199-95938199 on Build GRCh38
rs751846999 Chr.10: 95937953-95937953 on Build GRCh38
rs907177141 Chr.10: 95937968-95937968 on Build GRCh38
rs376030619 Chr.10: 95938054-95938054 on Build GRCh38
rs1047653052 Chr.10: 95937962-95937962 on Build GRCh38
rs562738035 Chr.10: 95938080-95938080 on Build GRCh38
rs1000939894 Chr.10: 95938234-95938234 on Build GRCh38
rs527454488 Chr.10: 95938329-95938329 on Build GRCh38
rs542962923 Chr.10: 95938066-95938066 on Build GRCh38
rs765085506 Chr.10: 95938078-95938078 on Build GRCh38
rs78719716 Chr.10: 95938006-95938006 on Build GRCh38
rs565103741 Chr.10: 95938308-95938308 on Build GRCh38
rs545308434 Chr.10: 95938260-95938260 on Build GRCh38
rs950773470 Chr.10: 95937988-95937988 on Build GRCh38
rs1026369988 Chr.10: 95937987-95937987 on Build GRCh38
rs923156395 Chr.10: 95937942-95937942 on Build GRCh38
rs934456601 Chr.10: 95937890-95937890 on Build GRCh38
rs758090505 Chr.10: 95938102-95938102 on Build GRCh38
rs188396436 Chr.10: 95938141-95938141 on Build GRCh38
rs770469744 Chr.10: 95937976-95937976 on Build GRCh38
rs192638260 Chr.10: 95938335-95938335 on Build GRCh38
rs1013237026 Chr.10: 95937935-95937935 on Build GRCh38
rs886465842 Chr.10: 95938273-95938273 on Build GRCh38
rs948602178 Chr.10: 95938146-95938146 on Build GRCh38
rs984906615 Chr.10: 95938007-95938007 on Build GRCh38
rs756679901 Chr.10: 95938227-95938227 on Build GRCh38
rs573962226 Chr.10: 95938042-95938042 on Build GRCh38
rs745372380 Chr.10: 95938117-95938117 on Build GRCh38
rs1054161289 Chr.10: 95938123-95938123 on Build GRCh38
rs182534542 Chr.10: 95937892-95937892 on Build GRCh38
rs554206984 Chr.10: 95937985-95937985 on Build GRCh38
rs750002281 Chr.10: 95938100-95938100 on Build GRCh38
rs771167901 Chr.10: 95937989-95937989 on Build GRCh38
rs780984520 Chr.10: 95938236-95938236 on Build GRCh38
rs751152323 Chr.10: 95938151-95938151 on Build GRCh38
rs976314519 Chr.10: 95937931-95937931 on Build GRCh38
rs753655014 Chr.10: 95938065-95938065 on Build GRCh38
rs1022906984 Chr.10: 95937963-95937963 on Build GRCh38
rs1021129054 Chr.10: 95938174-95938174 on Build GRCh38
rs772094765 Chr.10: 95938037-95938037 on Build GRCh38
rs748615113 Chr.10: 95937958-95937958 on Build GRCh38
rs868533554 Chr.10: 95938089-95938089 on Build GRCh38
rs760577816 Chr.10: 95938049-95938049 on Build GRCh38
rs986431541 Chr.10: 95938072-95938072 on Build GRCh38
rs773678051 Chr.10: 95937982-95937982 on Build GRCh38
rs759838397 Chr.10: 95938016-95938016 on Build GRCh38
rs1009704456 Chr.10: 95938118-95938121 on Build GRCh38
rs561152050 Chr.10: 95937893-95937893 on Build GRCh38

View Full Product Details