Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97333997-97334491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRAT2
Entrez Gene ID
23401
Gene Name
frequently rearranged in advanced T-cell lymphomas 2
Gene Aliases
-
UniGene
Hs.140720
Gene Chromosome Location
Chr.10: 97332497-97334701 on Build GRCh38
Transcripts
NM_012083.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6581227 c.554C>T p.S185L Chr.10: 97334019-97334019 on Build GRCh38
6299038 c.512G>A p.G171D Chr.10: 97334061-97334061 on Build GRCh38
6551216 c.235G>A p.A79T Chr.10: 97334338-97334338 on Build GRCh38
6240644 c.99G>T p.T33T Chr.10: 97334474-97334474 on Build GRCh38
6530916 c.529C>G p.R177G Chr.10: 97334044-97334044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760082910 Chr.10: 97334377-97334377 on Build GRCh38
rs909251895 Chr.10: 97334304-97334304 on Build GRCh38
rs768851925 Chr.10: 97334067-97334067 on Build GRCh38
rs768870341 Chr.10: 97334159-97334159 on Build GRCh38
rs752291854 Chr.10: 97334372-97334372 on Build GRCh38
rs750887635 Chr.10: 97334414-97334414 on Build GRCh38
rs968355048 Chr.10: 97334189-97334189 on Build GRCh38
rs566685296 Chr.10: 97334096-97334096 on Build GRCh38
rs775834984 Chr.10: 97334064-97334064 on Build GRCh38
rs565101660 Chr.10: 97334465-97334465 on Build GRCh38
rs773016444 Chr.10: 97334086-97334086 on Build GRCh38
rs766959091 Chr.10: 97334112-97334112 on Build GRCh38
rs779288381 Chr.10: 97334059-97334059 on Build GRCh38
rs955957042 Chr.10: 97334153-97334153 on Build GRCh38
rs754474171 Chr.10: 97334018-97334018 on Build GRCh38
rs1025220003 Chr.10: 97334109-97334109 on Build GRCh38
rs751853572 Chr.10: 97334473-97334473 on Build GRCh38
rs577297633 Chr.10: 97334207-97334207 on Build GRCh38
rs902350502 Chr.10: 97334043-97334043 on Build GRCh38
rs181573119 Chr.10: 97334326-97334326 on Build GRCh38
rs779258474 Chr.10: 97334139-97334139 on Build GRCh38
rs3740515 Chr.10: 97334401-97334401 on Build GRCh38
rs12767255 Chr.10: 97334128-97334128 on Build GRCh38
rs746109134 Chr.10: 97334061-97334061 on Build GRCh38
rs929404173 Chr.10: 97334272-97334272 on Build GRCh38
rs758678704 Chr.10: 97334149-97334149 on Build GRCh38
rs761751988 Chr.10: 97334073-97334073 on Build GRCh38
rs1002726820 Chr.10: 97334336-97334336 on Build GRCh38
rs544739077 Chr.10: 97334208-97334208 on Build GRCh38
rs780487854 Chr.10: 97334150-97334150 on Build GRCh38
rs369870649 Chr.10: 97334462-97334462 on Build GRCh38
rs561034462 Chr.10: 97334367-97334367 on Build GRCh38
rs942002233 Chr.10: 97334305-97334305 on Build GRCh38
rs1003082835 Chr.10: 97334144-97334144 on Build GRCh38
rs781372531 Chr.10: 97334162-97334162 on Build GRCh38
rs749732602 Chr.10: 97334037-97334037 on Build GRCh38
rs556881302 Chr.10: 97334216-97334216 on Build GRCh38
rs968364900 Chr.10: 97334409-97334409 on Build GRCh38
rs776714683 Chr.10: 97334071-97334071 on Build GRCh38
rs923394153 Chr.10: 97334218-97334218 on Build GRCh38
rs751211779 Chr.10: 97334094-97334094 on Build GRCh38
rs769806710 Chr.10: 97334195-97334195 on Build GRCh38
rs905733186 Chr.10: 97334333-97334333 on Build GRCh38
rs976348389 Chr.10: 97334472-97334472 on Build GRCh38
rs757580877 Chr.10: 97334041-97334041 on Build GRCh38
rs770649715 Chr.10: 97334227-97334227 on Build GRCh38
rs542858899 Chr.10: 97334363-97334363 on Build GRCh38
rs766102767 Chr.10: 97334092-97334092 on Build GRCh38
rs371878873 Chr.10: 97334066-97334066 on Build GRCh38
rs746293757 Chr.10: 97334143-97334143 on Build GRCh38
rs778321683 Chr.10: 97334026-97334026 on Build GRCh38
rs558774685 Chr.10: 97334185-97334185 on Build GRCh38
rs754369707 Chr.10: 97334126-97334126 on Build GRCh38
rs772380383 Chr.10: 97334063-97334063 on Build GRCh38
rs528434717 Chr.10: 97334407-97334407 on Build GRCh38
rs1047823688 Chr.10: 97334292-97334292 on Build GRCh38
rs901885500 Chr.10: 97334023-97334023 on Build GRCh38
rs979690798 Chr.10: 97334192-97334192 on Build GRCh38
rs758792803 Chr.10: 97334435-97334435 on Build GRCh38
rs905727833 Chr.10: 97334137-97334137 on Build GRCh38
rs764959921 Chr.10: 97334074-97334074 on Build GRCh38
rs554969892 Chr.10: 97334090-97334090 on Build GRCh38
rs534119007 Chr.10: 97334155-97334155 on Build GRCh38
rs1009981867 Chr.10: 97334388-97334388 on Build GRCh38

View Full Product Details