Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97599985-97600494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOGA1
Entrez Gene ID
112817
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases
C10orf65, DHDPS2, DHDPSL, HP3, NPL2
UniGene
Hs.180346
Gene Chromosome Location
Chr.10: 97584345-97612802 on Build GRCh38
Transcripts
NM_001134670.1 NM_138413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3442434 c.646G>A p.D216N Chr.10: 97600109-97600109 on Build GRCh38
5999160 c.692A>G p.Y231C Chr.10: 97600155-97600155 on Build GRCh38
3442433 c.636C>T p.T212T Chr.10: 97600099-97600099 on Build GRCh38
1162922 c.669G>A p.S223S Chr.10: 97600132-97600132 on Build GRCh38
7261078 c.662C>T p.A221V Chr.10: 97600125-97600125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553221586 Chr.10: 97600240-97600240 on Build GRCh38
rs577932546 Chr.10: 97600315-97600315 on Build GRCh38
rs758122412 Chr.10: 97600096-97600096 on Build GRCh38
rs199604936 Chr.10: 97600023-97600023 on Build GRCh38
rs985964047 Chr.10: 97600229-97600229 on Build GRCh38
rs779492256 Chr.10: 97600097-97600097 on Build GRCh38
rs576082301 Chr.10: 97600446-97600446 on Build GRCh38
rs185803104 Chr.10: 97600168-97600168 on Build GRCh38
rs771334502 Chr.10: 97600132-97600132 on Build GRCh38
rs769361123 Chr.10: 97600130-97600131 on Build GRCh38
rs189424938 Chr.10: 97600438-97600438 on Build GRCh38
rs142766113 Chr.10: 97600131-97600131 on Build GRCh38
rs1040324329 Chr.10: 97600255-97600255 on Build GRCh38
rs1027757723 Chr.10: 97600382-97600382 on Build GRCh38
rs774579806 Chr.10: 97600026-97600026 on Build GRCh38
rs375091787 Chr.10: 97600172-97600172 on Build GRCh38
rs778937599 Chr.10: 97600038-97600038 on Build GRCh38
rs753833846 Chr.10: 97600178-97600178 on Build GRCh38
rs753648051 Chr.10: 97600055-97600055 on Build GRCh38
rs759819900 Chr.10: 97600161-97600161 on Build GRCh38
rs1020790167 Chr.10: 97600033-97600033 on Build GRCh38
rs564444742 Chr.10: 97600317-97600317 on Build GRCh38
rs746545018 Chr.10: 97600098-97600098 on Build GRCh38
rs1024511389 Chr.10: 97600334-97600334 on Build GRCh38
rs752816955 Chr.10: 97600037-97600037 on Build GRCh38
rs1042137746 Chr.10: 97600275-97600275 on Build GRCh38
rs368607136 Chr.10: 97600035-97600035 on Build GRCh38
rs2297643 Chr.10: 97600230-97600230 on Build GRCh38
rs543192255 Chr.10: 97600452-97600452 on Build GRCh38
rs551638828 Chr.10: 97600034-97600034 on Build GRCh38
rs181077522 Chr.10: 97600461-97600461 on Build GRCh38
rs776660476 Chr.10: 97600029-97600030 on Build GRCh38
rs1003854237 Chr.10: 97600318-97600318 on Build GRCh38
rs1000706512 Chr.10: 97600301-97600301 on Build GRCh38
rs967028585 Chr.10: 97600060-97600060 on Build GRCh38
rs1010696394 Chr.10: 97600313-97600313 on Build GRCh38
rs774667887 Chr.10: 97600139-97600139 on Build GRCh38
rs990830655 Chr.10: 97600165-97600165 on Build GRCh38
rs375002620 Chr.10: 97600040-97600040 on Build GRCh38
rs750145949 Chr.10: 97600094-97600094 on Build GRCh38
rs1034451669 Chr.10: 97600307-97600307 on Build GRCh38
rs765159808 Chr.10: 97600210-97600210 on Build GRCh38
rs957797762 Chr.10: 97600009-97600009 on Build GRCh38
rs1027694505 Chr.10: 97600406-97600406 on Build GRCh38
rs775790008 Chr.10: 97600258-97600258 on Build GRCh38
rs771246710 Chr.10: 97600025-97600025 on Build GRCh38
rs764104379 Chr.10: 97600053-97600053 on Build GRCh38
rs781419688 Chr.10: 97600218-97600218 on Build GRCh38
rs775016553 Chr.10: 97600193-97600201 on Build GRCh38
rs1014419641 Chr.10: 97600316-97600316 on Build GRCh38
rs761816106 Chr.10: 97600193-97600193 on Build GRCh38
rs757147223 Chr.10: 97600057-97600057 on Build GRCh38
rs778797375 Chr.10: 97600086-97600086 on Build GRCh38
rs367698800 Chr.10: 97600041-97600041 on Build GRCh38
rs187274275 Chr.10: 97600463-97600463 on Build GRCh38
rs762722138 Chr.10: 97600214-97600214 on Build GRCh38
rs73332768 Chr.10: 97600396-97600396 on Build GRCh38
rs962304321 Chr.10: 97600343-97600343 on Build GRCh38
rs534699077 Chr.10: 97600239-97600239 on Build GRCh38
rs1034904540 Chr.10: 97600394-97600394 on Build GRCh38
rs767987177 Chr.10: 97600105-97600105 on Build GRCh38
rs778377512 Chr.10: 97600129-97600129 on Build GRCh38
rs904586074 Chr.10: 97600280-97600280 on Build GRCh38
rs990778431 Chr.10: 97600397-97600397 on Build GRCh38
rs546057563 Chr.10: 97600353-97600355 on Build GRCh38
rs57616820 Chr.10: 97600170-97600170 on Build GRCh38
rs749695055 Chr.10: 97600266-97600266 on Build GRCh38
rs750233813 Chr.10: 97600213-97600213 on Build GRCh38
rs371213768 Chr.10: 97600036-97600036 on Build GRCh38
rs760812475 Chr.10: 97600171-97600171 on Build GRCh38
rs971743987 Chr.10: 97600354-97600354 on Build GRCh38
rs759146945 Chr.10: 97600110-97600111 on Build GRCh38

View Full Product Details