Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 98456811-98457305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPSE2
Entrez Gene ID
60495
Gene Name
heparanase 2 (inactive)
Gene Aliases
HPA2, HPR2, UFS, UFS1
UniGene
Hs.500750
Gene Chromosome Location
Chr.10: 98457077-99235875 on Build GRCh38
Transcripts
NM_001166244.1 NM_001166245.1 NM_001166246.1 NM_021828.4 XM_006717937.2 XM_011540029.1 XM_011540030.1 XM_011540031.2 XM_011540033.2 XM_017016497.1 XM_017016498.1

SNP Detail

SNP ID Chromosome Location
rs529354276 Chr.10: 98457106-98457106 on Build GRCh38
rs765699994 Chr.10: 98457127-98457127 on Build GRCh38
rs4375377 Chr.10: 98457048-98457048 on Build GRCh38
rs566919198 Chr.10: 98457247-98457247 on Build GRCh38
rs907456899 Chr.10: 98457097-98457097 on Build GRCh38
rs1030622899 Chr.10: 98457273-98457273 on Build GRCh38
rs753889413 Chr.10: 98457073-98457073 on Build GRCh38
rs1006241930 Chr.10: 98457137-98457137 on Build GRCh38
rs996005310 Chr.10: 98457260-98457260 on Build GRCh38
rs4611143 Chr.10: 98457141-98457141 on Build GRCh38
rs923658056 Chr.10: 98456966-98456966 on Build GRCh38
rs964535812 Chr.10: 98456878-98456878 on Build GRCh38
rs12221117 Chr.10: 98456912-98456912 on Build GRCh38
rs989529022 Chr.10: 98457233-98457233 on Build GRCh38
rs974618264 Chr.10: 98456891-98456891 on Build GRCh38
rs75469488 Chr.10: 98457216-98457216 on Build GRCh38
rs755470044 Chr.10: 98457110-98457110 on Build GRCh38
rs143986616 Chr.10: 98457207-98457208 on Build GRCh38
rs1044668902 Chr.10: 98457062-98457062 on Build GRCh38
rs1027115717 Chr.10: 98457111-98457111 on Build GRCh38
rs894256965 Chr.10: 98457076-98457076 on Build GRCh38
rs936453467 Chr.10: 98456982-98456982 on Build GRCh38
rs4598598 Chr.10: 98457060-98457060 on Build GRCh38
rs12221115 Chr.10: 98456902-98456902 on Build GRCh38
rs564813774 Chr.10: 98456967-98456967 on Build GRCh38
rs886530971 Chr.10: 98456869-98456869 on Build GRCh38
rs554370970 Chr.10: 98456876-98456876 on Build GRCh38
rs78986047 Chr.10: 98457217-98457217 on Build GRCh38
rs1005978494 Chr.10: 98456872-98456872 on Build GRCh38
rs12221114 Chr.10: 98456849-98456849 on Build GRCh38
rs886497485 Chr.10: 98457136-98457136 on Build GRCh38
rs900334979 Chr.10: 98457187-98457187 on Build GRCh38
rs995669869 Chr.10: 98457087-98457087 on Build GRCh38
rs954524082 Chr.10: 98456910-98456910 on Build GRCh38
rs887280603 Chr.10: 98457118-98457118 on Build GRCh38
rs906835169 Chr.10: 98457025-98457025 on Build GRCh38
rs4611142 Chr.10: 98457024-98457024 on Build GRCh38

View Full Product Details