Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101525928-101526441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTRC
Entrez Gene ID
8945
Gene Name
beta-transducin repeat containing E3 ubiquitin protein ligase
Gene Aliases
BETA-TRCP, FBW1A, FBXW1, FBXW1A, FWD1, bTrCP, bTrCP1, betaTrCP
UniGene
Hs.643802
Gene Chromosome Location
Chr.10: 101353808-101557321 on Build GRCh38
Transcripts
NM_001256856.1 NM_003939.4 NM_033637.3 XM_006718054.2 XM_011540320.2 XM_017016870.1 XM_017016871.1 XM_017016872.1 XM_017016873.1 XM_017016874.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4746028 c.710C>G p.S237C Chr.10: 101526166-101526166 on Build GRCh38
1158896 c.694A>G p.M232V Chr.10: 101526150-101526150 on Build GRCh38
1627284 c.641A>G p.E214G Chr.10: 101526097-101526097 on Build GRCh38
6245922 c.591G>T p.L197L Chr.10: 101526047-101526047 on Build GRCh38
7247657 c.687C>T p.I229I Chr.10: 101526143-101526143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930417105 Chr.10: 101526316-101526316 on Build GRCh38
rs567133115 Chr.10: 101526254-101526254 on Build GRCh38
rs1029215678 Chr.10: 101526113-101526113 on Build GRCh38
rs149903447 Chr.10: 101526190-101526190 on Build GRCh38
rs777223538 Chr.10: 101526038-101526038 on Build GRCh38
rs746460151 Chr.10: 101526205-101526205 on Build GRCh38
rs371000178 Chr.10: 101526165-101526165 on Build GRCh38
rs991677905 Chr.10: 101526289-101526289 on Build GRCh38
rs769908228 Chr.10: 101526072-101526072 on Build GRCh38
rs984529653 Chr.10: 101526356-101526356 on Build GRCh38
rs538998223 Chr.10: 101526271-101526271 on Build GRCh38
rs764456335 Chr.10: 101525967-101525967 on Build GRCh38
rs780579198 Chr.10: 101526157-101526157 on Build GRCh38
rs575405884 Chr.10: 101526280-101526280 on Build GRCh38
rs757745628 Chr.10: 101526338-101526339 on Build GRCh38
rs552991186 Chr.10: 101526182-101526182 on Build GRCh38
rs759367212 Chr.10: 101526061-101526061 on Build GRCh38
rs912145452 Chr.10: 101526408-101526408 on Build GRCh38
rs552921727 Chr.10: 101526349-101526349 on Build GRCh38
rs771540911 Chr.10: 101526022-101526022 on Build GRCh38
rs764295137 Chr.10: 101526085-101526085 on Build GRCh38
rs762828910 Chr.10: 101526382-101526385 on Build GRCh38
rs986120839 Chr.10: 101526404-101526404 on Build GRCh38
rs761948466 Chr.10: 101526106-101526106 on Build GRCh38
rs973874284 Chr.10: 101526341-101526341 on Build GRCh38
rs763109497 Chr.10: 101525966-101525966 on Build GRCh38
rs747705989 Chr.10: 101526016-101526016 on Build GRCh38
rs904832037 Chr.10: 101526228-101526228 on Build GRCh38
rs944996211 Chr.10: 101526412-101526412 on Build GRCh38
rs756647532 Chr.10: 101526002-101526002 on Build GRCh38
rs751796602 Chr.10: 101525977-101525977 on Build GRCh38
rs755428307 Chr.10: 101526180-101526180 on Build GRCh38
rs770557078 Chr.10: 101526050-101526050 on Build GRCh38
rs757456144 Chr.10: 101525986-101525986 on Build GRCh38
rs368671101 Chr.10: 101526115-101526115 on Build GRCh38
rs781427499 Chr.10: 101525988-101525988 on Build GRCh38
rs745696423 Chr.10: 101526230-101526230 on Build GRCh38
rs373809510 Chr.10: 101526229-101526229 on Build GRCh38
rs542449919 Chr.10: 101526352-101526352 on Build GRCh38
rs777995345 Chr.10: 101526294-101526294 on Build GRCh38
rs760129765 Chr.10: 101526080-101526080 on Build GRCh38
rs944855506 Chr.10: 101525996-101525996 on Build GRCh38
rs751603535 Chr.10: 101526096-101526096 on Build GRCh38
rs920046547 Chr.10: 101526302-101526302 on Build GRCh38
rs931965311 Chr.10: 101526353-101526353 on Build GRCh38
rs374149155 Chr.10: 101526073-101526073 on Build GRCh38
rs1015403067 Chr.10: 101526282-101526282 on Build GRCh38
rs756527898 Chr.10: 101526224-101526224 on Build GRCh38
rs17767748 Chr.10: 101526143-101526143 on Build GRCh38
rs981689959 Chr.10: 101526257-101526257 on Build GRCh38
rs756450284 Chr.10: 101526144-101526144 on Build GRCh38
rs750735634 Chr.10: 101525999-101525999 on Build GRCh38
rs577533940 Chr.10: 101526221-101526221 on Build GRCh38
rs867755281 Chr.10: 101526130-101526130 on Build GRCh38
rs151289382 Chr.10: 101526042-101526042 on Build GRCh38
rs370875449 Chr.10: 101526049-101526049 on Build GRCh38
rs377123253 Chr.10: 101526005-101526005 on Build GRCh38
rs776292989 Chr.10: 101526057-101526057 on Build GRCh38
rs556929371 Chr.10: 101526274-101526274 on Build GRCh38

View Full Product Details