Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101775371-101775881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF8
Entrez Gene ID
2253
Gene Name
fibroblast growth factor 8
Gene Aliases
AIGF, FGF-8, HBGF-8, HH6, KAL6
UniGene
Hs.57710
Gene Chromosome Location
Chr.10: 101770130-101780369 on Build GRCh38
Transcripts
NM_001206389.1 NM_006119.4 NM_033163.3 NM_033164.3 NM_033165.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6372486 c.35T>C p.L12P Chr.10: 101775774-101775774 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs950465740 Chr.10: 101775644-101775644 on Build GRCh38
rs866678020 Chr.10: 101775762-101775762 on Build GRCh38
rs908863258 Chr.10: 101775668-101775668 on Build GRCh38
rs1023174578 Chr.10: 101775536-101775536 on Build GRCh38
rs934746020 Chr.10: 101775806-101775806 on Build GRCh38
rs921814082 Chr.10: 101775750-101775750 on Build GRCh38
rs756492101 Chr.10: 101775702-101775702 on Build GRCh38
rs567972135 Chr.10: 101775826-101775826 on Build GRCh38
rs891177630 Chr.10: 101775431-101775431 on Build GRCh38
rs548378720 Chr.10: 101775397-101775397 on Build GRCh38
rs184829568 Chr.10: 101775423-101775423 on Build GRCh38
rs756541067 Chr.10: 101775728-101775728 on Build GRCh38
rs3218227 Chr.10: 101775706-101775706 on Build GRCh38
rs961939072 Chr.10: 101775598-101775598 on Build GRCh38
rs987293051 Chr.10: 101775775-101775775 on Build GRCh38
rs538019672 Chr.10: 101775662-101775662 on Build GRCh38
rs999275082 Chr.10: 101775438-101775438 on Build GRCh38
rs933158155 Chr.10: 101775755-101775755 on Build GRCh38
rs570874523 Chr.10: 101775648-101775648 on Build GRCh38
rs1053084734 Chr.10: 101775442-101775442 on Build GRCh38
rs535351281 Chr.10: 101775858-101775858 on Build GRCh38
rs888548294 Chr.10: 101775824-101775824 on Build GRCh38
rs1006481789 Chr.10: 101775543-101775543 on Build GRCh38
rs764086706 Chr.10: 101775674-101775674 on Build GRCh38
rs1011825076 Chr.10: 101775507-101775507 on Build GRCh38
rs953247735 Chr.10: 101775609-101775609 on Build GRCh38
rs749605885 Chr.10: 101775781-101775781 on Build GRCh38
rs893418713 Chr.10: 101775448-101775448 on Build GRCh38
rs137852659 Chr.10: 101775769-101775769 on Build GRCh38
rs953104114 Chr.10: 101775859-101775859 on Build GRCh38
rs759091259 Chr.10: 101775844-101775844 on Build GRCh38
rs1029990029 Chr.10: 101775606-101775606 on Build GRCh38
rs897739202 Chr.10: 101775841-101775842 on Build GRCh38
rs948801248 Chr.10: 101775771-101775771 on Build GRCh38
rs894555606 Chr.10: 101775471-101775471 on Build GRCh38
rs776389277 Chr.10: 101775841-101775841 on Build GRCh38
rs748640762 Chr.10: 101775837-101775838 on Build GRCh38
rs769442062 Chr.10: 101775857-101775857 on Build GRCh38
rs915937495 Chr.10: 101775726-101775726 on Build GRCh38
rs757356983 Chr.10: 101775701-101775701 on Build GRCh38
rs1045282821 Chr.10: 101775533-101775533 on Build GRCh38
rs985923351 Chr.10: 101775626-101775626 on Build GRCh38
rs112280066 Chr.10: 101775597-101775597 on Build GRCh38
rs780259162 Chr.10: 101775772-101775772 on Build GRCh38
rs1048444141 Chr.10: 101775797-101775797 on Build GRCh38
rs750604378 Chr.10: 101775727-101775727 on Build GRCh38
rs779323337 Chr.10: 101775829-101775829 on Build GRCh38
rs1041943009 Chr.10: 101775412-101775412 on Build GRCh38
rs746533135 Chr.10: 101775832-101775832 on Build GRCh38
rs1014771957 Chr.10: 101775576-101775576 on Build GRCh38
rs751694549 Chr.10: 101775692-101775692 on Build GRCh38
rs914714094 Chr.10: 101775660-101775660 on Build GRCh38
rs1057150717 Chr.10: 101775723-101775723 on Build GRCh38
rs770331509 Chr.10: 101775838-101775838 on Build GRCh38
rs997105525 Chr.10: 101775599-101775599 on Build GRCh38
rs1035854168 Chr.10: 101775602-101775602 on Build GRCh38
rs932785003 Chr.10: 101775413-101775413 on Build GRCh38
rs552627920 Chr.10: 101775542-101775542 on Build GRCh38
rs963292794 Chr.10: 101775687-101775687 on Build GRCh38

View Full Product Details