Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101827554-101828066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNIP2
Entrez Gene ID
30819
Gene Name
potassium voltage-gated channel interacting protein 2
Gene Aliases
KCHIP2
UniGene
Hs.97044
Gene Chromosome Location
Chr.10: 101825971-101843920 on Build GRCh38
Transcripts
NM_014591.4 NM_173191.2 NM_173192.2 NM_173193.2 NM_173194.2 NM_173195.2 NM_173197.2 XM_005269729.2 XM_005269730.2 XM_006717812.2 XM_011539731.2 XM_017016161.1 NR_045118.1

Gene Symbol
KCNIP2-AS1
Entrez Gene ID
100289509
Gene Name
KCNIP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.568881
Gene Chromosome Location
Chr.10: 101819068-101828889 on Build GRCh38
Transcripts
NM_014591.4 NM_173191.2 NM_173192.2 NM_173193.2 NM_173194.2 NM_173195.2 NM_173197.2 XM_005269729.2 XM_005269730.2 XM_006717812.2 XM_011539731.2 XM_017016161.1 NR_045118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6691033 c.532C>T p.R178W Chr.10: 101827930-101827930 on Build GRCh38
282089 c.692C>T p.T231M Chr.10: 101827944-101827944 on Build GRCh38
5731360 c.527C>T p.A176V Chr.10: 101827935-101827935 on Build GRCh38
3790417 c.553G>C p.E185Q Chr.10: 101827903-101827903 on Build GRCh38
6691035 c.403C>T p.R135W Chr.10: 101827930-101827930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149050202 Chr.10: 101827929-101827929 on Build GRCh38
rs781697732 Chr.10: 101827776-101827776 on Build GRCh38
rs187864183 Chr.10: 101827847-101827847 on Build GRCh38
rs747659884 Chr.10: 101827798-101827798 on Build GRCh38
rs767843104 Chr.10: 101827900-101827900 on Build GRCh38
rs951526453 Chr.10: 101827749-101827749 on Build GRCh38
rs746575299 Chr.10: 101827899-101827899 on Build GRCh38
rs767900144 Chr.10: 101828043-101828043 on Build GRCh38
rs768461287 Chr.10: 101828020-101828020 on Build GRCh38
rs572725527 Chr.10: 101827815-101827815 on Build GRCh38
rs762753361 Chr.10: 101827880-101827880 on Build GRCh38
rs201481729 Chr.10: 101827930-101827930 on Build GRCh38
rs756477891 Chr.10: 101827907-101827907 on Build GRCh38
rs1034058019 Chr.10: 101827626-101827626 on Build GRCh38
rs775478545 Chr.10: 101827875-101827875 on Build GRCh38
rs751628876 Chr.10: 101827894-101827894 on Build GRCh38
rs745716009 Chr.10: 101827955-101827955 on Build GRCh38
rs950229053 Chr.10: 101827779-101827779 on Build GRCh38
rs752790110 Chr.10: 101827721-101827721 on Build GRCh38
rs751859470 Chr.10: 101827769-101827769 on Build GRCh38
rs112426578 Chr.10: 101827807-101827807 on Build GRCh38
rs992336586 Chr.10: 101827728-101827728 on Build GRCh38
rs757485761 Chr.10: 101827774-101827774 on Build GRCh38
rs780795809 Chr.10: 101827944-101827944 on Build GRCh38
rs369344782 Chr.10: 101827898-101827898 on Build GRCh38
rs776074231 Chr.10: 101827920-101827920 on Build GRCh38
rs2305189 Chr.10: 101827795-101827795 on Build GRCh38
rs769441984 Chr.10: 101827661-101827661 on Build GRCh38
rs747375622 Chr.10: 101827632-101827632 on Build GRCh38
rs771658331 Chr.10: 101827670-101827670 on Build GRCh38
rs781654565 Chr.10: 101827646-101827646 on Build GRCh38
rs374874936 Chr.10: 101827717-101827717 on Build GRCh38
rs1055645900 Chr.10: 101828010-101828010 on Build GRCh38
rs114881842 Chr.10: 101827838-101827838 on Build GRCh38
rs937234511 Chr.10: 101827973-101827973 on Build GRCh38
rs758477233 Chr.10: 101827737-101827737 on Build GRCh38
rs766195922 Chr.10: 101827694-101827694 on Build GRCh38
rs371574418 Chr.10: 101827649-101827649 on Build GRCh38
rs778052779 Chr.10: 101827618-101827618 on Build GRCh38
rs775110308 Chr.10: 101827970-101827970 on Build GRCh38
rs754398821 Chr.10: 101827928-101827928 on Build GRCh38
rs773791083 Chr.10: 101827657-101827657 on Build GRCh38
rs759484365 Chr.10: 101827707-101827707 on Build GRCh38
rs756751375 Chr.10: 101827790-101827790 on Build GRCh38
rs769656412 Chr.10: 101827956-101827956 on Build GRCh38
rs369666641 Chr.10: 101827787-101827787 on Build GRCh38
rs752768127 Chr.10: 101827596-101827596 on Build GRCh38
rs762957930 Chr.10: 101828019-101828019 on Build GRCh38
rs373476932 Chr.10: 101827845-101827845 on Build GRCh38
rs770637574 Chr.10: 101827943-101827943 on Build GRCh38
rs776347807 Chr.10: 101827705-101827705 on Build GRCh38
rs375884771 Chr.10: 101827799-101827799 on Build GRCh38
rs764275518 Chr.10: 101827759-101827759 on Build GRCh38
rs772896557 Chr.10: 101827679-101827679 on Build GRCh38
rs759326939 Chr.10: 101827864-101827864 on Build GRCh38
rs761953139 Chr.10: 101828031-101828031 on Build GRCh38
rs115921182 Chr.10: 101827906-101827906 on Build GRCh38
rs979357444 Chr.10: 101828036-101828036 on Build GRCh38
rs368114335 Chr.10: 101827603-101827603 on Build GRCh38
rs760269458 Chr.10: 101827682-101827682 on Build GRCh38
rs774985846 Chr.10: 101827749-101827749 on Build GRCh38
rs769663599 Chr.10: 101827869-101827869 on Build GRCh38
rs747685150 Chr.10: 101827662-101827662 on Build GRCh38
rs762280651 Chr.10: 101827883-101827883 on Build GRCh38
rs144960412 Chr.10: 101827924-101827924 on Build GRCh38
rs776468967 Chr.10: 101827856-101827856 on Build GRCh38
rs770579729 Chr.10: 101827914-101827914 on Build GRCh38
rs536763907 Chr.10: 101827642-101827642 on Build GRCh38
rs374027418 Chr.10: 101828022-101828022 on Build GRCh38
rs1001597455 Chr.10: 101827584-101827584 on Build GRCh38
rs764236283 Chr.10: 101827891-101827891 on Build GRCh38

View Full Product Details