Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101829540-101830068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNIP2
Entrez Gene ID
30819
Gene Name
potassium voltage-gated channel interacting protein 2
Gene Aliases
KCHIP2
UniGene
Hs.97044
Gene Chromosome Location
Chr.10: 101825971-101843920 on Build GRCh38
Transcripts
NM_014591.4 NM_173191.2 NM_173192.2 NM_173193.2 NM_173194.2 NM_173195.2 NM_173197.2 XM_005269729.2 XM_005269730.2 XM_006717812.2 XM_011539731.2 XM_017016161.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6367085 c.247A>G p.R83G Chr.10: 101829865-101829865 on Build GRCh38
4912053 c.224C>A p.A75D Chr.10: 101829888-101829888 on Build GRCh38
302458 c.244delC p.H82fs*105 Chr.10: 101829868-101829868 on Build GRCh38
4951232 c.170-2A>T p.? Chr.10: 101829944-101829944 on Build GRCh38
5369951 c.190C>T p.L64F Chr.10: 101829922-101829922 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs577386801 Chr.10: 101830040-101830040 on Build GRCh38
rs759534660 Chr.10: 101829883-101829883 on Build GRCh38
rs1048035903 Chr.10: 101829859-101829859 on Build GRCh38
rs1032733131 Chr.10: 101829997-101829997 on Build GRCh38
rs888138861 Chr.10: 101829868-101829868 on Build GRCh38
rs1034137281 Chr.10: 101829795-101829795 on Build GRCh38
rs570628954 Chr.10: 101829771-101829771 on Build GRCh38
rs553844476 Chr.10: 101829826-101829826 on Build GRCh38
rs1018031045 Chr.10: 101829935-101829935 on Build GRCh38
rs565453539 Chr.10: 101829996-101829996 on Build GRCh38
rs572130897 Chr.10: 101829902-101829902 on Build GRCh38
rs764217442 Chr.10: 101829945-101829945 on Build GRCh38
rs867019660 Chr.10: 101829982-101829982 on Build GRCh38
rs376696004 Chr.10: 101829958-101829958 on Build GRCh38
rs763097435 Chr.10: 101829937-101829937 on Build GRCh38
rs906807328 Chr.10: 101829638-101829638 on Build GRCh38
rs746326473 Chr.10: 101830038-101830038 on Build GRCh38
rs746616814 Chr.10: 101829831-101829831 on Build GRCh38
rs966939877 Chr.10: 101829990-101829990 on Build GRCh38
rs747606945 Chr.10: 101829815-101829815 on Build GRCh38
rs929916944 Chr.10: 101829858-101829858 on Build GRCh38
rs574677484 Chr.10: 101829809-101829809 on Build GRCh38
rs770746068 Chr.10: 101829870-101829870 on Build GRCh38
rs537720840 Chr.10: 101829804-101829804 on Build GRCh38
rs952525843 Chr.10: 101829994-101829994 on Build GRCh38
rs535441934 Chr.10: 101829817-101829817 on Build GRCh38
rs181922684 Chr.10: 101829806-101829806 on Build GRCh38
rs777382927 Chr.10: 101829828-101829828 on Build GRCh38
rs769608348 Chr.10: 101829892-101829892 on Build GRCh38
rs895153180 Chr.10: 101829820-101829821 on Build GRCh38
rs199767134 Chr.10: 101829984-101829984 on Build GRCh38
rs867618475 Chr.10: 101829885-101829885 on Build GRCh38
rs868327134 Chr.10: 101829760-101829760 on Build GRCh38
rs994849339 Chr.10: 101829824-101829824 on Build GRCh38
rs776223482 Chr.10: 101829882-101829882 on Build GRCh38
rs34566627 Chr.10: 101829583-101829584 on Build GRCh38
rs916358318 Chr.10: 101829588-101829588 on Build GRCh38
rs746957721 Chr.10: 101829864-101829864 on Build GRCh38
rs1007012928 Chr.10: 101829907-101829907 on Build GRCh38
rs945545863 Chr.10: 101829584-101829584 on Build GRCh38
rs982605190 Chr.10: 101830011-101830011 on Build GRCh38
rs756517192 Chr.10: 101829793-101829793 on Build GRCh38
rs868040497 Chr.10: 101829593-101829593 on Build GRCh38
rs978354239 Chr.10: 101829993-101829993 on Build GRCh38
rs1003539135 Chr.10: 101829719-101829719 on Build GRCh38
rs558287378 Chr.10: 101829581-101829581 on Build GRCh38
rs560298441 Chr.10: 101830015-101830015 on Build GRCh38
rs185283450 Chr.10: 101829936-101829936 on Build GRCh38
rs145543468 Chr.10: 101829928-101829928 on Build GRCh38

View Full Product Details