Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102227541-102228043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL3
Entrez Gene ID
83401
Gene Name
ELOVL fatty acid elongase 3
Gene Aliases
CIG-30, CIG30
UniGene
Hs.302130
Gene Chromosome Location
Chr.10: 102224767-102229589 on Build GRCh38
Transcripts
NM_152310.2 XM_011540245.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
371007 c.204C>A p.I68I Chr.10: 102227728-102227728 on Build GRCh38
5919859 c.107C>T p.T36I Chr.10: 102227631-102227631 on Build GRCh38
914140 c.219C>T p.C73C Chr.10: 102227743-102227743 on Build GRCh38
3433713 c.115C>T p.P39S Chr.10: 102227639-102227639 on Build GRCh38
4460000 c.114C>T p.F38F Chr.10: 102227638-102227638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760858592 Chr.10: 102227626-102227626 on Build GRCh38
rs780903547 Chr.10: 102227578-102227578 on Build GRCh38
rs749786533 Chr.10: 102227713-102227713 on Build GRCh38
rs760914912 Chr.10: 102227711-102227711 on Build GRCh38
rs950659122 Chr.10: 102227829-102227829 on Build GRCh38
rs570641098 Chr.10: 102227828-102227828 on Build GRCh38
rs751900625 Chr.10: 102227762-102227762 on Build GRCh38
rs527925433 Chr.10: 102227604-102227604 on Build GRCh38
rs755743002 Chr.10: 102227770-102227770 on Build GRCh38
rs1052076619 Chr.10: 102227958-102227958 on Build GRCh38
rs941864890 Chr.10: 102227868-102227868 on Build GRCh38
rs761710097 Chr.10: 102227608-102227608 on Build GRCh38
rs765368871 Chr.10: 102227725-102227725 on Build GRCh38
rs932500308 Chr.10: 102227868-102227868 on Build GRCh38
rs575590142 Chr.10: 102227742-102227742 on Build GRCh38
rs959822041 Chr.10: 102227709-102227709 on Build GRCh38
rs758759044 Chr.10: 102227660-102227660 on Build GRCh38
rs767565579 Chr.10: 102227609-102227609 on Build GRCh38
rs762307295 Chr.10: 102227891-102227891 on Build GRCh38
rs776634398 Chr.10: 102227718-102227718 on Build GRCh38
rs959551099 Chr.10: 102227997-102227997 on Build GRCh38
rs761898830 Chr.10: 102227930-102227930 on Build GRCh38
rs774432163 Chr.10: 102227696-102227696 on Build GRCh38
rs771881239 Chr.10: 102227710-102227710 on Build GRCh38
rs528557257 Chr.10: 102227857-102227857 on Build GRCh38
rs540055284 Chr.10: 102227890-102227890 on Build GRCh38
rs1032534189 Chr.10: 102227580-102227580 on Build GRCh38
rs1011724411 Chr.10: 102227749-102227749 on Build GRCh38
rs753191554 Chr.10: 102227731-102227731 on Build GRCh38
rs564698986 Chr.10: 102227907-102227907 on Build GRCh38
rs145666911 Chr.10: 102227931-102227931 on Build GRCh38
rs185337007 Chr.10: 102227993-102227993 on Build GRCh38
rs1041372132 Chr.10: 102227846-102227846 on Build GRCh38
rs533437585 Chr.10: 102227996-102227996 on Build GRCh38
rs997619848 Chr.10: 102227634-102227634 on Build GRCh38
rs150930565 Chr.10: 102227717-102227717 on Build GRCh38
rs759617475 Chr.10: 102227721-102227721 on Build GRCh38
rs974011960 Chr.10: 102228011-102228011 on Build GRCh38
rs1053536780 Chr.10: 102227898-102227898 on Build GRCh38
rs755761829 Chr.10: 102227669-102227669 on Build GRCh38
rs768653709 Chr.10: 102227693-102227693 on Build GRCh38
rs766455075 Chr.10: 102227628-102227628 on Build GRCh38
rs532794438 Chr.10: 102227654-102227654 on Build GRCh38
rs902195192 Chr.10: 102227960-102227960 on Build GRCh38
rs983380490 Chr.10: 102227795-102227795 on Build GRCh38
rs376359483 Chr.10: 102227592-102227592 on Build GRCh38
rs192318756 Chr.10: 102227648-102227648 on Build GRCh38
rs201241444 Chr.10: 102227589-102227589 on Build GRCh38
rs957547966 Chr.10: 102227699-102227699 on Build GRCh38
rs559483130 Chr.10: 102227617-102227617 on Build GRCh38
rs370654761 Chr.10: 102227583-102227583 on Build GRCh38
rs373730220 Chr.10: 102227679-102227679 on Build GRCh38
rs764487617 Chr.10: 102227751-102227751 on Build GRCh38
rs549925937 Chr.10: 102228012-102228012 on Build GRCh38
rs1020594365 Chr.10: 102227922-102227922 on Build GRCh38
rs750371471 Chr.10: 102227721-102227722 on Build GRCh38
rs773476724 Chr.10: 102227610-102227610 on Build GRCh38
rs370240986 Chr.10: 102227596-102227596 on Build GRCh38
rs753899817 Chr.10: 102227643-102227643 on Build GRCh38
rs143448083 Chr.10: 102227668-102227668 on Build GRCh38
rs748161170 Chr.10: 102227697-102227697 on Build GRCh38
rs909270079 Chr.10: 102227862-102227862 on Build GRCh38
rs973500804 Chr.10: 102227724-102227724 on Build GRCh38
rs778621546 Chr.10: 102227808-102227808 on Build GRCh38
rs745575714 Chr.10: 102227582-102227582 on Build GRCh38
rs779575679 Chr.10: 102227672-102227672 on Build GRCh38
rs1038851229 Chr.10: 102227882-102227882 on Build GRCh38
rs752993519 Chr.10: 102227659-102227659 on Build GRCh38
rs768745641 Chr.10: 102227593-102227593 on Build GRCh38
rs999118473 Chr.10: 102227970-102227970 on Build GRCh38
rs368580874 Chr.10: 102227805-102227805 on Build GRCh38
rs368166326 Chr.10: 102227777-102227777 on Build GRCh38
rs370783391 Chr.10: 102227794-102227794 on Build GRCh38
rs117979772 Chr.10: 102227803-102227803 on Build GRCh38
rs1009327838 Chr.10: 102227917-102227917 on Build GRCh38

View Full Product Details