Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102864176-102864717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORCS7
Entrez Gene ID
119032
Gene Name
BLOC-1 related complex subunit 7
Gene Aliases
C10orf32
UniGene
Hs.34492
Gene Chromosome Location
Chr.10: 102854210-102864961 on Build GRCh38
Transcripts
NM_001136200.1 NM_144591.3 NR_037644.1

Gene Symbol
BORCS7-ASMT
Entrez Gene ID
100528007
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
Gene Aliases
C10orf32-AS3MT, C10orf32-ASMT
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102854210-102901899 on Build GRCh38
Transcripts
NM_001136200.1 NM_144591.3 NR_037644.1

SNP Detail

SNP ID Chromosome Location
rs569704211 Chr.10: 102864647-102864647 on Build GRCh38
rs1010121870 Chr.10: 102864359-102864359 on Build GRCh38
rs866030939 Chr.10: 102864388-102864388 on Build GRCh38
rs754526869 Chr.10: 102864367-102864367 on Build GRCh38
rs138030676 Chr.10: 102864532-102864532 on Build GRCh38
rs533107831 Chr.10: 102864595-102864595 on Build GRCh38
rs907506346 Chr.10: 102864241-102864241 on Build GRCh38
rs529344540 Chr.10: 102864515-102864515 on Build GRCh38
rs537734074 Chr.10: 102864597-102864597 on Build GRCh38
rs574244347 Chr.10: 102864646-102864646 on Build GRCh38
rs922775433 Chr.10: 102864248-102864248 on Build GRCh38
rs41287474 Chr.10: 102864600-102864600 on Build GRCh38
rs891096706 Chr.10: 102864439-102864439 on Build GRCh38
rs937147587 Chr.10: 102864298-102864298 on Build GRCh38
rs955851861 Chr.10: 102864270-102864270 on Build GRCh38
rs951562377 Chr.10: 102864549-102864549 on Build GRCh38
rs150359631 Chr.10: 102864406-102864406 on Build GRCh38
rs935064737 Chr.10: 102864475-102864475 on Build GRCh38
rs80142826 Chr.10: 102864274-102864274 on Build GRCh38
rs559688462 Chr.10: 102864565-102864565 on Build GRCh38
rs949767990 Chr.10: 102864245-102864245 on Build GRCh38
rs972924600 Chr.10: 102864243-102864243 on Build GRCh38
rs1027076677 Chr.10: 102864535-102864535 on Build GRCh38
rs947845425 Chr.10: 102864550-102864550 on Build GRCh38
rs964136279 Chr.10: 102864531-102864531 on Build GRCh38
rs923722368 Chr.10: 102864467-102864467 on Build GRCh38
rs961423868 Chr.10: 102864238-102864238 on Build GRCh38
rs191692489 Chr.10: 102864425-102864425 on Build GRCh38
rs1054210790 Chr.10: 102864681-102864681 on Build GRCh38
rs866943446 Chr.10: 102864260-102864260 on Build GRCh38
rs530507706 Chr.10: 102864680-102864680 on Build GRCh38
rs1008251527 Chr.10: 102864510-102864510 on Build GRCh38
rs951593358 Chr.10: 102864226-102864226 on Build GRCh38
rs4917985 Chr.10: 102864315-102864315 on Build GRCh38
rs576484646 Chr.10: 102864337-102864337 on Build GRCh38
rs927026186 Chr.10: 102864655-102864655 on Build GRCh38
rs554092099 Chr.10: 102864201-102864201 on Build GRCh38
rs558507342 Chr.10: 102864412-102864412 on Build GRCh38
rs983062773 Chr.10: 102864237-102864237 on Build GRCh38
rs142871406 Chr.10: 102864326-102864326 on Build GRCh38
rs927095278 Chr.10: 102864280-102864280 on Build GRCh38

View Full Product Details