Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103446694-103447187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALHM2
Entrez Gene ID
51063
Gene Name
calcium homeostasis modulator 2
Gene Aliases
FAM26B
UniGene
Hs.241545
Gene Chromosome Location
Chr.10: 103446785-103452419 on Build GRCh38
Transcripts
NM_015916.4 NR_024552.1 NR_046344.1 XM_006717880.2 XM_006717883.2 XM_006717884.3 XM_011539848.2 XM_017016306.1 XM_017016307.1 XM_017016308.1 XM_017016309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6201027 c.937C>A p.P313T Chr.10: 103447187-103447187 on Build GRCh38
6619087 c.967T>G p.S323A Chr.10: 103447157-103447157 on Build GRCh38
2059912 c.945C>T p.N315N Chr.10: 103447179-103447179 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774136884 Chr.10: 103447024-103447024 on Build GRCh38
rs1031701687 Chr.10: 103446770-103446770 on Build GRCh38
rs538819372 Chr.10: 103446925-103446925 on Build GRCh38
rs1039542546 Chr.10: 103447059-103447059 on Build GRCh38
rs577789094 Chr.10: 103446879-103446879 on Build GRCh38
rs879642023 Chr.10: 103446937-103446937 on Build GRCh38
rs1041375090 Chr.10: 103446990-103446990 on Build GRCh38
rs1013557624 Chr.10: 103447160-103447160 on Build GRCh38
rs914290349 Chr.10: 103446932-103446932 on Build GRCh38
rs540750564 Chr.10: 103446725-103446725 on Build GRCh38
rs971997586 Chr.10: 103446884-103446884 on Build GRCh38
rs768566714 Chr.10: 103446740-103446740 on Build GRCh38
rs6580 Chr.10: 103447117-103447117 on Build GRCh38
rs542696088 Chr.10: 103446952-103446952 on Build GRCh38
rs557157128 Chr.10: 103446928-103446928 on Build GRCh38
rs768212131 Chr.10: 103447133-103447133 on Build GRCh38
rs888836977 Chr.10: 103446977-103446977 on Build GRCh38
rs184715548 Chr.10: 103446876-103446876 on Build GRCh38
rs748834217 Chr.10: 103446996-103446996 on Build GRCh38
rs771980308 Chr.10: 103447145-103447145 on Build GRCh38
rs563789065 Chr.10: 103446803-103446803 on Build GRCh38
rs1051386510 Chr.10: 103446960-103446960 on Build GRCh38
rs771390694 Chr.10: 103447121-103447121 on Build GRCh38
rs779559082 Chr.10: 103447078-103447078 on Build GRCh38
rs113071440 Chr.10: 103446756-103446756 on Build GRCh38
rs949606074 Chr.10: 103446841-103446841 on Build GRCh38
rs761355035 Chr.10: 103447140-103447140 on Build GRCh38
rs575431292 Chr.10: 103446944-103446944 on Build GRCh38
rs371260243 Chr.10: 103446936-103446936 on Build GRCh38
rs748910135 Chr.10: 103447130-103447130 on Build GRCh38
rs1001108417 Chr.10: 103446768-103446768 on Build GRCh38
rs766131645 Chr.10: 103447165-103447165 on Build GRCh38
rs560728020 Chr.10: 103447035-103447035 on Build GRCh38
rs951478089 Chr.10: 103446732-103446732 on Build GRCh38
rs533426305 Chr.10: 103447159-103447159 on Build GRCh38
rs1018411815 Chr.10: 103446723-103446723 on Build GRCh38
rs773814424 Chr.10: 103447138-103447138 on Build GRCh38
rs757863850 Chr.10: 103447102-103447103 on Build GRCh38
rs572796307 Chr.10: 103447099-103447099 on Build GRCh38
rs140359433 Chr.10: 103446794-103446794 on Build GRCh38
rs534651894 Chr.10: 103446827-103446827 on Build GRCh38
rs904009813 Chr.10: 103447077-103447077 on Build GRCh38
rs937135008 Chr.10: 103446896-103446896 on Build GRCh38
rs1040068404 Chr.10: 103447083-103447093 on Build GRCh38
rs904091004 Chr.10: 103446765-103446765 on Build GRCh38
rs956726481 Chr.10: 103446778-103446778 on Build GRCh38
rs753591491 Chr.10: 103447167-103447167 on Build GRCh38
rs567609259 Chr.10: 103446823-103446823 on Build GRCh38
rs778472486 Chr.10: 103447102-103447102 on Build GRCh38
rs372230850 Chr.10: 103447126-103447126 on Build GRCh38
rs372810153 Chr.10: 103446844-103446844 on Build GRCh38
rs532772319 Chr.10: 103446892-103446892 on Build GRCh38
rs559982700 Chr.10: 103447158-103447158 on Build GRCh38

View Full Product Details