Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116883218-116883751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO4
Entrez Gene ID
387712
Gene Name
enolase family member 4
Gene Aliases
C10orf134
UniGene
Hs.693248
Gene Chromosome Location
Chr.10: 116848230-116911803 on Build GRCh38
Transcripts
XM_006717835.3 XM_006717836.3 XM_011539797.2 XM_011539799.2 XM_017016241.1 NM_001127211.2 NM_001258298.1 NM_018330.6

Gene Symbol
SHTN1
Entrez Gene ID
57698
Gene Name
shootin 1
Gene Aliases
KIAA1598
UniGene
Hs.501140
Gene Chromosome Location
Chr.10: 116883377-117126586 on Build GRCh38
Transcripts
XM_006717835.3 XM_006717836.3 XM_011539797.2 XM_011539799.2 XM_017016241.1 NM_001127211.2 NM_001258298.1 NM_018330.6

SNP Detail

SNP ID Chromosome Location
rs989496576 Chr.10: 116883461-116883461 on Build GRCh38
rs975371853 Chr.10: 116883445-116883445 on Build GRCh38
rs115529496 Chr.10: 116883685-116883685 on Build GRCh38
rs914078359 Chr.10: 116883466-116883466 on Build GRCh38
rs994601593 Chr.10: 116883590-116883590 on Build GRCh38
rs1055532615 Chr.10: 116883264-116883265 on Build GRCh38
rs1018863070 Chr.10: 116883400-116883400 on Build GRCh38
rs537505437 Chr.10: 116883450-116883450 on Build GRCh38
rs775936398 Chr.10: 116883523-116883523 on Build GRCh38
rs941267018 Chr.10: 116883603-116883603 on Build GRCh38
rs569818658 Chr.10: 116883435-116883435 on Build GRCh38
rs1056045053 Chr.10: 116883606-116883606 on Build GRCh38
rs142357804 Chr.10: 116883663-116883663 on Build GRCh38
rs922268908 Chr.10: 116883487-116883487 on Build GRCh38
rs945523864 Chr.10: 116883469-116883469 on Build GRCh38
rs921343510 Chr.10: 116883249-116883249 on Build GRCh38
rs1045784458 Chr.10: 116883648-116883648 on Build GRCh38
rs976489160 Chr.10: 116883471-116883471 on Build GRCh38
rs574328598 Chr.10: 116883662-116883662 on Build GRCh38
rs888354982 Chr.10: 116883262-116883262 on Build GRCh38
rs1007786564 Chr.10: 116883720-116883720 on Build GRCh38
rs539611886 Chr.10: 116883251-116883251 on Build GRCh38
rs945633451 Chr.10: 116883291-116883291 on Build GRCh38
rs886312272 Chr.10: 116883617-116883617 on Build GRCh38
rs72831678 Chr.10: 116883322-116883322 on Build GRCh38
rs1005379765 Chr.10: 116883263-116883263 on Build GRCh38
rs921375120 Chr.10: 116883449-116883449 on Build GRCh38
rs898808572 Chr.10: 116883563-116883563 on Build GRCh38
rs950183794 Chr.10: 116883637-116883637 on Build GRCh38
rs1053863928 Chr.10: 116883267-116883267 on Build GRCh38
rs553715463 Chr.10: 116883380-116883380 on Build GRCh38
rs967367621 Chr.10: 116883312-116883314 on Build GRCh38
rs572618523 Chr.10: 116883397-116883397 on Build GRCh38
rs1054777713 Chr.10: 116883527-116883527 on Build GRCh38
rs994694862 Chr.10: 116883369-116883369 on Build GRCh38
rs1008724490 Chr.10: 116883654-116883654 on Build GRCh38
rs989525638 Chr.10: 116883711-116883711 on Build GRCh38
rs761306621 Chr.10: 116883609-116883609 on Build GRCh38
rs1014309428 Chr.10: 116883277-116883277 on Build GRCh38
rs1026169154 Chr.10: 116883377-116883377 on Build GRCh38
rs1019308251 Chr.10: 116883661-116883661 on Build GRCh38
rs914048963 Chr.10: 116883287-116883287 on Build GRCh38
rs752291553 Chr.10: 116883591-116883591 on Build GRCh38
rs906192474 Chr.10: 116883313-116883313 on Build GRCh38
rs1001874477 Chr.10: 116883314-116883314 on Build GRCh38
rs1054915459 Chr.10: 116883318-116883318 on Build GRCh38
rs560907848 Chr.10: 116883526-116883526 on Build GRCh38
rs894579404 Chr.10: 116883272-116883272 on Build GRCh38
rs1046608924 Chr.10: 116883311-116883311 on Build GRCh38
rs1047123244 Chr.10: 116883493-116883493 on Build GRCh38
rs146481218 Chr.10: 116883674-116883674 on Build GRCh38

View Full Product Details