Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116883651-116884156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO4
Entrez Gene ID
387712
Gene Name
enolase family member 4
Gene Aliases
C10orf134
UniGene
Hs.693248
Gene Chromosome Location
Chr.10: 116848230-116911803 on Build GRCh38
Transcripts
XM_006717835.3 XM_006717836.3 XM_011539797.2 XM_011539799.2 XM_017016241.1 NM_001127211.2 NM_001258298.1 NM_018330.6

Gene Symbol
SHTN1
Entrez Gene ID
57698
Gene Name
shootin 1
Gene Aliases
KIAA1598
UniGene
Hs.501140
Gene Chromosome Location
Chr.10: 116883377-117126586 on Build GRCh38
Transcripts
XM_006717835.3 XM_006717836.3 XM_011539797.2 XM_011539799.2 XM_017016241.1 NM_001127211.2 NM_001258298.1 NM_018330.6

SNP Detail

SNP ID Chromosome Location
rs17678211 Chr.10: 116883732-116883732 on Build GRCh38
rs966203869 Chr.10: 116883758-116883758 on Build GRCh38
rs559117392 Chr.10: 116883790-116883790 on Build GRCh38
rs374564114 Chr.10: 116884000-116884000 on Build GRCh38
rs115529496 Chr.10: 116883685-116883685 on Build GRCh38
rs537023513 Chr.10: 116884093-116884093 on Build GRCh38
rs900306116 Chr.10: 116883993-116883993 on Build GRCh38
rs766298205 Chr.10: 116883795-116883795 on Build GRCh38
rs893119396 Chr.10: 116884004-116884004 on Build GRCh38
rs982440104 Chr.10: 116883841-116883841 on Build GRCh38
rs991380098 Chr.10: 116883920-116883920 on Build GRCh38
rs893846518 Chr.10: 116884014-116884014 on Build GRCh38
rs920265549 Chr.10: 116883837-116883837 on Build GRCh38
rs568339770 Chr.10: 116883919-116883920 on Build GRCh38
rs982286473 Chr.10: 116883791-116883791 on Build GRCh38
rs989525638 Chr.10: 116883711-116883711 on Build GRCh38
rs553769828 Chr.10: 116883988-116883988 on Build GRCh38
rs996059265 Chr.10: 116884003-116884003 on Build GRCh38
rs1026884351 Chr.10: 116884069-116884069 on Build GRCh38
rs967312805 Chr.10: 116883752-116883752 on Build GRCh38
rs754921450 Chr.10: 116884046-116884046 on Build GRCh38
rs867523494 Chr.10: 116883897-116883897 on Build GRCh38
rs949951101 Chr.10: 116883974-116883974 on Build GRCh38
rs934720159 Chr.10: 116883994-116883994 on Build GRCh38
rs568470254 Chr.10: 116883903-116883903 on Build GRCh38
rs576734703 Chr.10: 116883751-116883751 on Build GRCh38
rs866215333 Chr.10: 116883796-116883796 on Build GRCh38
rs572376104 Chr.10: 116883991-116883991 on Build GRCh38
rs966800544 Chr.10: 116884047-116884047 on Build GRCh38
rs79646754 Chr.10: 116883920-116883920 on Build GRCh38
rs1007786564 Chr.10: 116883720-116883720 on Build GRCh38
rs1047546500 Chr.10: 116883849-116883849 on Build GRCh38
rs1040227682 Chr.10: 116883990-116883990 on Build GRCh38
rs551069188 Chr.10: 116884103-116884103 on Build GRCh38
rs1021484340 Chr.10: 116884027-116884027 on Build GRCh38
rs915739896 Chr.10: 116883920-116883923 on Build GRCh38
rs79670792 Chr.10: 116883919-116883919 on Build GRCh38
rs9421253 Chr.10: 116883835-116883835 on Build GRCh38
rs1010852480 Chr.10: 116884015-116884015 on Build GRCh38
rs548066333 Chr.10: 116883838-116883838 on Build GRCh38
rs1032850517 Chr.10: 116884011-116884011 on Build GRCh38

View Full Product Details