Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 118681247-118681748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACUL1
Entrez Gene ID
143384
Gene Name
CDK2 associated cullin domain 1
Gene Aliases
C10orf46, CAC1
UniGene
Hs.420024
Gene Chromosome Location
Chr.10: 118680982-118755246 on Build GRCh38
Transcripts
NM_153810.4

SNP Detail

SNP ID Chromosome Location
rs978625986 Chr.10: 118681461-118681461 on Build GRCh38
rs928389816 Chr.10: 118681279-118681279 on Build GRCh38
rs543447092 Chr.10: 118681502-118681502 on Build GRCh38
rs993532998 Chr.10: 118681315-118681315 on Build GRCh38
rs776705157 Chr.10: 118681462-118681462 on Build GRCh38
rs896970458 Chr.10: 118681677-118681677 on Build GRCh38
rs554948828 Chr.10: 118681487-118681487 on Build GRCh38
rs995767809 Chr.10: 118681722-118681722 on Build GRCh38
rs571495335 Chr.10: 118681271-118681271 on Build GRCh38
rs199708518 Chr.10: 118681525-118681526 on Build GRCh38
rs183893514 Chr.10: 118681457-118681457 on Build GRCh38
rs545918524 Chr.10: 118681675-118681675 on Build GRCh38
rs189717980 Chr.10: 118681638-118681638 on Build GRCh38
rs906020657 Chr.10: 118681269-118681269 on Build GRCh38
rs950116280 Chr.10: 118681532-118681532 on Build GRCh38
rs772392737 Chr.10: 118681392-118681392 on Build GRCh38
rs1027342237 Chr.10: 118681481-118681481 on Build GRCh38
rs914101657 Chr.10: 118681305-118681306 on Build GRCh38
rs961540416 Chr.10: 118681470-118681470 on Build GRCh38
rs760797628 Chr.10: 118681400-118681400 on Build GRCh38
rs368564720 Chr.10: 118681527-118681527 on Build GRCh38
rs748581557 Chr.10: 118681388-118681388 on Build GRCh38
rs17097960 Chr.10: 118681437-118681437 on Build GRCh38
rs191562482 Chr.10: 118681288-118681288 on Build GRCh38
rs146932305 Chr.10: 118681420-118681420 on Build GRCh38
rs1033054389 Chr.10: 118681480-118681480 on Build GRCh38
rs914060444 Chr.10: 118681564-118681564 on Build GRCh38
rs11550771 Chr.10: 118681615-118681615 on Build GRCh38
rs575982405 Chr.10: 118681501-118681501 on Build GRCh38
rs765623618 Chr.10: 118681480-118681481 on Build GRCh38
rs752936795 Chr.10: 118681606-118681606 on Build GRCh38
rs898886744 Chr.10: 118681707-118681707 on Build GRCh38
rs989190083 Chr.10: 118681511-118681511 on Build GRCh38
rs1049718110 Chr.10: 118681644-118681644 on Build GRCh38
rs765612853 Chr.10: 118681515-118681515 on Build GRCh38
rs1025990450 Chr.10: 118681384-118681384 on Build GRCh38
rs199609134 Chr.10: 118681713-118681713 on Build GRCh38
rs112050425 Chr.10: 118681363-118681363 on Build GRCh38
rs932617984 Chr.10: 118681632-118681632 on Build GRCh38
rs891164039 Chr.10: 118681645-118681645 on Build GRCh38
rs566509430 Chr.10: 118681525-118681525 on Build GRCh38
rs945558218 Chr.10: 118681606-118681606 on Build GRCh38
rs1015377174 Chr.10: 118681466-118681466 on Build GRCh38
rs201789798 Chr.10: 118681709-118681709 on Build GRCh38
rs141437706 Chr.10: 118681289-118681289 on Build GRCh38
rs979778018 Chr.10: 118681521-118681521 on Build GRCh38
rs759690577 Chr.10: 118681474-118681474 on Build GRCh38
rs1043959654 Chr.10: 118681614-118681614 on Build GRCh38
rs997272228 Chr.10: 118681429-118681429 on Build GRCh38
rs988599949 Chr.10: 118681555-118681555 on Build GRCh38
rs1008257309 Chr.10: 118681663-118681663 on Build GRCh38
rs1038506024 Chr.10: 118681637-118681637 on Build GRCh38
rs891216258 Chr.10: 118681438-118681438 on Build GRCh38
rs980307461 Chr.10: 118681583-118681583 on Build GRCh38

View Full Product Details