Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 127175525-127176021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK1
Entrez Gene ID
1793
Gene Name
dedicator of cytokinesis 1
Gene Aliases
DOCK180, ced5
UniGene
Hs.159195
Gene Chromosome Location
Chr.10: 126905409-127452517 on Build GRCh38
Transcripts
NM_001290223.1 NM_001380.4 XM_011539422.2 XM_011539424.1 XM_017015813.1 XM_017015814.1 XM_017015815.1 XM_017015816.1 NM_001039762.2 XM_005252694.3 XM_017016537.1 XM_017016538.1 XM_017016539.1 XM_017016540.1 XM_017016541.1 XM_017016542.1 XM_017016543.1

Gene Symbol
FAM196A
Entrez Gene ID
642938
Gene Name
family with sequence similarity 196 member A
Gene Aliases
C10orf141
UniGene
Hs.613882
Gene Chromosome Location
Chr.10: 127135426-127196522 on Build GRCh38
Transcripts
NM_001290223.1 NM_001380.4 XM_011539422.2 XM_011539424.1 XM_017015813.1 XM_017015814.1 XM_017015815.1 XM_017015816.1 NM_001039762.2 XM_005252694.3 XM_017016537.1 XM_017016538.1 XM_017016539.1 XM_017016540.1 XM_017016541.1 XM_017016542.1 XM_017016543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4012588 c.650C>T p.S217F Chr.10: 127175746-127175746 on Build GRCh38
916001 c.471G>A p.E157E Chr.10: 127175925-127175925 on Build GRCh38
4012590 c.564G>T p.L188F Chr.10: 127175832-127175832 on Build GRCh38
123005 c.507G>C p.K169N Chr.10: 127175889-127175889 on Build GRCh38
5423348 c.703C>T p.P235S Chr.10: 127175693-127175693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756966142 Chr.10: 127175741-127175741 on Build GRCh38
rs766189937 Chr.10: 127175907-127175907 on Build GRCh38
rs184591766 Chr.10: 127175872-127175872 on Build GRCh38
rs140516988 Chr.10: 127175793-127175793 on Build GRCh38
rs374827963 Chr.10: 127175667-127175667 on Build GRCh38
rs374342932 Chr.10: 127175917-127175917 on Build GRCh38
rs769661665 Chr.10: 127175685-127175685 on Build GRCh38
rs771096143 Chr.10: 127175767-127175767 on Build GRCh38
rs369909713 Chr.10: 127175625-127175625 on Build GRCh38
rs755904233 Chr.10: 127175823-127175823 on Build GRCh38
rs376988297 Chr.10: 127175877-127175877 on Build GRCh38
rs778371072 Chr.10: 127175577-127175577 on Build GRCh38
rs77083569 Chr.10: 127175976-127175976 on Build GRCh38
rs901091271 Chr.10: 127175785-127175785 on Build GRCh38
rs773109284 Chr.10: 127175598-127175598 on Build GRCh38
rs940714348 Chr.10: 127175983-127175983 on Build GRCh38
rs760887375 Chr.10: 127175716-127175716 on Build GRCh38
rs371917458 Chr.10: 127175548-127175548 on Build GRCh38
rs201652122 Chr.10: 127175675-127175675 on Build GRCh38
rs760865513 Chr.10: 127175702-127175702 on Build GRCh38
rs777876080 Chr.10: 127175854-127175854 on Build GRCh38
rs773425911 Chr.10: 127175714-127175714 on Build GRCh38
rs368886635 Chr.10: 127175547-127175547 on Build GRCh38
rs370964104 Chr.10: 127175883-127175883 on Build GRCh38
rs924454038 Chr.10: 127175855-127175855 on Build GRCh38
rs1004876591 Chr.10: 127175612-127175612 on Build GRCh38
rs761165368 Chr.10: 127175626-127175626 on Build GRCh38
rs757987233 Chr.10: 127175731-127175731 on Build GRCh38
rs930758623 Chr.10: 127175757-127175757 on Build GRCh38
rs761300471 Chr.10: 127175904-127175904 on Build GRCh38
rs1027675265 Chr.10: 127175571-127175571 on Build GRCh38
rs747825338 Chr.10: 127175595-127175595 on Build GRCh38
rs201298990 Chr.10: 127175937-127175937 on Build GRCh38
rs777637245 Chr.10: 127175651-127175651 on Build GRCh38
rs761994515 Chr.10: 127175622-127175622 on Build GRCh38
rs764748949 Chr.10: 127175897-127175897 on Build GRCh38
rs750232225 Chr.10: 127175815-127175815 on Build GRCh38
rs139810222 Chr.10: 127175698-127175698 on Build GRCh38
rs770771122 Chr.10: 127175613-127175613 on Build GRCh38
rs565955024 Chr.10: 127175621-127175621 on Build GRCh38
rs539100904 Chr.10: 127175643-127175643 on Build GRCh38
rs766459538 Chr.10: 127175901-127175901 on Build GRCh38
rs549935946 Chr.10: 127175863-127175863 on Build GRCh38
rs531727184 Chr.10: 127175800-127175800 on Build GRCh38
rs745704190 Chr.10: 127175744-127175744 on Build GRCh38
rs762505186 Chr.10: 127175782-127175782 on Build GRCh38
rs779906190 Chr.10: 127175827-127175827 on Build GRCh38
rs748154370 Chr.10: 127175750-127175750 on Build GRCh38
rs765894265 Chr.10: 127175723-127175723 on Build GRCh38
rs778597060 Chr.10: 127175637-127175637 on Build GRCh38
rs752031911 Chr.10: 127175853-127175853 on Build GRCh38
rs776635279 Chr.10: 127175616-127175616 on Build GRCh38
rs754378814 Chr.10: 127175631-127175631 on Build GRCh38
rs11594560 Chr.10: 127175956-127175956 on Build GRCh38
rs935714014 Chr.10: 127175861-127175861 on Build GRCh38
rs777030866 Chr.10: 127175772-127175772 on Build GRCh38
rs748989064 Chr.10: 127175943-127175943 on Build GRCh38
rs150007430 Chr.10: 127175942-127175942 on Build GRCh38
rs150978210 Chr.10: 127175745-127175745 on Build GRCh38
rs752623435 Chr.10: 127175602-127175602 on Build GRCh38
rs200216671 Chr.10: 127175912-127175912 on Build GRCh38
rs767778753 Chr.10: 127175624-127175624 on Build GRCh38
rs758431011 Chr.10: 127175835-127175835 on Build GRCh38
rs770306176 Chr.10: 127175879-127175879 on Build GRCh38
rs372417174 Chr.10: 127175702-127175702 on Build GRCh38
rs772878195 Chr.10: 127175958-127175958 on Build GRCh38
rs1049642667 Chr.10: 127175762-127175762 on Build GRCh38
rs370957941 Chr.10: 127175799-127175799 on Build GRCh38
rs202008548 Chr.10: 127175899-127175899 on Build GRCh38
rs757299305 Chr.10: 127175918-127175918 on Build GRCh38
rs770775490 Chr.10: 127175676-127175676 on Build GRCh38
rs755851502 Chr.10: 127175906-127175906 on Build GRCh38
rs547642648 Chr.10: 127175554-127175554 on Build GRCh38
rs754152980 Chr.10: 127175736-127175736 on Build GRCh38
rs370158282 Chr.10: 127175909-127175909 on Build GRCh38
rs868339889 Chr.10: 127175888-127175888 on Build GRCh38
rs781426542 Chr.10: 127175928-127175928 on Build GRCh38
rs760287894 Chr.10: 127175972-127175972 on Build GRCh38
rs371362388 Chr.10: 127175681-127175681 on Build GRCh38
rs565895276 Chr.10: 127175901-127175901 on Build GRCh38
rs780028933 Chr.10: 127175746-127175746 on Build GRCh38
rs143176564 Chr.10: 127175794-127175794 on Build GRCh38
rs776154036 Chr.10: 127175775-127175775 on Build GRCh38
rs1028170000 Chr.10: 127175855-127175856 on Build GRCh38
rs866079227 Chr.10: 127175669-127175669 on Build GRCh38
rs375773085 Chr.10: 127175755-127175755 on Build GRCh38
rs771951257 Chr.10: 127175596-127175596 on Build GRCh38
rs746126759 Chr.10: 127175929-127175929 on Build GRCh38
rs147680044 Chr.10: 127175955-127175955 on Build GRCh38
rs762884651 Chr.10: 127175620-127175620 on Build GRCh38
rs1054618287 Chr.10: 127175885-127175885 on Build GRCh38
rs551807342 Chr.10: 127175905-127175905 on Build GRCh38
rs369739654 Chr.10: 127175900-127175900 on Build GRCh38
rs773370941 Chr.10: 127175756-127175756 on Build GRCh38
rs752251853 Chr.10: 127175724-127175724 on Build GRCh38
rs143897472 Chr.10: 127175570-127175570 on Build GRCh38
rs778773796 Chr.10: 127175916-127175916 on Build GRCh38
rs373570832 Chr.10: 127175867-127175867 on Build GRCh38
rs61738666 Chr.10: 127175721-127175721 on Build GRCh38
rs201163273 Chr.10: 127175697-127175697 on Build GRCh38
rs142627987 Chr.10: 127175735-127175735 on Build GRCh38
rs201790205 Chr.10: 127175773-127175773 on Build GRCh38
rs774728310 Chr.10: 127175946-127175946 on Build GRCh38
rs749242714 Chr.10: 127175831-127175831 on Build GRCh38
rs754517108 Chr.10: 127175575-127175575 on Build GRCh38
rs973403565 Chr.10: 127175545-127175545 on Build GRCh38
rs780666441 Chr.10: 127175564-127175564 on Build GRCh38
rs753671967 Chr.10: 127175908-127175908 on Build GRCh38
rs745852102 Chr.10: 127175684-127175684 on Build GRCh38
rs377391223 Chr.10: 127175644-127175644 on Build GRCh38
rs373167144 Chr.10: 127175630-127175630 on Build GRCh38
rs142182142 Chr.10: 127175720-127175720 on Build GRCh38
rs79463082 Chr.10: 127175701-127175701 on Build GRCh38
rs376501105 Chr.10: 127175561-127175561 on Build GRCh38
rs529533671 Chr.10: 127175891-127175891 on Build GRCh38
rs911695629 Chr.10: 127175617-127175617 on Build GRCh38

View Full Product Details