Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 127175922-127176429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK1
Entrez Gene ID
1793
Gene Name
dedicator of cytokinesis 1
Gene Aliases
DOCK180, ced5
UniGene
Hs.159195
Gene Chromosome Location
Chr.10: 126905409-127452517 on Build GRCh38
Transcripts
NM_001290223.1 NM_001380.4 XM_011539422.2 XM_011539424.1 XM_017015813.1 XM_017015814.1 XM_017015815.1 XM_017015816.1 NM_001039762.2 XM_005252694.3 XM_017016537.1 XM_017016538.1 XM_017016539.1 XM_017016540.1 XM_017016541.1 XM_017016542.1 XM_017016543.1

Gene Symbol
FAM196A
Entrez Gene ID
642938
Gene Name
family with sequence similarity 196 member A
Gene Aliases
C10orf141
UniGene
Hs.613882
Gene Chromosome Location
Chr.10: 127135426-127196522 on Build GRCh38
Transcripts
NM_001290223.1 NM_001380.4 XM_011539422.2 XM_011539424.1 XM_017015813.1 XM_017015814.1 XM_017015815.1 XM_017015816.1 NM_001039762.2 XM_005252694.3 XM_017016537.1 XM_017016538.1 XM_017016539.1 XM_017016540.1 XM_017016541.1 XM_017016542.1 XM_017016543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6352840 c.39G>A p.T13T Chr.10: 127176357-127176357 on Build GRCh38
684058 c.140T>A p.V47E Chr.10: 127176256-127176256 on Build GRCh38
203655 c.295G>A p.V99I Chr.10: 127176101-127176101 on Build GRCh38
916001 c.471G>A p.E157E Chr.10: 127175925-127175925 on Build GRCh38
916003 c.142C>T p.R48W Chr.10: 127176254-127176254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767609347 Chr.10: 127176237-127176237 on Build GRCh38
rs201807593 Chr.10: 127176189-127176189 on Build GRCh38
rs112335081 Chr.10: 127176255-127176255 on Build GRCh38
rs748011955 Chr.10: 127176308-127176308 on Build GRCh38
rs374772780 Chr.10: 127176123-127176123 on Build GRCh38
rs755223872 Chr.10: 127176357-127176359 on Build GRCh38
rs747882991 Chr.10: 127176354-127176354 on Build GRCh38
rs952293347 Chr.10: 127176232-127176232 on Build GRCh38
rs745443698 Chr.10: 127176199-127176199 on Build GRCh38
rs775136991 Chr.10: 127176152-127176152 on Build GRCh38
rs772686532 Chr.10: 127176068-127176068 on Build GRCh38
rs77083569 Chr.10: 127175976-127175976 on Build GRCh38
rs940714348 Chr.10: 127175983-127175983 on Build GRCh38
rs915757581 Chr.10: 127176341-127176341 on Build GRCh38
rs759084320 Chr.10: 127176224-127176224 on Build GRCh38
rs201518235 Chr.10: 127176096-127176096 on Build GRCh38
rs368666491 Chr.10: 127176257-127176257 on Build GRCh38
rs371707213 Chr.10: 127176353-127176353 on Build GRCh38
rs367840893 Chr.10: 127176172-127176172 on Build GRCh38
rs748982319 Chr.10: 127176211-127176211 on Build GRCh38
rs772004736 Chr.10: 127176355-127176355 on Build GRCh38
rs768460062 Chr.10: 127176302-127176302 on Build GRCh38
rs939722603 Chr.10: 127176362-127176362 on Build GRCh38
rs372783138 Chr.10: 127176378-127176378 on Build GRCh38
rs776269777 Chr.10: 127176079-127176079 on Build GRCh38
rs138559550 Chr.10: 127176344-127176344 on Build GRCh38
rs763089897 Chr.10: 127176376-127176376 on Build GRCh38
rs781715631 Chr.10: 127176291-127176291 on Build GRCh38
rs757749817 Chr.10: 127176289-127176289 on Build GRCh38
rs774387247 Chr.10: 127176223-127176223 on Build GRCh38
rs201704187 Chr.10: 127176253-127176253 on Build GRCh38
rs748631088 Chr.10: 127176061-127176061 on Build GRCh38
rs747545234 Chr.10: 127176196-127176196 on Build GRCh38
rs529298057 Chr.10: 127176338-127176338 on Build GRCh38
rs372345493 Chr.10: 127176297-127176297 on Build GRCh38
rs749004305 Chr.10: 127176275-127176275 on Build GRCh38
rs775216075 Chr.10: 127176208-127176208 on Build GRCh38
rs762876030 Chr.10: 127176154-127176154 on Build GRCh38
rs146174669 Chr.10: 127176377-127176377 on Build GRCh38
rs765309721 Chr.10: 127176254-127176254 on Build GRCh38
rs781103257 Chr.10: 127176028-127176028 on Build GRCh38
rs928320163 Chr.10: 127176125-127176125 on Build GRCh38
rs538123042 Chr.10: 127176342-127176342 on Build GRCh38
rs779422344 Chr.10: 127176051-127176051 on Build GRCh38
rs772671869 Chr.10: 127176063-127176063 on Build GRCh38
rs756270808 Chr.10: 127176107-127176107 on Build GRCh38
rs567593902 Chr.10: 127176336-127176336 on Build GRCh38
rs762779861 Chr.10: 127176102-127176102 on Build GRCh38
rs770823920 Chr.10: 127176360-127176360 on Build GRCh38
rs755543710 Chr.10: 127176337-127176337 on Build GRCh38
rs769537771 Chr.10: 127176149-127176149 on Build GRCh38
rs538792856 Chr.10: 127176236-127176236 on Build GRCh38
rs1007997134 Chr.10: 127176057-127176057 on Build GRCh38
rs751970879 Chr.10: 127176276-127176276 on Build GRCh38
rs757802603 Chr.10: 127176198-127176198 on Build GRCh38
rs141830486 Chr.10: 127176142-127176142 on Build GRCh38
rs11594560 Chr.10: 127175956-127175956 on Build GRCh38
rs778319223 Chr.10: 127176127-127176127 on Build GRCh38
rs150007430 Chr.10: 127175942-127175942 on Build GRCh38
rs748989064 Chr.10: 127175943-127175943 on Build GRCh38
rs367902185 Chr.10: 127176018-127176018 on Build GRCh38
rs370769092 Chr.10: 127176031-127176031 on Build GRCh38
rs753847704 Chr.10: 127176306-127176307 on Build GRCh38
rs148136993 Chr.10: 127176274-127176274 on Build GRCh38
rs762172825 Chr.10: 127176335-127176335 on Build GRCh38
rs745480635 Chr.10: 127176148-127176148 on Build GRCh38
rs79489715 Chr.10: 127176321-127176321 on Build GRCh38
rs977355583 Chr.10: 127176069-127176069 on Build GRCh38
rs751919597 Chr.10: 127176333-127176333 on Build GRCh38
rs772878195 Chr.10: 127175958-127175958 on Build GRCh38
rs989910874 Chr.10: 127176086-127176086 on Build GRCh38
rs889229055 Chr.10: 127176188-127176188 on Build GRCh38
rs549319983 Chr.10: 127176143-127176143 on Build GRCh38
rs746449530 Chr.10: 127176134-127176134 on Build GRCh38
rs372715738 Chr.10: 127176178-127176178 on Build GRCh38
rs754130370 Chr.10: 127176117-127176117 on Build GRCh38
rs150536495 Chr.10: 127176305-127176305 on Build GRCh38
rs143780805 Chr.10: 127176179-127176179 on Build GRCh38
rs754007845 Chr.10: 127176177-127176177 on Build GRCh38
rs760287894 Chr.10: 127175972-127175972 on Build GRCh38
rs750530104 Chr.10: 127176104-127176104 on Build GRCh38
rs139369079 Chr.10: 127176101-127176101 on Build GRCh38
rs768912755 Chr.10: 127176045-127176045 on Build GRCh38
rs757321590 Chr.10: 127176023-127176023 on Build GRCh38
rs764060092 Chr.10: 127176162-127176162 on Build GRCh38
rs768598681 Chr.10: 127176217-127176217 on Build GRCh38
rs76646118 Chr.10: 127176323-127176323 on Build GRCh38
rs762119851 Chr.10: 127176379-127176379 on Build GRCh38
rs147680044 Chr.10: 127175955-127175955 on Build GRCh38
rs760783594 Chr.10: 127176167-127176167 on Build GRCh38
rs746933569 Chr.10: 127176359-127176359 on Build GRCh38
rs2489386 Chr.10: 127176357-127176357 on Build GRCh38
rs749601333 Chr.10: 127176038-127176038 on Build GRCh38
rs200747773 Chr.10: 127176084-127176084 on Build GRCh38
rs769939553 Chr.10: 127176318-127176318 on Build GRCh38
rs770566069 Chr.10: 127176073-127176073 on Build GRCh38
rs780369218 Chr.10: 127176115-127176115 on Build GRCh38
rs759475811 Chr.10: 127176315-127176315 on Build GRCh38
rs541317255 Chr.10: 127176351-127176351 on Build GRCh38
rs779113272 Chr.10: 127176119-127176119 on Build GRCh38
rs759392442 Chr.10: 127176017-127176017 on Build GRCh38
rs117242435 Chr.10: 127176046-127176046 on Build GRCh38
rs759326307 Chr.10: 127176083-127176083 on Build GRCh38
rs774728310 Chr.10: 127175946-127175946 on Build GRCh38
rs753615944 Chr.10: 127176004-127176004 on Build GRCh38
rs770696831 Chr.10: 127176292-127176292 on Build GRCh38
rs748593127 Chr.10: 127176122-127176122 on Build GRCh38
rs776558943 Chr.10: 127176238-127176238 on Build GRCh38
rs200815248 Chr.10: 127176164-127176164 on Build GRCh38
rs773204442 Chr.10: 127176313-127176313 on Build GRCh38
rs769329853 Chr.10: 127176200-127176200 on Build GRCh38
rs756393627 Chr.10: 127176032-127176032 on Build GRCh38
rs755325636 Chr.10: 127176118-127176118 on Build GRCh38
rs772129249 Chr.10: 127176312-127176312 on Build GRCh38
rs199952144 Chr.10: 127176213-127176213 on Build GRCh38
rs534565452 Chr.10: 127176072-127176072 on Build GRCh38
rs775627557 Chr.10: 127176369-127176369 on Build GRCh38
rs778825802 Chr.10: 127176306-127176306 on Build GRCh38
rs375404920 Chr.10: 127176187-127176187 on Build GRCh38
rs141706041 Chr.10: 127176300-127176300 on Build GRCh38

View Full Product Details