Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 290589-291088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGGHG
Entrez Gene ID
80162
Gene Name
protein-glucosylgalactosylhydroxylysine glucosidase
Gene Aliases
ATHL1
UniGene
Hs.353181
Gene Chromosome Location
Chr.11: 287760-295688 on Build GRCh38
Transcripts
NM_025092.4 XM_011520383.2 XM_011520384.2 XM_011520385.2 XM_011520386.2 XM_011520387.2 XM_017018355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3979552 c.757C>G p.R253G Chr.11: 290964-290964 on Build GRCh38
5831731 c.507_508insG p.P172fs*22 Chr.11: 290714-290715 on Build GRCh38
4032267 c.714C>G p.L238L Chr.11: 290921-290921 on Build GRCh38
6606772 c.787C>A p.L263M Chr.11: 290994-290994 on Build GRCh38
4958156 c.823T>C p.Y275H Chr.11: 291030-291030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369804174 Chr.11: 290952-290952 on Build GRCh38
rs761188249 Chr.11: 291026-291026 on Build GRCh38
rs377492501 Chr.11: 290850-290850 on Build GRCh38
rs370664133 Chr.11: 291029-291029 on Build GRCh38
rs753503749 Chr.11: 290798-290798 on Build GRCh38
rs369270826 Chr.11: 290800-290800 on Build GRCh38
rs746436580 Chr.11: 291011-291011 on Build GRCh38
rs529572127 Chr.11: 290893-290893 on Build GRCh38
rs756483084 Chr.11: 290683-290683 on Build GRCh38
rs778959511 Chr.11: 290909-290909 on Build GRCh38
rs376440498 Chr.11: 290699-290699 on Build GRCh38
rs757991282 Chr.11: 290808-290808 on Build GRCh38
rs145342416 Chr.11: 290913-290913 on Build GRCh38
rs372433025 Chr.11: 290828-290828 on Build GRCh38
rs978246689 Chr.11: 290875-290875 on Build GRCh38
rs750931203 Chr.11: 290760-290760 on Build GRCh38
rs755812817 Chr.11: 290964-290964 on Build GRCh38
rs143771488 Chr.11: 291063-291063 on Build GRCh38
rs114375326 Chr.11: 290802-290802 on Build GRCh38
rs370178886 Chr.11: 290966-290966 on Build GRCh38
rs950682709 Chr.11: 290761-290761 on Build GRCh38
rs151001509 Chr.11: 290847-290847 on Build GRCh38
rs542864967 Chr.11: 290682-290682 on Build GRCh38
rs767952967 Chr.11: 290739-290739 on Build GRCh38
rs746738497 Chr.11: 290718-290718 on Build GRCh38
rs10902120 Chr.11: 290816-290816 on Build GRCh38
rs773299991 Chr.11: 290716-290716 on Build GRCh38
rs763779735 Chr.11: 291021-291021 on Build GRCh38
rs573768364 Chr.11: 290977-290977 on Build GRCh38
rs553210478 Chr.11: 291058-291058 on Build GRCh38
rs748770834 Chr.11: 290629-290629 on Build GRCh38
rs759961334 Chr.11: 290886-290886 on Build GRCh38
rs556885979 Chr.11: 290975-290975 on Build GRCh38
rs768052220 Chr.11: 290639-290639 on Build GRCh38
rs72636978 Chr.11: 290918-290918 on Build GRCh38
rs984846977 Chr.11: 290763-290763 on Build GRCh38
rs376282580 Chr.11: 291052-291052 on Build GRCh38
rs773209706 Chr.11: 290660-290660 on Build GRCh38
rs771078047 Chr.11: 290999-290999 on Build GRCh38
rs901637826 Chr.11: 290720-290720 on Build GRCh38
rs72636977 Chr.11: 290891-290891 on Build GRCh38
rs769592797 Chr.11: 290715-290731 on Build GRCh38
rs779611436 Chr.11: 290684-290684 on Build GRCh38
rs763134680 Chr.11: 290722-290722 on Build GRCh38
rs756113972 Chr.11: 291044-291044 on Build GRCh38
rs748054613 Chr.11: 290646-290646 on Build GRCh38
rs202237606 Chr.11: 290797-290797 on Build GRCh38
rs765656541 Chr.11: 290936-290936 on Build GRCh38
rs759783981 Chr.11: 290931-290931 on Build GRCh38
rs746965720 Chr.11: 290815-290815 on Build GRCh38
rs762680096 Chr.11: 291018-291018 on Build GRCh38
rs779525417 Chr.11: 290627-290627 on Build GRCh38
rs373537355 Chr.11: 290965-290965 on Build GRCh38
rs1041117526 Chr.11: 290902-290902 on Build GRCh38
rs564622719 Chr.11: 290822-290822 on Build GRCh38
rs752085391 Chr.11: 290671-290671 on Build GRCh38
rs545000615 Chr.11: 290772-290772 on Build GRCh38
rs145779251 Chr.11: 290942-290942 on Build GRCh38
rs572792323 Chr.11: 290721-290721 on Build GRCh38
rs766732835 Chr.11: 290874-290874 on Build GRCh38
rs114473173 Chr.11: 290955-290955 on Build GRCh38
rs777813224 Chr.11: 290813-290813 on Build GRCh38
rs762735613 Chr.11: 290943-290943 on Build GRCh38
rs776958341 Chr.11: 291003-291003 on Build GRCh38
rs777987620 Chr.11: 290983-290983 on Build GRCh38
rs762368477 Chr.11: 290737-290737 on Build GRCh38
rs140256419 Chr.11: 290901-290901 on Build GRCh38
rs766843025 Chr.11: 290644-290644 on Build GRCh38
rs759823292 Chr.11: 290664-290664 on Build GRCh38
rs902785150 Chr.11: 290654-290654 on Build GRCh38
rs267602839 Chr.11: 290984-290984 on Build GRCh38
rs771873020 Chr.11: 290659-290659 on Build GRCh38
rs751170554 Chr.11: 290951-290951 on Build GRCh38
rs780857717 Chr.11: 290628-290628 on Build GRCh38
rs765501558 Chr.11: 290667-290667 on Build GRCh38
rs752094767 Chr.11: 290801-290801 on Build GRCh38
rs757328725 Chr.11: 290900-290900 on Build GRCh38
rs890414409 Chr.11: 291033-291033 on Build GRCh38
rs769635958 Chr.11: 290908-290908 on Build GRCh38
rs745319007 Chr.11: 290694-290695 on Build GRCh38
rs936966688 Chr.11: 290889-290889 on Build GRCh38
rs377335658 Chr.11: 291015-291015 on Build GRCh38
rs1015962072 Chr.11: 290769-290769 on Build GRCh38
rs200688444 Chr.11: 290823-290823 on Build GRCh38
rs761215956 Chr.11: 290868-290868 on Build GRCh38
rs768748877 Chr.11: 290736-290736 on Build GRCh38
rs149921679 Chr.11: 290715-290715 on Build GRCh38
rs773317933 Chr.11: 290921-290921 on Build GRCh38
rs766154794 Chr.11: 291045-291045 on Build GRCh38
rs774965357 Chr.11: 290715-290715 on Build GRCh38
rs140895331 Chr.11: 290877-290877 on Build GRCh38
rs889854093 Chr.11: 290702-290702 on Build GRCh38
rs775861335 Chr.11: 290668-290668 on Build GRCh38
rs762280608 Chr.11: 290747-290748 on Build GRCh38
rs767862686 Chr.11: 290674-290674 on Build GRCh38
rs754460238 Chr.11: 290708-290708 on Build GRCh38
rs774913967 Chr.11: 290925-290925 on Build GRCh38
rs372880922 Chr.11: 290693-290693 on Build GRCh38
rs879089088 Chr.11: 290920-290920 on Build GRCh38
rs199731720 Chr.11: 291040-291040 on Build GRCh38
rs771288111 Chr.11: 290928-290928 on Build GRCh38
rs767289701 Chr.11: 290953-290953 on Build GRCh38
rs745591761 Chr.11: 290905-290905 on Build GRCh38
rs750302296 Chr.11: 291039-291039 on Build GRCh38
rs563385736 Chr.11: 290894-290894 on Build GRCh38
rs150075556 Chr.11: 290888-290888 on Build GRCh38
rs927574054 Chr.11: 290907-290907 on Build GRCh38
rs201239213 Chr.11: 290948-290948 on Build GRCh38
rs963505068 Chr.11: 290759-290759 on Build GRCh38
rs774149920 Chr.11: 291022-291022 on Build GRCh38
rs772303912 Chr.11: 291015-291015 on Build GRCh38
rs751311658 Chr.11: 290688-290688 on Build GRCh38
rs997328459 Chr.11: 290916-290916 on Build GRCh38
rs778332421 Chr.11: 290713-290713 on Build GRCh38

View Full Product Details