Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 292738-293232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGGHG
Entrez Gene ID
80162
Gene Name
protein-glucosylgalactosylhydroxylysine glucosidase
Gene Aliases
ATHL1
UniGene
Hs.353181
Gene Chromosome Location
Chr.11: 287760-295688 on Build GRCh38
Transcripts
NM_025092.4 XM_011520383.2 XM_011520384.2 XM_011520385.2 XM_011520386.2 XM_011520387.2 XM_017018355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1585679 c.1174C>T p.R392* Chr.11: 292901-292901 on Build GRCh38
7356836 c.704A>C p.E235A Chr.11: 292962-292962 on Build GRCh38
1585678 c.1338G>A p.Q446Q Chr.11: 293230-293230 on Build GRCh38
5815856 c.1203G>A p.R401R Chr.11: 292930-292930 on Build GRCh38
4165759 c.740-4C>A p.? Chr.11: 293159-293159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767417994 Chr.11: 293006-293006 on Build GRCh38
rs371578139 Chr.11: 293014-293014 on Build GRCh38
rs754322093 Chr.11: 293147-293147 on Build GRCh38
rs373890304 Chr.11: 293137-293137 on Build GRCh38
rs879014487 Chr.11: 292879-292879 on Build GRCh38
rs756982817 Chr.11: 292835-292835 on Build GRCh38
rs758706797 Chr.11: 293019-293019 on Build GRCh38
rs967541951 Chr.11: 292968-292968 on Build GRCh38
rs145280533 Chr.11: 292973-292973 on Build GRCh38
rs773541695 Chr.11: 292855-292855 on Build GRCh38
rs371300411 Chr.11: 293177-293177 on Build GRCh38
rs750468855 Chr.11: 293134-293134 on Build GRCh38
rs897271779 Chr.11: 292821-292821 on Build GRCh38
rs374388402 Chr.11: 292922-292922 on Build GRCh38
rs761874563 Chr.11: 292895-292895 on Build GRCh38
rs1035428612 Chr.11: 293191-293191 on Build GRCh38
rs112633783 Chr.11: 293023-293023 on Build GRCh38
rs775263042 Chr.11: 292956-292956 on Build GRCh38
rs758196372 Chr.11: 292831-292831 on Build GRCh38
rs142215595 Chr.11: 292887-292887 on Build GRCh38
rs757988892 Chr.11: 293175-293175 on Build GRCh38
rs115207440 Chr.11: 292861-292861 on Build GRCh38
rs368063489 Chr.11: 293142-293142 on Build GRCh38
rs548505527 Chr.11: 293101-293101 on Build GRCh38
rs752913524 Chr.11: 293016-293016 on Build GRCh38
rs755326444 Chr.11: 293095-293095 on Build GRCh38
rs751755330 Chr.11: 293083-293083 on Build GRCh38
rs146770651 Chr.11: 292892-292892 on Build GRCh38
rs568096396 Chr.11: 292809-292809 on Build GRCh38
rs758268420 Chr.11: 293126-293127 on Build GRCh38
rs772168575 Chr.11: 292854-292854 on Build GRCh38
rs1032754211 Chr.11: 293157-293157 on Build GRCh38
rs779185127 Chr.11: 293154-293154 on Build GRCh38
rs375624960 Chr.11: 293102-293102 on Build GRCh38
rs753935715 Chr.11: 292969-292969 on Build GRCh38
rs779120646 Chr.11: 292848-292848 on Build GRCh38
rs140222962 Chr.11: 292962-292962 on Build GRCh38
rs760410673 Chr.11: 293013-293013 on Build GRCh38
rs777407751 Chr.11: 293176-293176 on Build GRCh38
rs371621845 Chr.11: 293117-293117 on Build GRCh38
rs1035396168 Chr.11: 292972-292972 on Build GRCh38
rs765479578 Chr.11: 293016-293016 on Build GRCh38
rs570565217 Chr.11: 292836-292836 on Build GRCh38
rs913463388 Chr.11: 292779-292779 on Build GRCh38
rs538556338 Chr.11: 292902-292902 on Build GRCh38
rs759992696 Chr.11: 292868-292868 on Build GRCh38
rs756264753 Chr.11: 293135-293135 on Build GRCh38
rs370862533 Chr.11: 292837-292837 on Build GRCh38
rs11602423 Chr.11: 292798-292798 on Build GRCh38
rs931187039 Chr.11: 293169-293169 on Build GRCh38
rs770255000 Chr.11: 293037-293037 on Build GRCh38
rs752838756 Chr.11: 293061-293061 on Build GRCh38
rs780718419 Chr.11: 293091-293091 on Build GRCh38
rs773876628 Chr.11: 293048-293048 on Build GRCh38
rs771504751 Chr.11: 293067-293067 on Build GRCh38
rs771411404 Chr.11: 292932-292932 on Build GRCh38
rs777871404 Chr.11: 293022-293022 on Build GRCh38
rs140787613 Chr.11: 293193-293193 on Build GRCh38
rs776583068 Chr.11: 293108-293108 on Build GRCh38
rs779067390 Chr.11: 292907-292907 on Build GRCh38
rs758449268 Chr.11: 292921-292921 on Build GRCh38
rs538101900 Chr.11: 293069-293069 on Build GRCh38
rs772373475 Chr.11: 293088-293088 on Build GRCh38
rs780628796 Chr.11: 293041-293041 on Build GRCh38
rs755345955 Chr.11: 293149-293149 on Build GRCh38
rs764881216 Chr.11: 292832-292832 on Build GRCh38
rs373964830 Chr.11: 292856-292856 on Build GRCh38
rs769508282 Chr.11: 292883-292883 on Build GRCh38
rs542587412 Chr.11: 293076-293076 on Build GRCh38
rs764338395 Chr.11: 293079-293079 on Build GRCh38
rs978766392 Chr.11: 293081-293081 on Build GRCh38
rs750965016 Chr.11: 293089-293089 on Build GRCh38
rs771197997 Chr.11: 292857-292857 on Build GRCh38
rs117666347 Chr.11: 293096-293096 on Build GRCh38
rs760645336 Chr.11: 293072-293072 on Build GRCh38
rs370073465 Chr.11: 292869-292869 on Build GRCh38
rs150442930 Chr.11: 293210-293210 on Build GRCh38
rs561846271 Chr.11: 293106-293106 on Build GRCh38
rs767199549 Chr.11: 292901-292901 on Build GRCh38
rs779493472 Chr.11: 293077-293077 on Build GRCh38
rs781691940 Chr.11: 293025-293025 on Build GRCh38
rs746379558 Chr.11: 293085-293085 on Build GRCh38
rs200591136 Chr.11: 292971-292971 on Build GRCh38
rs11246056 Chr.11: 292805-292805 on Build GRCh38
rs746537072 Chr.11: 293105-293105 on Build GRCh38
rs12801980 Chr.11: 293188-293188 on Build GRCh38
rs369165114 Chr.11: 292853-292853 on Build GRCh38
rs777428964 Chr.11: 293199-293210 on Build GRCh38
rs546669379 Chr.11: 293126-293126 on Build GRCh38
rs747832026 Chr.11: 293172-293172 on Build GRCh38
rs773109050 Chr.11: 293068-293068 on Build GRCh38
rs35259273 Chr.11: 293001-293002 on Build GRCh38
rs989113188 Chr.11: 292778-292778 on Build GRCh38
rs770451652 Chr.11: 293180-293180 on Build GRCh38
rs1043428459 Chr.11: 292802-292802 on Build GRCh38
rs761239181 Chr.11: 292996-292996 on Build GRCh38
rs897081635 Chr.11: 293155-293155 on Build GRCh38
rs866813005 Chr.11: 293195-293195 on Build GRCh38
rs376887602 Chr.11: 293009-293009 on Build GRCh38
rs749593314 Chr.11: 293092-293092 on Build GRCh38
rs748932118 Chr.11: 293045-293045 on Build GRCh38
rs999915410 Chr.11: 293141-293141 on Build GRCh38
rs543435538 Chr.11: 292789-292789 on Build GRCh38
rs756740748 Chr.11: 293090-293090 on Build GRCh38
rs1037247636 Chr.11: 293130-293130 on Build GRCh38
rs576943817 Chr.11: 292998-292998 on Build GRCh38
rs377674052 Chr.11: 292838-292838 on Build GRCh38
rs747653005 Chr.11: 293060-293060 on Build GRCh38
rs554959633 Chr.11: 292933-292933 on Build GRCh38
rs755906539 Chr.11: 292906-292906 on Build GRCh38
rs774605192 Chr.11: 293118-293118 on Build GRCh38
rs971003098 Chr.11: 293043-293043 on Build GRCh38
rs767688976 Chr.11: 293127-293127 on Build GRCh38
rs878937063 Chr.11: 292880-292880 on Build GRCh38
rs559995922 Chr.11: 292991-292991 on Build GRCh38
rs762506921 Chr.11: 292888-292888 on Build GRCh38
rs889593256 Chr.11: 292969-292969 on Build GRCh38
rs770309826 Chr.11: 292955-292955 on Build GRCh38
rs759330127 Chr.11: 293116-293116 on Build GRCh38
rs746428216 Chr.11: 293026-293026 on Build GRCh38
rs763712888 Chr.11: 292889-292889 on Build GRCh38
rs1036979320 Chr.11: 292882-292882 on Build GRCh38
rs752937899 Chr.11: 292912-292912 on Build GRCh38
rs746199014 Chr.11: 292940-292940 on Build GRCh38
rs533545798 Chr.11: 292820-292820 on Build GRCh38
rs547376637 Chr.11: 292823-292823 on Build GRCh38

View Full Product Details