Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 869782-870287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHID1
Entrez Gene ID
66005
Gene Name
chitinase domain containing 1
Gene Aliases
GL008, SI-CLP, SICLP
UniGene
Hs.144468
Gene Chromosome Location
Chr.11: 867859-915058 on Build GRCh38
Transcripts
NM_001142674.1 NM_001142675.1 NM_001142676.1 NM_001142677.1 NM_023947.3 XM_005253072.2 XM_005253073.4 XM_006718282.2 XM_011520299.1 XM_011520300.1 XM_017018166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2221925 c.1122C>G p.G374G Chr.11: 869918-869918 on Build GRCh38
6628383 c.1122C>T p.G374G Chr.11: 869918-869918 on Build GRCh38
5373761 c.1083+1G>A p.? Chr.11: 870120-870120 on Build GRCh38
2149267 c.1103A>G p.E368G Chr.11: 869937-869937 on Build GRCh38
6628376 c.1213G>A p.V405I Chr.11: 869917-869917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756897842 Chr.11: 870195-870195 on Build GRCh38
rs940226890 Chr.11: 870259-870259 on Build GRCh38
rs759370428 Chr.11: 870182-870182 on Build GRCh38
rs780648242 Chr.11: 869858-869859 on Build GRCh38
rs556360418 Chr.11: 869839-869839 on Build GRCh38
rs754969137 Chr.11: 869990-869990 on Build GRCh38
rs751161259 Chr.11: 870116-870116 on Build GRCh38
rs568144760 Chr.11: 870076-870076 on Build GRCh38
rs748714060 Chr.11: 870144-870144 on Build GRCh38
rs142286202 Chr.11: 869944-869944 on Build GRCh38
rs769175892 Chr.11: 870075-870075 on Build GRCh38
rs202154539 Chr.11: 870097-870097 on Build GRCh38
rs771863477 Chr.11: 869937-869937 on Build GRCh38
rs750014050 Chr.11: 869975-869975 on Build GRCh38
rs763441480 Chr.11: 869948-869948 on Build GRCh38
rs545997593 Chr.11: 869892-869892 on Build GRCh38
rs778633553 Chr.11: 869928-869928 on Build GRCh38
rs745897578 Chr.11: 869973-869973 on Build GRCh38
rs371660142 Chr.11: 869870-869870 on Build GRCh38
rs370593417 Chr.11: 870156-870156 on Build GRCh38
rs750125161 Chr.11: 870206-870206 on Build GRCh38
rs754145886 Chr.11: 869907-869907 on Build GRCh38
rs755324095 Chr.11: 870210-870210 on Build GRCh38
rs761978057 Chr.11: 869955-869955 on Build GRCh38
rs1037987785 Chr.11: 869838-869838 on Build GRCh38
rs773470775 Chr.11: 870088-870088 on Build GRCh38
rs745740853 Chr.11: 869830-869830 on Build GRCh38
rs757477413 Chr.11: 869906-869906 on Build GRCh38
rs777138598 Chr.11: 869869-869869 on Build GRCh38
rs765170022 Chr.11: 870187-870187 on Build GRCh38
rs749233909 Chr.11: 869921-869921 on Build GRCh38
rs377421622 Chr.11: 869807-869807 on Build GRCh38
rs754109159 Chr.11: 869891-869917 on Build GRCh38
rs754840899 Chr.11: 870117-870117 on Build GRCh38
rs1051277546 Chr.11: 869833-869833 on Build GRCh38
rs920153613 Chr.11: 870266-870266 on Build GRCh38
rs777998047 Chr.11: 869809-869809 on Build GRCh38
rs980054010 Chr.11: 870013-870013 on Build GRCh38
rs148160696 Chr.11: 869837-869837 on Build GRCh38
rs767245236 Chr.11: 869915-869915 on Build GRCh38
rs182950163 Chr.11: 869863-869863 on Build GRCh38
rs770193297 Chr.11: 870006-870006 on Build GRCh38
rs747733825 Chr.11: 869929-869929 on Build GRCh38
rs111245740 Chr.11: 870244-870244 on Build GRCh38
rs200870523 Chr.11: 869828-869828 on Build GRCh38
rs1047934105 Chr.11: 870115-870115 on Build GRCh38
rs560853222 Chr.11: 870011-870011 on Build GRCh38
rs762991237 Chr.11: 870191-870191 on Build GRCh38
rs934223496 Chr.11: 869895-869895 on Build GRCh38
rs776443991 Chr.11: 870166-870166 on Build GRCh38
rs143781870 Chr.11: 869836-869836 on Build GRCh38
rs764097989 Chr.11: 869953-869953 on Build GRCh38
rs755395155 Chr.11: 869953-869954 on Build GRCh38
rs779051397 Chr.11: 869973-869974 on Build GRCh38
rs563877459 Chr.11: 870264-870264 on Build GRCh38
rs908714079 Chr.11: 870254-870254 on Build GRCh38
rs375375230 Chr.11: 869846-869846 on Build GRCh38
rs187154087 Chr.11: 869918-869918 on Build GRCh38
rs751780638 Chr.11: 869893-869893 on Build GRCh38
rs780290124 Chr.11: 870073-870073 on Build GRCh38
rs188873073 Chr.11: 870193-870193 on Build GRCh38
rs570735805 Chr.11: 870103-870103 on Build GRCh38
rs372856810 Chr.11: 869988-869988 on Build GRCh38
rs573982321 Chr.11: 869998-869998 on Build GRCh38
rs373291423 Chr.11: 870077-870077 on Build GRCh38
rs555520541 Chr.11: 870194-870194 on Build GRCh38
rs200243605 Chr.11: 869959-869959 on Build GRCh38
rs750652235 Chr.11: 869961-869961 on Build GRCh38
rs775388800 Chr.11: 869880-869880 on Build GRCh38
rs771825733 Chr.11: 870227-870227 on Build GRCh38
rs867162831 Chr.11: 869934-869937 on Build GRCh38
rs369487093 Chr.11: 870111-870111 on Build GRCh38
rs754522495 Chr.11: 869922-869922 on Build GRCh38
rs572134914 Chr.11: 870199-870199 on Build GRCh38
rs201415316 Chr.11: 870094-870094 on Build GRCh38
rs763053936 Chr.11: 869885-869885 on Build GRCh38
rs957446169 Chr.11: 870027-870027 on Build GRCh38
rs781179463 Chr.11: 869854-869854 on Build GRCh38
rs114900330 Chr.11: 869808-869808 on Build GRCh38
rs373432738 Chr.11: 870189-870189 on Build GRCh38
rs752274383 Chr.11: 870000-870000 on Build GRCh38
rs138166915 Chr.11: 870141-870141 on Build GRCh38
rs374883925 Chr.11: 869943-869943 on Build GRCh38
rs905764501 Chr.11: 870142-870142 on Build GRCh38
rs200188854 Chr.11: 870001-870001 on Build GRCh38
rs113580509 Chr.11: 870095-870095 on Build GRCh38
rs376166791 Chr.11: 870213-870213 on Build GRCh38
rs141753287 Chr.11: 870095-870095 on Build GRCh38
rs771415920 Chr.11: 869856-869856 on Build GRCh38
rs1044084682 Chr.11: 870084-870084 on Build GRCh38
rs760496947 Chr.11: 869967-869967 on Build GRCh38
rs541029433 Chr.11: 870246-870246 on Build GRCh38
rs768091549 Chr.11: 869840-869840 on Build GRCh38
rs371419951 Chr.11: 869974-869974 on Build GRCh38
rs770175300 Chr.11: 870041-870041 on Build GRCh38
rs765793433 Chr.11: 869871-869871 on Build GRCh38
rs748585661 Chr.11: 870215-870215 on Build GRCh38
rs115994760 Chr.11: 870136-870136 on Build GRCh38
rs562937742 Chr.11: 870031-870031 on Build GRCh38
rs141109433 Chr.11: 869977-869977 on Build GRCh38
rs746685245 Chr.11: 870004-870004 on Build GRCh38
rs1023102864 Chr.11: 869873-869873 on Build GRCh38
rs140476082 Chr.11: 870158-870158 on Build GRCh38
rs1032977173 Chr.11: 870148-870148 on Build GRCh38
rs773026411 Chr.11: 870238-870238 on Build GRCh38
rs556045828 Chr.11: 869917-869917 on Build GRCh38
rs948349166 Chr.11: 870081-870081 on Build GRCh38
rs747866649 Chr.11: 869850-869850 on Build GRCh38
rs757036352 Chr.11: 869825-869825 on Build GRCh38
rs886359843 Chr.11: 869989-869989 on Build GRCh38
rs201931075 Chr.11: 870074-870074 on Build GRCh38
rs955630312 Chr.11: 870237-870237 on Build GRCh38
rs144295528 Chr.11: 870154-870154 on Build GRCh38
rs759497130 Chr.11: 870102-870102 on Build GRCh38
rs750256778 Chr.11: 869916-869916 on Build GRCh38
rs780419403 Chr.11: 870120-870120 on Build GRCh38
rs986928767 Chr.11: 870243-870243 on Build GRCh38
rs769049099 Chr.11: 870137-870137 on Build GRCh38
rs376509515 Chr.11: 870087-870087 on Build GRCh38
rs746273102 Chr.11: 869942-869942 on Build GRCh38
rs150184057 Chr.11: 870028-870028 on Build GRCh38
rs751815626 Chr.11: 869819-869819 on Build GRCh38
rs115326504 Chr.11: 870040-870040 on Build GRCh38
rs986944190 Chr.11: 870257-870257 on Build GRCh38
rs774431066 Chr.11: 869954-869954 on Build GRCh38
rs987080839 Chr.11: 870072-870072 on Build GRCh38
rs748378823 Chr.11: 870090-870090 on Build GRCh38
rs532653739 Chr.11: 870015-870015 on Build GRCh38
rs149585062 Chr.11: 870145-870145 on Build GRCh38

View Full Product Details