Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 976969-977466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2A2
Entrez Gene ID
161
Gene Name
adaptor related protein complex 2 alpha 2 subunit
Gene Aliases
ADTAB, CLAPA2, HIP-9, HIP9, HYPJ
UniGene
Hs.19121
Gene Chromosome Location
Chr.11: 925809-1012245 on Build GRCh38
Transcripts
NM_001242837.1 NM_012305.3 XM_011519929.1 XM_011519930.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5065780 c.485A>G p.D162G Chr.11: 977106-977106 on Build GRCh38
1183126 c.565A>G p.T189A Chr.11: 977186-977186 on Build GRCh38
3954011 c.492G>A p.V164V Chr.11: 977113-977113 on Build GRCh38
318714 c.536C>T p.S179F Chr.11: 977157-977157 on Build GRCh38
1358044 c.572G>A p.R191Q Chr.11: 977193-977193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757988092 Chr.11: 977155-977155 on Build GRCh38
rs1037673431 Chr.11: 977278-977278 on Build GRCh38
rs776376498 Chr.11: 977238-977238 on Build GRCh38
rs771596836 Chr.11: 977081-977081 on Build GRCh38
rs755959634 Chr.11: 977063-977063 on Build GRCh38
rs1016398569 Chr.11: 977042-977042 on Build GRCh38
rs565356652 Chr.11: 977323-977323 on Build GRCh38
rs1006216264 Chr.11: 977037-977037 on Build GRCh38
rs567073979 Chr.11: 977394-977394 on Build GRCh38
rs542944302 Chr.11: 977162-977162 on Build GRCh38
rs373655031 Chr.11: 977134-977134 on Build GRCh38
rs774980955 Chr.11: 977174-977174 on Build GRCh38
rs980107183 Chr.11: 977024-977024 on Build GRCh38
rs376831929 Chr.11: 977049-977049 on Build GRCh38
rs187108877 Chr.11: 977048-977048 on Build GRCh38
rs374082996 Chr.11: 977203-977203 on Build GRCh38
rs926055314 Chr.11: 977025-977025 on Build GRCh38
rs1045582181 Chr.11: 977355-977355 on Build GRCh38
rs1031201339 Chr.11: 977086-977086 on Build GRCh38
rs376931521 Chr.11: 977059-977059 on Build GRCh38
rs775185111 Chr.11: 977252-977252 on Build GRCh38
rs779961122 Chr.11: 977067-977067 on Build GRCh38
rs993161820 Chr.11: 977008-977008 on Build GRCh38
rs770352763 Chr.11: 977101-977101 on Build GRCh38
rs777534562 Chr.11: 977082-977082 on Build GRCh38
rs748170846 Chr.11: 977179-977179 on Build GRCh38
rs1046994249 Chr.11: 977026-977026 on Build GRCh38
rs901103135 Chr.11: 977013-977013 on Build GRCh38
rs114731185 Chr.11: 977322-977322 on Build GRCh38
rs772196174 Chr.11: 977193-977193 on Build GRCh38
rs200699117 Chr.11: 977253-977253 on Build GRCh38
rs201561885 Chr.11: 977068-977068 on Build GRCh38
rs942550350 Chr.11: 977006-977006 on Build GRCh38
rs751748343 Chr.11: 977052-977052 on Build GRCh38
rs909796731 Chr.11: 977268-977268 on Build GRCh38
rs1024217713 Chr.11: 977015-977015 on Build GRCh38
rs6597960 Chr.11: 977036-977036 on Build GRCh38
rs116495280 Chr.11: 976990-976990 on Build GRCh38
rs752129687 Chr.11: 977154-977154 on Build GRCh38
rs879057921 Chr.11: 977313-977313 on Build GRCh38
rs774326430 Chr.11: 977119-977119 on Build GRCh38
rs1010540270 Chr.11: 977381-977381 on Build GRCh38
rs999641192 Chr.11: 977351-977351 on Build GRCh38
rs1047509101 Chr.11: 977334-977335 on Build GRCh38
rs535949215 Chr.11: 977408-977408 on Build GRCh38
rs951231123 Chr.11: 977407-977407 on Build GRCh38
rs778305562 Chr.11: 977069-977069 on Build GRCh38
rs537293684 Chr.11: 977380-977380 on Build GRCh38
rs373069772 Chr.11: 977088-977088 on Build GRCh38
rs910967465 Chr.11: 977005-977005 on Build GRCh38
rs987267694 Chr.11: 977438-977438 on Build GRCh38
rs942013932 Chr.11: 977271-977271 on Build GRCh38
rs776199398 Chr.11: 977045-977045 on Build GRCh38
rs970502632 Chr.11: 977016-977016 on Build GRCh38
rs911353367 Chr.11: 977272-977272 on Build GRCh38
rs777289425 Chr.11: 977159-977159 on Build GRCh38
rs1041242562 Chr.11: 977007-977007 on Build GRCh38
rs371169279 Chr.11: 977171-977171 on Build GRCh38
rs929856673 Chr.11: 977023-977023 on Build GRCh38
rs1032892933 Chr.11: 977384-977384 on Build GRCh38
rs929811250 Chr.11: 977210-977210 on Build GRCh38
rs376826459 Chr.11: 977141-977141 on Build GRCh38
rs906724021 Chr.11: 977344-977344 on Build GRCh38
rs922448861 Chr.11: 977176-977176 on Build GRCh38
rs572231533 Chr.11: 977309-977309 on Build GRCh38
rs756084673 Chr.11: 977074-977075 on Build GRCh38
rs528447735 Chr.11: 977302-977302 on Build GRCh38
rs746665248 Chr.11: 977161-977161 on Build GRCh38
rs549459789 Chr.11: 977142-977142 on Build GRCh38
rs778892476 Chr.11: 977352-977352 on Build GRCh38
rs551684477 Chr.11: 977321-977321 on Build GRCh38
rs370958520 Chr.11: 977125-977125 on Build GRCh38
rs930304667 Chr.11: 977326-977326 on Build GRCh38
rs746225400 Chr.11: 977099-977099 on Build GRCh38
rs922367303 Chr.11: 977403-977403 on Build GRCh38
rs766117962 Chr.11: 977230-977230 on Build GRCh38
rs776075998 Chr.11: 977102-977102 on Build GRCh38
rs1001452627 Chr.11: 977383-977383 on Build GRCh38
rs886170794 Chr.11: 977335-977335 on Build GRCh38
rs910977400 Chr.11: 977148-977148 on Build GRCh38
rs780587992 Chr.11: 977167-977167 on Build GRCh38
rs377743784 Chr.11: 977110-977110 on Build GRCh38
rs766171655 Chr.11: 977130-977130 on Build GRCh38
rs759428299 Chr.11: 977243-977243 on Build GRCh38
rs762001403 Chr.11: 977124-977124 on Build GRCh38
rs114762092 Chr.11: 977386-977386 on Build GRCh38
rs760394119 Chr.11: 977128-977128 on Build GRCh38
rs913319108 Chr.11: 977217-977217 on Build GRCh38
rs760907083 Chr.11: 977211-977211 on Build GRCh38
rs769021235 Chr.11: 977172-977172 on Build GRCh38
rs529975838 Chr.11: 977400-977400 on Build GRCh38
rs745983771 Chr.11: 977049-977050 on Build GRCh38
rs757783352 Chr.11: 977274-977274 on Build GRCh38
rs963725999 Chr.11: 977399-977399 on Build GRCh38
rs911398357 Chr.11: 977441-977441 on Build GRCh38
rs747793140 Chr.11: 977076-977076 on Build GRCh38
rs958248293 Chr.11: 977364-977364 on Build GRCh38
rs761964683 Chr.11: 977054-977054 on Build GRCh38

View Full Product Details