Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 988360-988855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2A2
Entrez Gene ID
161
Gene Name
adaptor related protein complex 2 alpha 2 subunit
Gene Aliases
ADTAB, CLAPA2, HIP-9, HIP9, HYPJ
UniGene
Hs.19121
Gene Chromosome Location
Chr.11: 925809-1012245 on Build GRCh38
Transcripts
NM_001242837.1 NM_012305.3 XM_011519929.1 XM_011519930.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6070730 c.1159C>T p.R387W Chr.11: 988579-988579 on Build GRCh38
4037822 c.1193G>A p.R398H Chr.11: 988613-988613 on Build GRCh38
4462705 c.1254C>T p.S418S Chr.11: 988674-988674 on Build GRCh38
4523610 c.1219G>A p.E407K Chr.11: 988639-988639 on Build GRCh38
6849950 c.1253C>T p.S418F Chr.11: 988673-988673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908801367 Chr.11: 988420-988420 on Build GRCh38
rs368388767 Chr.11: 988599-988599 on Build GRCh38
rs961610848 Chr.11: 988431-988431 on Build GRCh38
rs763234910 Chr.11: 988594-988595 on Build GRCh38
rs918620269 Chr.11: 988756-988756 on Build GRCh38
rs536770387 Chr.11: 988471-988471 on Build GRCh38
rs747605211 Chr.11: 988826-988827 on Build GRCh38
rs770771246 Chr.11: 988605-988605 on Build GRCh38
rs777411161 Chr.11: 988540-988540 on Build GRCh38
rs749816068 Chr.11: 988570-988570 on Build GRCh38
rs1032760928 Chr.11: 988813-988813 on Build GRCh38
rs959591236 Chr.11: 988829-988830 on Build GRCh38
rs75750770 Chr.11: 988742-988742 on Build GRCh38
rs975489878 Chr.11: 988392-988393 on Build GRCh38
rs926942701 Chr.11: 988757-988757 on Build GRCh38
rs756614085 Chr.11: 988566-988566 on Build GRCh38
rs766218989 Chr.11: 988532-988532 on Build GRCh38
rs748276511 Chr.11: 988584-988584 on Build GRCh38
rs915678660 Chr.11: 988688-988688 on Build GRCh38
rs567722539 Chr.11: 988405-988405 on Build GRCh38
rs866531685 Chr.11: 988703-988703 on Build GRCh38
rs751262548 Chr.11: 988620-988620 on Build GRCh38
rs765130178 Chr.11: 988609-988609 on Build GRCh38
rs762614728 Chr.11: 988615-988615 on Build GRCh38
rs1015727759 Chr.11: 988613-988613 on Build GRCh38
rs780490962 Chr.11: 988621-988621 on Build GRCh38
rs924463138 Chr.11: 988727-988727 on Build GRCh38
rs774137653 Chr.11: 988729-988729 on Build GRCh38
rs376073560 Chr.11: 988580-988580 on Build GRCh38
rs374078818 Chr.11: 988632-988632 on Build GRCh38
rs938470698 Chr.11: 988452-988453 on Build GRCh38
rs747250419 Chr.11: 988682-988682 on Build GRCh38
rs1028535574 Chr.11: 988397-988397 on Build GRCh38
rs762196661 Chr.11: 988626-988626 on Build GRCh38
rs972990573 Chr.11: 988700-988700 on Build GRCh38
rs777620962 Chr.11: 988559-988559 on Build GRCh38
rs572932767 Chr.11: 988547-988547 on Build GRCh38
rs768577549 Chr.11: 988574-988574 on Build GRCh38
rs937087510 Chr.11: 988761-988761 on Build GRCh38
rs551317283 Chr.11: 988794-988794 on Build GRCh38
rs759104340 Chr.11: 988596-988596 on Build GRCh38
rs931559096 Chr.11: 988724-988724 on Build GRCh38
rs1005626062 Chr.11: 988783-988783 on Build GRCh38
rs545817355 Chr.11: 988771-988771 on Build GRCh38
rs1055449597 Chr.11: 988461-988461 on Build GRCh38
rs952838457 Chr.11: 988398-988398 on Build GRCh38
rs758908829 Chr.11: 988536-988536 on Build GRCh38
rs896860670 Chr.11: 988455-988455 on Build GRCh38
rs560274090 Chr.11: 988765-988765 on Build GRCh38
rs895683923 Chr.11: 988766-988766 on Build GRCh38
rs866811703 Chr.11: 988707-988707 on Build GRCh38
rs774379835 Chr.11: 988503-988503 on Build GRCh38
rs144441591 Chr.11: 988619-988619 on Build GRCh38
rs755436493 Chr.11: 988380-988380 on Build GRCh38
rs776381664 Chr.11: 988607-988607 on Build GRCh38
rs1000087804 Chr.11: 988799-988799 on Build GRCh38
rs371478838 Chr.11: 988723-988723 on Build GRCh38
rs954070625 Chr.11: 988816-988816 on Build GRCh38
rs371138229 Chr.11: 988518-988518 on Build GRCh38
rs11538724 Chr.11: 988657-988657 on Build GRCh38
rs538516270 Chr.11: 988770-988770 on Build GRCh38
rs764823391 Chr.11: 988543-988543 on Build GRCh38
rs748221608 Chr.11: 988639-988639 on Build GRCh38
rs753815361 Chr.11: 988534-988534 on Build GRCh38
rs972146905 Chr.11: 988434-988434 on Build GRCh38
rs778985845 Chr.11: 988404-988404 on Build GRCh38
rs750808260 Chr.11: 988563-988563 on Build GRCh38
rs996958618 Chr.11: 988540-988541 on Build GRCh38
rs759551186 Chr.11: 988608-988608 on Build GRCh38
rs1001264133 Chr.11: 988549-988549 on Build GRCh38
rs1042874291 Chr.11: 988479-988479 on Build GRCh38
rs947234457 Chr.11: 988478-988478 on Build GRCh38
rs965855885 Chr.11: 988384-988384 on Build GRCh38
rs1041679719 Chr.11: 988782-988782 on Build GRCh38
rs201537556 Chr.11: 988650-988650 on Build GRCh38
rs1035246452 Chr.11: 988827-988827 on Build GRCh38
rs773847194 Chr.11: 988569-988569 on Build GRCh38
rs928312337 Chr.11: 988437-988437 on Build GRCh38
rs62640021 Chr.11: 988638-988638 on Build GRCh38
rs773707661 Chr.11: 988817-988817 on Build GRCh38
rs904455683 Chr.11: 988798-988798 on Build GRCh38
rs139783880 Chr.11: 988772-988772 on Build GRCh38
rs529819420 Chr.11: 988777-988777 on Build GRCh38
rs770589109 Chr.11: 988715-988715 on Build GRCh38
rs760392178 Chr.11: 988603-988603 on Build GRCh38
rs553025156 Chr.11: 988510-988510 on Build GRCh38
rs752237271 Chr.11: 988612-988612 on Build GRCh38
rs867485594 Chr.11: 988583-988583 on Build GRCh38
rs966764407 Chr.11: 988600-988600 on Build GRCh38
rs758095675 Chr.11: 988557-988557 on Build GRCh38
rs762051977 Chr.11: 988508-988508 on Build GRCh38
rs574519019 Chr.11: 988741-988741 on Build GRCh38
rs760520847 Chr.11: 988528-988528 on Build GRCh38
rs776458440 Chr.11: 988717-988717 on Build GRCh38
rs998750713 Chr.11: 988523-988523 on Build GRCh38

View Full Product Details