Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3659841-3660377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART1
Entrez Gene ID
417
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
ART2, ARTC1, CD296, RT6
UniGene
Hs.382188
Gene Chromosome Location
Chr.11: 3645110-3665595 on Build GRCh38
Transcripts
NM_004314.2 XM_011520114.2 XM_017017763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6305007 c.784G>T p.A262S Chr.11: 3660303-3660303 on Build GRCh38
4663567 c.735_736delCT p.F246fs*1 Chr.11: 3660254-3660255 on Build GRCh38
4748938 c.389A>G p.E130G Chr.11: 3659908-3659908 on Build GRCh38
4970268 c.575G>A p.R192Q Chr.11: 3660094-3660094 on Build GRCh38
3447112 c.696C>T p.Y232Y Chr.11: 3660215-3660215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767771808 Chr.11: 3659982-3659982 on Build GRCh38
rs141734720 Chr.11: 3660170-3660170 on Build GRCh38
rs111942933 Chr.11: 3659939-3659939 on Build GRCh38
rs1044310817 Chr.11: 3660079-3660094 on Build GRCh38
rs141732093 Chr.11: 3660028-3660028 on Build GRCh38
rs762197868 Chr.11: 3660037-3660037 on Build GRCh38
rs762428866 Chr.11: 3660248-3660248 on Build GRCh38
rs750686385 Chr.11: 3659987-3659987 on Build GRCh38
rs148703944 Chr.11: 3659926-3659926 on Build GRCh38
rs774844448 Chr.11: 3660085-3660085 on Build GRCh38
rs900074617 Chr.11: 3660161-3660161 on Build GRCh38
rs781422161 Chr.11: 3659922-3659922 on Build GRCh38
rs577558078 Chr.11: 3659968-3659968 on Build GRCh38
rs371263931 Chr.11: 3660066-3660066 on Build GRCh38
rs202219245 Chr.11: 3660087-3660087 on Build GRCh38
rs542871399 Chr.11: 3660073-3660073 on Build GRCh38
rs554173168 Chr.11: 3659995-3659995 on Build GRCh38
rs761598555 Chr.11: 3659933-3659933 on Build GRCh38
rs943927322 Chr.11: 3659862-3659862 on Build GRCh38
rs750734969 Chr.11: 3660043-3660043 on Build GRCh38
rs757189893 Chr.11: 3660121-3660121 on Build GRCh38
rs2280134 Chr.11: 3660289-3660289 on Build GRCh38
rs996977250 Chr.11: 3659895-3659895 on Build GRCh38
rs145405723 Chr.11: 3660054-3660054 on Build GRCh38
rs748021891 Chr.11: 3660010-3660010 on Build GRCh38
rs927702624 Chr.11: 3660091-3660091 on Build GRCh38
rs142006583 Chr.11: 3660093-3660093 on Build GRCh38
rs773884895 Chr.11: 3660138-3660138 on Build GRCh38
rs201158047 Chr.11: 3660322-3660322 on Build GRCh38
rs769388795 Chr.11: 3660082-3660082 on Build GRCh38
rs771377517 Chr.11: 3660201-3660201 on Build GRCh38
rs760968477 Chr.11: 3660094-3660094 on Build GRCh38
rs143216187 Chr.11: 3660069-3660069 on Build GRCh38
rs764347824 Chr.11: 3660230-3660230 on Build GRCh38
rs751912272 Chr.11: 3659914-3659914 on Build GRCh38
rs775467427 Chr.11: 3659865-3659865 on Build GRCh38
rs766036791 Chr.11: 3659944-3659944 on Build GRCh38
rs745586244 Chr.11: 3660124-3660124 on Build GRCh38
rs201487637 Chr.11: 3660000-3660000 on Build GRCh38
rs748952979 Chr.11: 3660198-3660198 on Build GRCh38
rs138825846 Chr.11: 3660079-3660079 on Build GRCh38
rs754155931 Chr.11: 3660097-3660097 on Build GRCh38
rs267602868 Chr.11: 3660249-3660249 on Build GRCh38
rs892156505 Chr.11: 3660115-3660115 on Build GRCh38
rs974096183 Chr.11: 3660248-3660248 on Build GRCh38
rs763183394 Chr.11: 3660162-3660162 on Build GRCh38
rs566663445 Chr.11: 3660199-3660199 on Build GRCh38
rs750142725 Chr.11: 3660177-3660177 on Build GRCh38
rs777306904 Chr.11: 3660329-3660329 on Build GRCh38
rs776376314 Chr.11: 3660116-3660117 on Build GRCh38
rs766135070 Chr.11: 3660306-3660306 on Build GRCh38
rs760397227 Chr.11: 3659943-3659943 on Build GRCh38
rs759029297 Chr.11: 3660029-3660029 on Build GRCh38
rs774042346 Chr.11: 3659932-3659932 on Build GRCh38
rs759245792 Chr.11: 3660119-3660120 on Build GRCh38
rs35619488 Chr.11: 3659896-3659896 on Build GRCh38
rs199699575 Chr.11: 3660023-3660023 on Build GRCh38
rs35136756 Chr.11: 3659885-3659885 on Build GRCh38
rs374138196 Chr.11: 3660072-3660072 on Build GRCh38
rs759834685 Chr.11: 3660155-3660155 on Build GRCh38
rs368030263 Chr.11: 3660130-3660130 on Build GRCh38
rs757914337 Chr.11: 3660323-3660323 on Build GRCh38
rs770721141 Chr.11: 3660076-3660076 on Build GRCh38
rs764813601 Chr.11: 3660318-3660318 on Build GRCh38
rs780600255 Chr.11: 3660347-3660347 on Build GRCh38
rs747616456 Chr.11: 3660271-3660271 on Build GRCh38
rs769221472 Chr.11: 3660006-3660006 on Build GRCh38
rs748677669 Chr.11: 3660132-3660132 on Build GRCh38
rs753925166 Chr.11: 3660056-3660056 on Build GRCh38
rs772551570 Chr.11: 3660030-3660030 on Build GRCh38
rs763239595 Chr.11: 3660239-3660239 on Build GRCh38
rs200211996 Chr.11: 3659949-3659949 on Build GRCh38
rs767921923 Chr.11: 3660038-3660038 on Build GRCh38
rs750943398 Chr.11: 3660334-3660334 on Build GRCh38
rs369235320 Chr.11: 3660350-3660350 on Build GRCh38
rs756598197 Chr.11: 3660340-3660340 on Build GRCh38
rs754656464 Chr.11: 3660315-3660315 on Build GRCh38
rs149229160 Chr.11: 3660065-3660065 on Build GRCh38
rs543916143 Chr.11: 3660088-3660088 on Build GRCh38
rs751530347 Chr.11: 3660120-3660120 on Build GRCh38
rs774993469 Chr.11: 3660036-3660036 on Build GRCh38
rs778023397 Chr.11: 3659898-3659898 on Build GRCh38
rs371356884 Chr.11: 3659925-3659925 on Build GRCh38
rs767154046 Chr.11: 3659884-3659884 on Build GRCh38
rs758416985 Chr.11: 3660111-3660111 on Build GRCh38
rs768964355 Chr.11: 3660291-3660291 on Build GRCh38
rs991136254 Chr.11: 3659972-3659972 on Build GRCh38
rs774170580 Chr.11: 3659882-3659882 on Build GRCh38
rs202112440 Chr.11: 3660122-3660122 on Build GRCh38
rs781161695 Chr.11: 3659860-3659860 on Build GRCh38
rs139601876 Chr.11: 3660260-3660260 on Build GRCh38
rs752835498 Chr.11: 3660159-3660159 on Build GRCh38
rs769373079 Chr.11: 3660181-3660181 on Build GRCh38
rs150574054 Chr.11: 3660253-3660253 on Build GRCh38
rs888495221 Chr.11: 3659919-3659919 on Build GRCh38
rs779510660 Chr.11: 3660183-3660183 on Build GRCh38
rs752333152 Chr.11: 3660319-3660319 on Build GRCh38
rs765406319 Chr.11: 3660102-3660102 on Build GRCh38
rs760627847 Chr.11: 3660305-3660305 on Build GRCh38
rs776836109 Chr.11: 3660152-3660152 on Build GRCh38
rs141183632 Chr.11: 3659991-3659991 on Build GRCh38
rs761164939 Chr.11: 3660145-3660145 on Build GRCh38
rs139469651 Chr.11: 3660302-3660302 on Build GRCh38
rs757796309 Chr.11: 3660272-3660272 on Build GRCh38
rs774573466 Chr.11: 3660294-3660294 on Build GRCh38
rs754603309 Chr.11: 3660266-3660266 on Build GRCh38
rs771324525 Chr.11: 3660150-3660150 on Build GRCh38
rs761390636 Chr.11: 3659883-3659883 on Build GRCh38
rs145152353 Chr.11: 3660015-3660015 on Build GRCh38
rs772525381 Chr.11: 3660089-3660089 on Build GRCh38
rs777059550 Chr.11: 3660210-3660210 on Build GRCh38
rs757425295 Chr.11: 3659921-3659921 on Build GRCh38
rs768011147 Chr.11: 3660315-3660316 on Build GRCh38
rs376555468 Chr.11: 3659861-3659861 on Build GRCh38
rs746683411 Chr.11: 3660021-3660021 on Build GRCh38
rs753529163 Chr.11: 3660263-3660263 on Build GRCh38
rs754871942 Chr.11: 3660005-3660005 on Build GRCh38
rs777796511 Chr.11: 3660117-3660117 on Build GRCh38
rs537517512 Chr.11: 3659937-3659937 on Build GRCh38
rs767619916 Chr.11: 3660303-3660303 on Build GRCh38
rs77748211 Chr.11: 3659899-3659899 on Build GRCh38
rs550030209 Chr.11: 3659893-3659893 on Build GRCh38
rs147920431 Chr.11: 3660020-3660020 on Build GRCh38
rs746383575 Chr.11: 3660277-3660277 on Build GRCh38
rs769009481 Chr.11: 3660351-3660351 on Build GRCh38
rs963054140 Chr.11: 3660217-3660217 on Build GRCh38
rs752681438 Chr.11: 3660110-3660110 on Build GRCh38
rs1051217304 Chr.11: 3660337-3660337 on Build GRCh38
rs186835333 Chr.11: 3660176-3660176 on Build GRCh38
rs778415511 Chr.11: 3660270-3660270 on Build GRCh38
rs773087367 Chr.11: 3660304-3660304 on Build GRCh38
rs781609382 Chr.11: 3660276-3660276 on Build GRCh38
rs775062993 Chr.11: 3660247-3660248 on Build GRCh38
rs151146245 Chr.11: 3659984-3659984 on Build GRCh38
rs749144608 Chr.11: 3660055-3660055 on Build GRCh38
rs779643913 Chr.11: 3660131-3660131 on Build GRCh38
rs770182602 Chr.11: 3660237-3660237 on Build GRCh38
rs545730767 Chr.11: 3659974-3659974 on Build GRCh38
rs769740196 Chr.11: 3659862-3659862 on Build GRCh38
rs746738511 Chr.11: 3659861-3659862 on Build GRCh38
rs748155690 Chr.11: 3660068-3660068 on Build GRCh38
rs200800426 Chr.11: 3660252-3660252 on Build GRCh38
rs139321904 Chr.11: 3659867-3659867 on Build GRCh38
rs138349653 Chr.11: 3660148-3660148 on Build GRCh38
rs755915909 Chr.11: 3660254-3660254 on Build GRCh38
rs768373376 Chr.11: 3659878-3659878 on Build GRCh38
rs376054375 Chr.11: 3659963-3659963 on Build GRCh38
rs760204892 Chr.11: 3659892-3659892 on Build GRCh38
rs377610501 Chr.11: 3660307-3660307 on Build GRCh38
rs757702310 Chr.11: 3659975-3659975 on Build GRCh38
rs755793863 Chr.11: 3660178-3660178 on Build GRCh38
rs759645257 Chr.11: 3660101-3660101 on Build GRCh38
rs770886227 Chr.11: 3659909-3659909 on Build GRCh38
rs1010554048 Chr.11: 3660127-3660127 on Build GRCh38
rs145683174 Chr.11: 3660137-3660137 on Build GRCh38
rs199558962 Chr.11: 3660168-3660168 on Build GRCh38
rs113817011 Chr.11: 3660090-3660090 on Build GRCh38
rs753322141 Chr.11: 3659894-3659894 on Build GRCh38
rs201450193 Chr.11: 3660227-3660227 on Build GRCh38
rs761461664 Chr.11: 3660203-3660203 on Build GRCh38
rs561441207 Chr.11: 3660078-3660078 on Build GRCh38
rs765311915 Chr.11: 3660158-3660158 on Build GRCh38
rs144426322 Chr.11: 3660290-3660290 on Build GRCh38
rs150462911 Chr.11: 3660033-3660033 on Build GRCh38

View Full Product Details