Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3660265-3660767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART1
Entrez Gene ID
417
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
ART2, ARTC1, CD296, RT6
UniGene
Hs.382188
Gene Chromosome Location
Chr.11: 3645110-3665595 on Build GRCh38
Transcripts
NM_004314.2 XM_011520114.2 XM_017017763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6305007 c.784G>T p.A262S Chr.11: 3660303-3660303 on Build GRCh38
4396461 c.800G>A p.R267Q Chr.11: 3660319-3660319 on Build GRCh38
5041395 c.769C>G p.L257V Chr.11: 3660288-3660288 on Build GRCh38
5910719 c.772G>A p.A258T Chr.11: 3660291-3660291 on Build GRCh38
5050795 c.788G>A p.R263H Chr.11: 3660307-3660307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182787832 Chr.11: 3660641-3660641 on Build GRCh38
rs940011198 Chr.11: 3660517-3660517 on Build GRCh38
rs1051309278 Chr.11: 3660575-3660575 on Build GRCh38
rs1034917874 Chr.11: 3660475-3660475 on Build GRCh38
rs10834744 Chr.11: 3660620-3660620 on Build GRCh38
rs773317551 Chr.11: 3660370-3660370 on Build GRCh38
rs759384567 Chr.11: 3660378-3660378 on Build GRCh38
rs1055728860 Chr.11: 3660539-3660539 on Build GRCh38
rs752333152 Chr.11: 3660319-3660319 on Build GRCh38
rs987984587 Chr.11: 3660495-3660495 on Build GRCh38
rs760627847 Chr.11: 3660305-3660305 on Build GRCh38
rs752299466 Chr.11: 3660576-3660576 on Build GRCh38
rs139469651 Chr.11: 3660302-3660302 on Build GRCh38
rs747465325 Chr.11: 3660656-3660656 on Build GRCh38
rs774573466 Chr.11: 3660294-3660294 on Build GRCh38
rs780515733 Chr.11: 3660651-3660651 on Build GRCh38
rs946447735 Chr.11: 3660524-3660524 on Build GRCh38
rs754379630 Chr.11: 3660405-3660405 on Build GRCh38
rs373559116 Chr.11: 3660622-3660622 on Build GRCh38
rs2280134 Chr.11: 3660289-3660289 on Build GRCh38
rs768011147 Chr.11: 3660315-3660316 on Build GRCh38
rs201158047 Chr.11: 3660322-3660322 on Build GRCh38
rs767619916 Chr.11: 3660303-3660303 on Build GRCh38
rs932306023 Chr.11: 3660545-3660545 on Build GRCh38
rs12786225 Chr.11: 3660400-3660400 on Build GRCh38
rs968215427 Chr.11: 3660700-3660700 on Build GRCh38
rs751452807 Chr.11: 3660520-3660520 on Build GRCh38
rs377476664 Chr.11: 3660371-3660371 on Build GRCh38
rs772081296 Chr.11: 3660365-3660365 on Build GRCh38
rs769009481 Chr.11: 3660351-3660351 on Build GRCh38
rs1051217304 Chr.11: 3660337-3660337 on Build GRCh38
rs531406237 Chr.11: 3660704-3660704 on Build GRCh38
rs1020762333 Chr.11: 3660699-3660699 on Build GRCh38
rs773087367 Chr.11: 3660304-3660304 on Build GRCh38
rs940215054 Chr.11: 3660589-3660589 on Build GRCh38
rs953896047 Chr.11: 3660653-3660653 on Build GRCh38
rs548857801 Chr.11: 3660643-3660643 on Build GRCh38
rs752388349 Chr.11: 3660386-3660386 on Build GRCh38
rs200477622 Chr.11: 3660357-3660357 on Build GRCh38
rs751044850 Chr.11: 3660395-3660395 on Build GRCh38
rs953751000 Chr.11: 3660722-3660722 on Build GRCh38
rs754845833 Chr.11: 3660534-3660534 on Build GRCh38
rs777306904 Chr.11: 3660329-3660329 on Build GRCh38
rs1016863607 Chr.11: 3660660-3660660 on Build GRCh38
rs1028145399 Chr.11: 3660425-3660425 on Build GRCh38
rs766135070 Chr.11: 3660306-3660306 on Build GRCh38
rs960569456 Chr.11: 3660485-3660485 on Build GRCh38
rs565235061 Chr.11: 3660595-3660595 on Build GRCh38
rs750933517 Chr.11: 3660395-3660396 on Build GRCh38
rs1016131400 Chr.11: 3660384-3660384 on Build GRCh38
rs1031308350 Chr.11: 3660618-3660618 on Build GRCh38
rs560797269 Chr.11: 3660659-3660659 on Build GRCh38
rs371110308 Chr.11: 3660394-3660394 on Build GRCh38
rs757914337 Chr.11: 3660323-3660323 on Build GRCh38
rs576698850 Chr.11: 3660493-3660493 on Build GRCh38
rs377610501 Chr.11: 3660307-3660307 on Build GRCh38
rs764813601 Chr.11: 3660318-3660318 on Build GRCh38
rs762648456 Chr.11: 3660393-3660393 on Build GRCh38
rs780600255 Chr.11: 3660347-3660347 on Build GRCh38
rs78604524 Chr.11: 3660396-3660396 on Build GRCh38
rs779387734 Chr.11: 3660406-3660406 on Build GRCh38
rs980714216 Chr.11: 3660442-3660442 on Build GRCh38
rs780646905 Chr.11: 3660401-3660401 on Build GRCh38
rs199730802 Chr.11: 3660377-3660377 on Build GRCh38
rs577226950 Chr.11: 3660612-3660612 on Build GRCh38
rs750943398 Chr.11: 3660334-3660334 on Build GRCh38
rs979233802 Chr.11: 3660703-3660703 on Build GRCh38
rs369235320 Chr.11: 3660350-3660350 on Build GRCh38
rs754656464 Chr.11: 3660315-3660315 on Build GRCh38
rs756598197 Chr.11: 3660340-3660340 on Build GRCh38
rs969308551 Chr.11: 3660437-3660437 on Build GRCh38
rs370145785 Chr.11: 3660376-3660376 on Build GRCh38
rs373715082 Chr.11: 3660364-3660364 on Build GRCh38
rs2280135 Chr.11: 3660385-3660385 on Build GRCh38
rs984035680 Chr.11: 3660465-3660465 on Build GRCh38
rs144426322 Chr.11: 3660290-3660290 on Build GRCh38
rs61878557 Chr.11: 3660535-3660535 on Build GRCh38
rs1034970089 Chr.11: 3660648-3660648 on Build GRCh38
rs1023045063 Chr.11: 3660623-3660623 on Build GRCh38
rs939421027 Chr.11: 3660538-3660538 on Build GRCh38
rs768964355 Chr.11: 3660291-3660291 on Build GRCh38

View Full Product Details