Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32601592-32602100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC73
Entrez Gene ID
493860
Gene Name
coiled-coil domain containing 73
Gene Aliases
NY-SAR-79
UniGene
Hs.706808
Gene Chromosome Location
Chr.11: 32602080-32794658 on Build GRCh38
Transcripts
NM_001008391.3 NM_001307929.1 NM_006360.5 XM_011519840.2

Gene Symbol
EIF3M
Entrez Gene ID
10480
Gene Name
eukaryotic translation initiation factor 3 subunit M
Gene Aliases
B5, GA17, PCID1, TANGO7, hfl-B5
UniGene
Hs.502244
Gene Chromosome Location
Chr.11: 32583767-32602873 on Build GRCh38
Transcripts
NM_001008391.3 NM_001307929.1 NM_006360.5 XM_011519840.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866956 c.944C>T p.A315V Chr.11: 32601762-32601762 on Build GRCh38
7304902 c.946G>A p.V316I Chr.11: 32601764-32601764 on Build GRCh38
6394843 c.969C>A p.C323* Chr.11: 32601787-32601787 on Build GRCh38
6258343 c.944-1G>T p.? Chr.11: 32601761-32601761 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373325255 Chr.11: 32601857-32601857 on Build GRCh38
rs975670163 Chr.11: 32601813-32601813 on Build GRCh38
rs115065212 Chr.11: 32602013-32602013 on Build GRCh38
rs369869761 Chr.11: 32601752-32601752 on Build GRCh38
rs569072491 Chr.11: 32601915-32601915 on Build GRCh38
rs926060332 Chr.11: 32601630-32601630 on Build GRCh38
rs988849454 Chr.11: 32601703-32601703 on Build GRCh38
rs755678406 Chr.11: 32601763-32601763 on Build GRCh38
rs377514127 Chr.11: 32601858-32601858 on Build GRCh38
rs936070058 Chr.11: 32601701-32601701 on Build GRCh38
rs994304133 Chr.11: 32601792-32601792 on Build GRCh38
rs77129113 Chr.11: 32601755-32601755 on Build GRCh38
rs898274730 Chr.11: 32601783-32601783 on Build GRCh38
rs749823142 Chr.11: 32601802-32601802 on Build GRCh38
rs892686770 Chr.11: 32601943-32601943 on Build GRCh38
rs760786011 Chr.11: 32601731-32601731 on Build GRCh38
rs754612117 Chr.11: 32601753-32601753 on Build GRCh38
rs763020018 Chr.11: 32601822-32601823 on Build GRCh38
rs1802364 Chr.11: 32601784-32601784 on Build GRCh38
rs1045545534 Chr.11: 32601911-32601911 on Build GRCh38
rs1038495355 Chr.11: 32601774-32601774 on Build GRCh38
rs370912230 Chr.11: 32601616-32601617 on Build GRCh38
rs762912374 Chr.11: 32601864-32601864 on Build GRCh38
rs764174112 Chr.11: 32601859-32601859 on Build GRCh38
rs1003830391 Chr.11: 32601626-32601631 on Build GRCh38
rs1056717739 Chr.11: 32601929-32601929 on Build GRCh38
rs1035676188 Chr.11: 32601681-32601681 on Build GRCh38
rs879495926 Chr.11: 32601894-32601894 on Build GRCh38
rs989312909 Chr.11: 32601979-32601979 on Build GRCh38
rs201586926 Chr.11: 32601739-32601739 on Build GRCh38
rs937582315 Chr.11: 32602053-32602053 on Build GRCh38
rs1021944592 Chr.11: 32601993-32601993 on Build GRCh38
rs531410535 Chr.11: 32601927-32601927 on Build GRCh38
rs758276803 Chr.11: 32601740-32601740 on Build GRCh38
rs950054543 Chr.11: 32601723-32601723 on Build GRCh38
rs956541285 Chr.11: 32601936-32601936 on Build GRCh38
rs771497467 Chr.11: 32601807-32601807 on Build GRCh38
rs778478271 Chr.11: 32601764-32601764 on Build GRCh38
rs74673753 Chr.11: 32602075-32602075 on Build GRCh38
rs766293592 Chr.11: 32601871-32601871 on Build GRCh38
rs776626176 Chr.11: 32601861-32601861 on Build GRCh38
rs774607688 Chr.11: 32601823-32601823 on Build GRCh38
rs913283991 Chr.11: 32601986-32601986 on Build GRCh38
rs913239549 Chr.11: 32601708-32601708 on Build GRCh38
rs981406687 Chr.11: 32602005-32602005 on Build GRCh38
rs1028482777 Chr.11: 32602073-32602073 on Build GRCh38
rs772230629 Chr.11: 32601826-32601826 on Build GRCh38
rs141937118 Chr.11: 32601737-32601737 on Build GRCh38
rs779830531 Chr.11: 32601743-32601743 on Build GRCh38
rs754774721 Chr.11: 32601874-32601874 on Build GRCh38
rs373077771 Chr.11: 32601729-32601729 on Build GRCh38
rs971809687 Chr.11: 32601998-32601998 on Build GRCh38
rs370151072 Chr.11: 32601825-32601825 on Build GRCh38
rs1018243043 Chr.11: 32601893-32601893 on Build GRCh38
rs775847565 Chr.11: 32601753-32601755 on Build GRCh38
rs77379922 Chr.11: 32601873-32601873 on Build GRCh38
rs963948043 Chr.11: 32601914-32601914 on Build GRCh38
rs1046786999 Chr.11: 32601854-32601854 on Build GRCh38
rs1009247826 Chr.11: 32601970-32601970 on Build GRCh38
rs370165322 Chr.11: 32601738-32601738 on Build GRCh38
rs930769960 Chr.11: 32601895-32601895 on Build GRCh38
rs920699761 Chr.11: 32601895-32601896 on Build GRCh38

View Full Product Details