Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32613231-32613746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC73
Entrez Gene ID
493860
Gene Name
coiled-coil domain containing 73
Gene Aliases
NY-SAR-79
UniGene
Hs.706808
Gene Chromosome Location
Chr.11: 32602080-32794658 on Build GRCh38
Transcripts
NM_001008391.3 XM_011520139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3725908 c.2725C>G p.P909A Chr.11: 32613593-32613593 on Build GRCh38
687511 c.2574C>T p.F858F Chr.11: 32613744-32613744 on Build GRCh38
6068558 c.2674G>T p.D892Y Chr.11: 32613644-32613644 on Build GRCh38
5416255 c.2896+10C>T p.? Chr.11: 32613412-32613412 on Build GRCh38
4032396 c.2802G>C p.E934D Chr.11: 32613516-32613516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543831421 Chr.11: 32613277-32613277 on Build GRCh38
rs554076892 Chr.11: 32613706-32613706 on Build GRCh38
rs374249888 Chr.11: 32613687-32613687 on Build GRCh38
rs759632812 Chr.11: 32613660-32613660 on Build GRCh38
rs771471823 Chr.11: 32613476-32613476 on Build GRCh38
rs373559418 Chr.11: 32613394-32613394 on Build GRCh38
rs188950262 Chr.11: 32613550-32613550 on Build GRCh38
rs769082521 Chr.11: 32613596-32613596 on Build GRCh38
rs777747155 Chr.11: 32613551-32613551 on Build GRCh38
rs779725038 Chr.11: 32613698-32613698 on Build GRCh38
rs756017348 Chr.11: 32613412-32613412 on Build GRCh38
rs763207450 Chr.11: 32613546-32613546 on Build GRCh38
rs1016995662 Chr.11: 32613615-32613615 on Build GRCh38
rs757130265 Chr.11: 32613675-32613675 on Build GRCh38
rs750169936 Chr.11: 32613694-32613694 on Build GRCh38
rs757044342 Chr.11: 32613555-32613555 on Build GRCh38
rs750657243 Chr.11: 32613537-32613537 on Build GRCh38
rs764007313 Chr.11: 32613665-32613665 on Build GRCh38
rs796835219 Chr.11: 32613650-32613650 on Build GRCh38
rs190344047 Chr.11: 32613326-32613326 on Build GRCh38
rs559813944 Chr.11: 32613371-32613371 on Build GRCh38
rs949402936 Chr.11: 32613520-32613520 on Build GRCh38
rs201070131 Chr.11: 32613674-32613674 on Build GRCh38
rs746566241 Chr.11: 32613701-32613701 on Build GRCh38
rs758034879 Chr.11: 32613697-32613697 on Build GRCh38
rs545937623 Chr.11: 32613359-32613359 on Build GRCh38
rs574814157 Chr.11: 32613543-32613543 on Build GRCh38
rs954369794 Chr.11: 32613259-32613259 on Build GRCh38
rs776908901 Chr.11: 32613506-32613506 on Build GRCh38
rs183956425 Chr.11: 32613533-32613533 on Build GRCh38
rs117521878 Chr.11: 32613442-32613442 on Build GRCh38
rs777456429 Chr.11: 32613416-32613416 on Build GRCh38
rs367851392 Chr.11: 32613556-32613556 on Build GRCh38
rs745534217 Chr.11: 32613571-32613571 on Build GRCh38
rs766579514 Chr.11: 32613627-32613627 on Build GRCh38
rs563721359 Chr.11: 32613283-32613283 on Build GRCh38
rs370560446 Chr.11: 32613613-32613613 on Build GRCh38
rs552007629 Chr.11: 32613323-32613323 on Build GRCh38
rs192445382 Chr.11: 32613572-32613572 on Build GRCh38
rs867448483 Chr.11: 32613464-32613464 on Build GRCh38
rs781251660 Chr.11: 32613381-32613381 on Build GRCh38
rs140972550 Chr.11: 32613385-32613385 on Build GRCh38
rs201737265 Chr.11: 32613547-32613547 on Build GRCh38
rs562028928 Chr.11: 32613453-32613453 on Build GRCh38
rs947246069 Chr.11: 32613310-32613310 on Build GRCh38
rs759413955 Chr.11: 32613460-32613460 on Build GRCh38
rs371209448 Chr.11: 32613444-32613444 on Build GRCh38
rs779453153 Chr.11: 32613575-32613575 on Build GRCh38
rs777271417 Chr.11: 32613597-32613597 on Build GRCh38
rs910466508 Chr.11: 32613293-32613293 on Build GRCh38
rs916137207 Chr.11: 32613262-32613262 on Build GRCh38
rs955862951 Chr.11: 32613712-32613712 on Build GRCh38
rs776266105 Chr.11: 32613408-32613411 on Build GRCh38
rs796463909 Chr.11: 32613565-32613565 on Build GRCh38
rs780788128 Chr.11: 32613707-32613707 on Build GRCh38
rs752451288 Chr.11: 32613647-32613647 on Build GRCh38
rs774347046 Chr.11: 32613644-32613644 on Build GRCh38
rs770293555 Chr.11: 32613610-32613610 on Build GRCh38
rs770621459 Chr.11: 32613443-32613443 on Build GRCh38
rs764792181 Chr.11: 32613635-32613635 on Build GRCh38
rs531147786 Chr.11: 32613492-32613492 on Build GRCh38
rs978624547 Chr.11: 32613680-32613680 on Build GRCh38
rs753675981 Chr.11: 32613554-32613554 on Build GRCh38
rs769157609 Chr.11: 32613505-32613505 on Build GRCh38
rs778507778 Chr.11: 32613452-32613452 on Build GRCh38
rs149853604 Chr.11: 32613532-32613532 on Build GRCh38
rs376242210 Chr.11: 32613454-32613454 on Build GRCh38
rs775007614 Chr.11: 32613483-32613483 on Build GRCh38
rs773641187 Chr.11: 32613612-32613612 on Build GRCh38
rs762323220 Chr.11: 32613600-32613600 on Build GRCh38
rs746504322 Chr.11: 32613585-32613585 on Build GRCh38
rs924259135 Chr.11: 32613350-32613350 on Build GRCh38

View Full Product Details