Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34888388-34888885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APIP
Entrez Gene ID
51074
Gene Name
APAF1 interacting protein
Gene Aliases
APIP2, CGI-29, CGI29, MMRP19, hAPIP
UniGene
Hs.447794
Gene Chromosome Location
Chr.11: 34882295-34916411 on Build GRCh38
Transcripts
NM_015957.3 XM_011520154.2 XM_017017875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4432012 c.270G>C p.M90I Chr.11: 34888858-34888858 on Build GRCh38
5650435 c.267G>A p.S89S Chr.11: 34888810-34888810 on Build GRCh38
7266858 c.276_279delTTGT p.C93fs*2 Chr.11: 34888849-34888852 on Build GRCh38
1183214 c.311C>A p.A104D Chr.11: 34888766-34888766 on Build GRCh38
7266857 c.225_228delTTGT p.C76fs*2 Chr.11: 34888849-34888852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1977420 Chr.11: 34888850-34888850 on Build GRCh38
rs959566327 Chr.11: 34888580-34888580 on Build GRCh38
rs535567205 Chr.11: 34888573-34888573 on Build GRCh38
rs764007986 Chr.11: 34888459-34888459 on Build GRCh38
rs749849808 Chr.11: 34888724-34888724 on Build GRCh38
rs1571134 Chr.11: 34888530-34888530 on Build GRCh38
rs58295829 Chr.11: 34888602-34888602 on Build GRCh38
rs767095224 Chr.11: 34888451-34888451 on Build GRCh38
rs147583783 Chr.11: 34888524-34888524 on Build GRCh38
rs778796827 Chr.11: 34888849-34888852 on Build GRCh38
rs184899673 Chr.11: 34888762-34888762 on Build GRCh38
rs374612734 Chr.11: 34888474-34888474 on Build GRCh38
rs1007367461 Chr.11: 34888603-34888603 on Build GRCh38
rs1040323160 Chr.11: 34888507-34888507 on Build GRCh38
rs753755687 Chr.11: 34888769-34888769 on Build GRCh38
rs780231727 Chr.11: 34888477-34888477 on Build GRCh38
rs959578163 Chr.11: 34888432-34888432 on Build GRCh38
rs993081799 Chr.11: 34888489-34888489 on Build GRCh38
rs1025249155 Chr.11: 34888610-34888610 on Build GRCh38
rs781571523 Chr.11: 34888416-34888416 on Build GRCh38
rs546686089 Chr.11: 34888793-34888794 on Build GRCh38
rs371778492 Chr.11: 34888460-34888460 on Build GRCh38
rs375428893 Chr.11: 34888729-34888729 on Build GRCh38
rs771551654 Chr.11: 34888843-34888843 on Build GRCh38
rs765119740 Chr.11: 34888780-34888780 on Build GRCh38
rs371800567 Chr.11: 34888738-34888738 on Build GRCh38
rs762537723 Chr.11: 34888430-34888430 on Build GRCh38
rs940116271 Chr.11: 34888784-34888784 on Build GRCh38
rs140661576 Chr.11: 34888816-34888816 on Build GRCh38
rs575264512 Chr.11: 34888585-34888585 on Build GRCh38
rs770321292 Chr.11: 34888731-34888731 on Build GRCh38
rs916062953 Chr.11: 34888523-34888523 on Build GRCh38
rs753034544 Chr.11: 34888811-34888811 on Build GRCh38
rs773858985 Chr.11: 34888733-34888733 on Build GRCh38
rs577071512 Chr.11: 34888411-34888411 on Build GRCh38
rs781461162 Chr.11: 34888810-34888810 on Build GRCh38
rs757738924 Chr.11: 34888830-34888830 on Build GRCh38
rs1571135 Chr.11: 34888670-34888670 on Build GRCh38
rs200854934 Chr.11: 34888704-34888704 on Build GRCh38
rs985579920 Chr.11: 34888688-34888688 on Build GRCh38
rs958067608 Chr.11: 34888681-34888681 on Build GRCh38
rs992408482 Chr.11: 34888440-34888440 on Build GRCh38
rs201312580 Chr.11: 34888475-34888475 on Build GRCh38
rs370412975 Chr.11: 34888450-34888450 on Build GRCh38
rs943262395 Chr.11: 34888504-34888509 on Build GRCh38
rs142737858 Chr.11: 34888422-34888422 on Build GRCh38
rs751613962 Chr.11: 34888479-34888479 on Build GRCh38
rs11032915 Chr.11: 34888740-34888740 on Build GRCh38
rs925366647 Chr.11: 34888482-34888482 on Build GRCh38
rs755188467 Chr.11: 34888703-34888703 on Build GRCh38
rs151010286 Chr.11: 34888778-34888778 on Build GRCh38
rs768283418 Chr.11: 34888794-34888794 on Build GRCh38
rs536434649 Chr.11: 34888831-34888831 on Build GRCh38
rs749688218 Chr.11: 34888827-34888827 on Build GRCh38
rs202180413 Chr.11: 34888454-34888454 on Build GRCh38
rs369991137 Chr.11: 34888748-34888748 on Build GRCh38
rs374995167 Chr.11: 34888436-34888436 on Build GRCh38
rs1037025847 Chr.11: 34888553-34888553 on Build GRCh38
rs564665836 Chr.11: 34888501-34888501 on Build GRCh38
rs773841994 Chr.11: 34888439-34888439 on Build GRCh38
rs149670209 Chr.11: 34888835-34888835 on Build GRCh38
rs968465628 Chr.11: 34888657-34888657 on Build GRCh38
rs551754743 Chr.11: 34888487-34888487 on Build GRCh38
rs748465761 Chr.11: 34888706-34888706 on Build GRCh38
rs199682939 Chr.11: 34888817-34888817 on Build GRCh38
rs1031297099 Chr.11: 34888678-34888678 on Build GRCh38
rs57876015 Chr.11: 34888561-34888561 on Build GRCh38
rs940107509 Chr.11: 34888551-34888551 on Build GRCh38
rs1054254076 Chr.11: 34888499-34888499 on Build GRCh38
rs372208635 Chr.11: 34888765-34888765 on Build GRCh38
rs368397377 Chr.11: 34888711-34888711 on Build GRCh38
rs865796358 Chr.11: 34888860-34888860 on Build GRCh38
rs1030280568 Chr.11: 34888589-34888589 on Build GRCh38
rs1049052372 Chr.11: 34888526-34888526 on Build GRCh38
rs773867951 Chr.11: 34888427-34888427 on Build GRCh38
rs745374302 Chr.11: 34888838-34888838 on Build GRCh38
rs751590859 Chr.11: 34888785-34888785 on Build GRCh38
rs779305490 Chr.11: 34888832-34888832 on Build GRCh38
rs770338613 Chr.11: 34888707-34888707 on Build GRCh38
rs553220215 Chr.11: 34888754-34888754 on Build GRCh38
rs115074182 Chr.11: 34888653-34888653 on Build GRCh38
rs755026205 Chr.11: 34888794-34888794 on Build GRCh38
rs922583021 Chr.11: 34888802-34888802 on Build GRCh38
rs779793182 Chr.11: 34888724-34888725 on Build GRCh38

View Full Product Details